Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44786386-44786625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST4
Entrez Gene ID
266675
Gene Name
bestrophin 4
Gene Aliases
VMD2L2
UniGene
Hs.302513
Gene Chromosome Location
Chr.1: 44781836-44792828 on Build GRCh38
Transcripts
NM_153274.2 XM_017001020.1 XM_017001021.1 XM_017001022.1 XM_017001023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
398124 c.424T>G p.S142A Chr.1: 44786520-44786520 on Build GRCh38
4333364 c.367C>G p.L123V Chr.1: 44786577-44786577 on Build GRCh38
5800284 c.332C>T p.S111L Chr.1: 44786612-44786612 on Build GRCh38
6835145 c.415G>T p.V139L Chr.1: 44786529-44786529 on Build GRCh38
3944121 c.359G>C p.R120P Chr.1: 44786585-44786585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775267719 Chr.1: 44786535-44786535 on Build GRCh38
rs993887118 Chr.1: 44786599-44786599 on Build GRCh38
rs747323801 Chr.1: 44786448-44786448 on Build GRCh38
rs902516345 Chr.1: 44786418-44786418 on Build GRCh38
rs770329575 Chr.1: 44786595-44786595 on Build GRCh38
rs376337015 Chr.1: 44786485-44786485 on Build GRCh38
rs41306591 Chr.1: 44786487-44786487 on Build GRCh38
rs755167044 Chr.1: 44786459-44786459 on Build GRCh38
rs911545103 Chr.1: 44786577-44786577 on Build GRCh38
rs768295715 Chr.1: 44786500-44786501 on Build GRCh38
rs758926133 Chr.1: 44786564-44786564 on Build GRCh38
rs774303743 Chr.1: 44786499-44786499 on Build GRCh38
rs373896908 Chr.1: 44786590-44786590 on Build GRCh38
rs969873714 Chr.1: 44786494-44786494 on Build GRCh38
rs767215949 Chr.1: 44786519-44786519 on Build GRCh38
rs999901542 Chr.1: 44786458-44786458 on Build GRCh38
rs77461644 Chr.1: 44786473-44786473 on Build GRCh38
rs888403850 Chr.1: 44786492-44786492 on Build GRCh38
rs966216849 Chr.1: 44786589-44786589 on Build GRCh38
rs558041735 Chr.1: 44786554-44786554 on Build GRCh38
rs879474294 Chr.1: 44786522-44786522 on Build GRCh38
rs770943917 Chr.1: 44786488-44786488 on Build GRCh38
rs575564694 Chr.1: 44786484-44786484 on Build GRCh38
rs986380064 Chr.1: 44786512-44786528 on Build GRCh38
rs769966232 Chr.1: 44786479-44786479 on Build GRCh38
rs760217991 Chr.1: 44786536-44786536 on Build GRCh38
rs766811788 Chr.1: 44786585-44786585 on Build GRCh38
rs983497502 Chr.1: 44786551-44786551 on Build GRCh38
rs1027951216 Chr.1: 44786527-44786527 on Build GRCh38
rs753302001 Chr.1: 44786548-44786548 on Build GRCh38
rs758869193 Chr.1: 44786420-44786420 on Build GRCh38
rs759340108 Chr.1: 44786508-44786508 on Build GRCh38
rs780423944 Chr.1: 44786438-44786438 on Build GRCh38
rs781408099 Chr.1: 44786464-44786464 on Build GRCh38
rs906259275 Chr.1: 44786467-44786467 on Build GRCh38
rs1005476462 Chr.1: 44786541-44786541 on Build GRCh38

View Full Product Details