Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74591833-74592106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf77
Entrez Gene ID
146723
Gene Name
chromosome 17 open reading frame 77
Gene Aliases
-
UniGene
Hs.350775
Gene Chromosome Location
Chr.17: 74584679-74594209 on Build GRCh38
Transcripts
NM_001302809.1 NM_152460.2 NM_001115152.1

Gene Symbol
CD300LD
Entrez Gene ID
100131439
Gene Name
CD300 molecule like family member d
Gene Aliases
CD300D, CLM-4, CMRF35-A4, CMRF35A4
UniGene
Hs.531099
Gene Chromosome Location
Chr.17: 74579972-74592231 on Build GRCh38
Transcripts
NM_001302809.1 NM_152460.2 NM_001115152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4941600 c.50A>G p.N17S Chr.17: 74592096-74592096 on Build GRCh38
4000291 c.32C>T p.T11I Chr.17: 74592078-74592078 on Build GRCh38
3356953 c.57G>A p.A19A Chr.17: 74592103-74592103 on Build GRCh38
5527589 c.43C>T p.P15S Chr.17: 74592089-74592089 on Build GRCh38
3937552 c.15G>A p.A5A Chr.17: 74592061-74592061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752879258 Chr.17: 74592020-74592020 on Build GRCh38
rs377248113 Chr.17: 74592060-74592060 on Build GRCh38
rs749397825 Chr.17: 74591911-74591911 on Build GRCh38
rs749425211 Chr.17: 74592044-74592044 on Build GRCh38
rs1013355936 Chr.17: 74592001-74592001 on Build GRCh38
rs765083368 Chr.17: 74592083-74592083 on Build GRCh38
rs767345917 Chr.17: 74591983-74591983 on Build GRCh38
rs746562017 Chr.17: 74592072-74592072 on Build GRCh38
rs957998407 Chr.17: 74592048-74592048 on Build GRCh38
rs150016134 Chr.17: 74591891-74591891 on Build GRCh38
rs549246537 Chr.17: 74591951-74591951 on Build GRCh38
rs767308982 Chr.17: 74591993-74591993 on Build GRCh38
rs1046628415 Chr.17: 74591948-74591948 on Build GRCh38
rs1030768801 Chr.17: 74592032-74592032 on Build GRCh38
rs756236158 Chr.17: 74592031-74592031 on Build GRCh38
rs370110809 Chr.17: 74592079-74592079 on Build GRCh38
rs887274156 Chr.17: 74591906-74591906 on Build GRCh38
rs775955873 Chr.17: 74592062-74592062 on Build GRCh38
rs112399930 Chr.17: 74592074-74592074 on Build GRCh38
rs569459359 Chr.17: 74591963-74591963 on Build GRCh38
rs779082544 Chr.17: 74592055-74592055 on Build GRCh38
rs771343711 Chr.17: 74592047-74592047 on Build GRCh38
rs139236578 Chr.17: 74591865-74591865 on Build GRCh38
rs68124408 Chr.17: 74592078-74592078 on Build GRCh38
rs778091698 Chr.17: 74592041-74592041 on Build GRCh38
rs773205274 Chr.17: 74592085-74592085 on Build GRCh38
rs746228815 Chr.17: 74592061-74592061 on Build GRCh38

View Full Product Details