Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35909995-35910226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPSL
Entrez Gene ID
133690
Gene Name
calcyphosine like
Gene Aliases
-
UniGene
Hs.55150
Gene Chromosome Location
Chr.5: 35904295-35938779 on Build GRCh38
Transcripts
NM_001042625.1 NM_144647.3 XM_006714444.3 XM_006714445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7237970 c.316C>A p.P106T Chr.5: 35910075-35910075 on Build GRCh38
309849 c.327C>A p.S109S Chr.5: 35910064-35910064 on Build GRCh38
420727 c.349A>T p.M117L Chr.5: 35910042-35910042 on Build GRCh38
5036408 c.348C>T p.I116I Chr.5: 35910043-35910043 on Build GRCh38
3615790 c.316-2A>T p.? Chr.5: 35910077-35910077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200956929 Chr.5: 35910114-35910114 on Build GRCh38
rs76988839 Chr.5: 35910166-35910166 on Build GRCh38
rs117483383 Chr.5: 35910064-35910064 on Build GRCh38
rs372581109 Chr.5: 35910161-35910161 on Build GRCh38
rs748338288 Chr.5: 35910092-35910092 on Build GRCh38
rs372265672 Chr.5: 35910152-35910152 on Build GRCh38
rs771405172 Chr.5: 35910101-35910101 on Build GRCh38
rs1032701006 Chr.5: 35910036-35910036 on Build GRCh38
rs946057717 Chr.5: 35910169-35910169 on Build GRCh38
rs750270903 Chr.5: 35910065-35910065 on Build GRCh38
rs762377720 Chr.5: 35910031-35910031 on Build GRCh38
rs138096862 Chr.5: 35910167-35910167 on Build GRCh38
rs372118277 Chr.5: 35910040-35910040 on Build GRCh38
rs1043311808 Chr.5: 35910182-35910182 on Build GRCh38
rs1026329917 Chr.5: 35910170-35910170 on Build GRCh38
rs766239268 Chr.5: 35910073-35910073 on Build GRCh38
rs753887158 Chr.5: 35910074-35910074 on Build GRCh38
rs539120636 Chr.5: 35910057-35910057 on Build GRCh38
rs951639923 Chr.5: 35910190-35910190 on Build GRCh38
rs778063988 Chr.5: 35910097-35910097 on Build GRCh38
rs1044848933 Chr.5: 35910138-35910138 on Build GRCh38
rs755007573 Chr.5: 35910075-35910075 on Build GRCh38
rs938742108 Chr.5: 35910164-35910164 on Build GRCh38
rs768033968 Chr.5: 35910122-35910122 on Build GRCh38
rs766095343 Chr.5: 35910110-35910110 on Build GRCh38
rs575166416 Chr.5: 35910186-35910186 on Build GRCh38
rs758650769 Chr.5: 35910096-35910096 on Build GRCh38
rs751398372 Chr.5: 35910123-35910123 on Build GRCh38
rs756027620 Chr.5: 35910070-35910070 on Build GRCh38
rs747389173 Chr.5: 35910098-35910098 on Build GRCh38
rs577006534 Chr.5: 35910025-35910025 on Build GRCh38
rs774665279 Chr.5: 35910086-35910088 on Build GRCh38
rs920537681 Chr.5: 35910103-35910103 on Build GRCh38
rs779107309 Chr.5: 35910085-35910085 on Build GRCh38
rs141951671 Chr.5: 35910033-35910033 on Build GRCh38
rs904752205 Chr.5: 35910194-35910194 on Build GRCh38
rs767464889 Chr.5: 35910189-35910189 on Build GRCh38

View Full Product Details