Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 109331634-109331893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPPA4
Entrez Gene ID
55211
Gene Name
developmental pluripotency associated 4
Gene Aliases
2410091M23Rik
UniGene
Hs.317659
Gene Chromosome Location
Chr.3: 109326141-109337572 on Build GRCh38
Transcripts
NM_018189.3 XM_011512954.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1036164 c.353A>G p.Y118C Chr.3: 109331771-109331771 on Build GRCh38
4112123 c.320A>G p.K107R Chr.3: 109331890-109331890 on Build GRCh38
2917328 c.368C>T p.A123V Chr.3: 109331756-109331756 on Build GRCh38
1036166 c.341A>C p.K114T Chr.3: 109331783-109331783 on Build GRCh38
6451083 c.341A>T p.K114I Chr.3: 109331783-109331783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779103904 Chr.3: 109331835-109331835 on Build GRCh38
rs376868290 Chr.3: 109331858-109331858 on Build GRCh38
rs763806953 Chr.3: 109331787-109331787 on Build GRCh38
rs140276272 Chr.3: 109331828-109331828 on Build GRCh38
rs201143622 Chr.3: 109331852-109331852 on Build GRCh38
rs776933550 Chr.3: 109331741-109331741 on Build GRCh38
rs906247357 Chr.3: 109331796-109331796 on Build GRCh38
rs1014464238 Chr.3: 109331822-109331822 on Build GRCh38
rs555722815 Chr.3: 109331713-109331713 on Build GRCh38
rs377670885 Chr.3: 109331849-109331849 on Build GRCh38
rs1010501753 Chr.3: 109331699-109331699 on Build GRCh38
rs776921693 Chr.3: 109331852-109331852 on Build GRCh38
rs778079833 Chr.3: 109331848-109331848 on Build GRCh38
rs747386277 Chr.3: 109331717-109331717 on Build GRCh38
rs921972988 Chr.3: 109331864-109331864 on Build GRCh38
rs774777605 Chr.3: 109331713-109331714 on Build GRCh38
rs767350561 Chr.3: 109331812-109331812 on Build GRCh38
rs150309278 Chr.3: 109331850-109331850 on Build GRCh38
rs1051940765 Chr.3: 109331662-109331662 on Build GRCh38
rs754948429 Chr.3: 109331698-109331698 on Build GRCh38
rs751355798 Chr.3: 109331684-109331684 on Build GRCh38
rs201748770 Chr.3: 109331712-109331712 on Build GRCh38
rs756945864 Chr.3: 109331810-109331810 on Build GRCh38
rs1038343502 Chr.3: 109331668-109331668 on Build GRCh38
rs1024158915 Chr.3: 109331826-109331826 on Build GRCh38
rs192264330 Chr.3: 109331786-109331786 on Build GRCh38
rs1046191676 Chr.3: 109331761-109331761 on Build GRCh38
rs781050306 Chr.3: 109331686-109331686 on Build GRCh38
rs373678901 Chr.3: 109331830-109331830 on Build GRCh38
rs757215134 Chr.3: 109331685-109331685 on Build GRCh38
rs773706919 Chr.3: 109331716-109331716 on Build GRCh38
rs750277627 Chr.3: 109331823-109331823 on Build GRCh38
rs748304448 Chr.3: 109331838-109331838 on Build GRCh38
rs745833466 Chr.3: 109331694-109331694 on Build GRCh38
rs771337473 Chr.3: 109331731-109331731 on Build GRCh38
rs751357374 Chr.3: 109331798-109331798 on Build GRCh38
rs932103871 Chr.3: 109331661-109331662 on Build GRCh38
rs947677444 Chr.3: 109331748-109331748 on Build GRCh38
rs763858529 Chr.3: 109331683-109331683 on Build GRCh38
rs62276083 Chr.3: 109331843-109331843 on Build GRCh38
rs1034052539 Chr.3: 109331794-109331794 on Build GRCh38
rs775033673 Chr.3: 109331781-109331781 on Build GRCh38
rs958454027 Chr.3: 109331805-109331805 on Build GRCh38
rs760118460 Chr.3: 109331749-109331749 on Build GRCh38
rs368568568 Chr.3: 109331706-109331706 on Build GRCh38
rs764680501 Chr.3: 109331765-109331765 on Build GRCh38

View Full Product Details