Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70204086-70204355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGAT2L6
Entrez Gene ID
347516
Gene Name
diacylglycerol O-acyltransferase 2 like 6
Gene Aliases
DC3
UniGene
Hs.454355
Gene Chromosome Location
Chr.X: 70177483-70205703 on Build GRCh38
Transcripts
NM_198512.2

SNP Detail

SNP ID Chromosome Location
rs780650389 Chr.X: 70204309-70204309 on Build GRCh38
rs1052115991 Chr.X: 70204186-70204186 on Build GRCh38
rs933289355 Chr.X: 70204152-70204152 on Build GRCh38
rs752625850 Chr.X: 70204262-70204262 on Build GRCh38
rs372808364 Chr.X: 70204278-70204278 on Build GRCh38
rs761465643 Chr.X: 70204195-70204195 on Build GRCh38
rs746269242 Chr.X: 70204277-70204277 on Build GRCh38
rs939111923 Chr.X: 70204302-70204302 on Build GRCh38
rs756557853 Chr.X: 70204298-70204298 on Build GRCh38
rs372003647 Chr.X: 70204311-70204311 on Build GRCh38
rs756145952 Chr.X: 70204293-70204294 on Build GRCh38
rs764395247 Chr.X: 70204259-70204259 on Build GRCh38
rs183391954 Chr.X: 70204258-70204258 on Build GRCh38
rs781376047 Chr.X: 70204263-70204263 on Build GRCh38
rs772490998 Chr.X: 70204320-70204320 on Build GRCh38

View Full Product Details