Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143561249-143561516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSDMD
Entrez Gene ID
79792
Gene Name
gasdermin D
Gene Aliases
DF5L, DFNA5L, FKSG10, GSDMDC1
UniGene
Hs.118983
Gene Chromosome Location
Chr.8: 143553387-143563062 on Build GRCh38
Transcripts
NM_001166237.1 NM_024736.6 XM_011517301.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2868222 c.730G>A p.A244T Chr.8: 143561417-143561417 on Build GRCh38
3698872 c.685G>A p.V229I Chr.8: 143561372-143561372 on Build GRCh38
1132824 c.684C>T p.D228D Chr.8: 143561371-143561371 on Build GRCh38
4408957 c.736+10G>A p.? Chr.8: 143561433-143561433 on Build GRCh38
361312 c.729C>T p.P243P Chr.8: 143561416-143561416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202183230 Chr.8: 143561438-143561438 on Build GRCh38
rs202104158 Chr.8: 143561358-143561358 on Build GRCh38
rs140608348 Chr.8: 143561405-143561405 on Build GRCh38
rs577037853 Chr.8: 143561372-143561372 on Build GRCh38
rs758975277 Chr.8: 143561418-143561418 on Build GRCh38
rs746090558 Chr.8: 143561442-143561442 on Build GRCh38
rs138833870 Chr.8: 143561385-143561385 on Build GRCh38
rs757337585 Chr.8: 143561435-143561435 on Build GRCh38
rs746544177 Chr.8: 143561467-143561467 on Build GRCh38
rs759272374 Chr.8: 143561472-143561472 on Build GRCh38
rs900902485 Chr.8: 143561337-143561337 on Build GRCh38
rs754178997 Chr.8: 143561338-143561338 on Build GRCh38
rs747041959 Chr.8: 143561362-143561362 on Build GRCh38
rs1044354145 Chr.8: 143561272-143561272 on Build GRCh38
rs1042560120 Chr.8: 143561320-143561320 on Build GRCh38
rs370326863 Chr.8: 143561416-143561416 on Build GRCh38
rs766017133 Chr.8: 143561330-143561330 on Build GRCh38
rs760041290 Chr.8: 143561415-143561415 on Build GRCh38
rs372705443 Chr.8: 143561443-143561443 on Build GRCh38
rs2290414 Chr.8: 143561433-143561433 on Build GRCh38
rs747900168 Chr.8: 143561450-143561450 on Build GRCh38
rs147751545 Chr.8: 143561446-143561446 on Build GRCh38
rs771087871 Chr.8: 143561364-143561364 on Build GRCh38
rs143242888 Chr.8: 143561378-143561378 on Build GRCh38
rs1022588537 Chr.8: 143561268-143561268 on Build GRCh38
rs893670390 Chr.8: 143561311-143561311 on Build GRCh38
rs199855694 Chr.8: 143561356-143561356 on Build GRCh38
rs776320321 Chr.8: 143561411-143561411 on Build GRCh38
rs939034007 Chr.8: 143561420-143561420 on Build GRCh38
rs937897918 Chr.8: 143561303-143561303 on Build GRCh38
rs772021570 Chr.8: 143561456-143561456 on Build GRCh38
rs370341026 Chr.8: 143561357-143561357 on Build GRCh38
rs755351885 Chr.8: 143561343-143561343 on Build GRCh38
rs779607714 Chr.8: 143561345-143561345 on Build GRCh38
rs771682018 Chr.8: 143561313-143561313 on Build GRCh38
rs767128474 Chr.8: 143561419-143561419 on Build GRCh38
rs770475103 Chr.8: 143561470-143561470 on Build GRCh38
rs753049247 Chr.8: 143561417-143561417 on Build GRCh38
rs764912576 Chr.8: 143561475-143561475 on Build GRCh38
rs781426111 Chr.8: 143561367-143561367 on Build GRCh38
rs552794891 Chr.8: 143561386-143561386 on Build GRCh38
rs375513977 Chr.8: 143561351-143561351 on Build GRCh38
rs368469297 Chr.8: 143561457-143561457 on Build GRCh38
rs574516326 Chr.8: 143561392-143561392 on Build GRCh38
rs370809058 Chr.8: 143561308-143561308 on Build GRCh38
rs541556931 Chr.8: 143561393-143561393 on Build GRCh38
rs201583128 Chr.8: 143561371-143561371 on Build GRCh38
rs905134220 Chr.8: 143561290-143561290 on Build GRCh38

View Full Product Details