Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102793446-102793693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDELC1
Entrez Gene ID
79070
Gene Name
KDEL motif containing 1
Gene Aliases
EP58, ERp58, KDEL1
UniGene
Hs.408629
Gene Chromosome Location
Chr.13: 102784281-102799054 on Build GRCh38
Transcripts
NM_001318732.1 NM_024089.2 XM_005254075.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1300008 c.555G>A p.Q185Q Chr.13: 102793640-102793640 on Build GRCh38
2048327 c.545G>C p.R182T Chr.13: 102793650-102793650 on Build GRCh38
2048328 c.512A>G p.D171G Chr.13: 102793683-102793683 on Build GRCh38
5362266 c.554A>G p.Q185R Chr.13: 102793641-102793641 on Build GRCh38
4045774 c.593A>C p.K198T Chr.13: 102793602-102793602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551617190 Chr.13: 102793556-102793556 on Build GRCh38
rs958855856 Chr.13: 102793649-102793649 on Build GRCh38
rs774669534 Chr.13: 102793619-102793619 on Build GRCh38
rs768906813 Chr.13: 102793570-102793573 on Build GRCh38
rs764425823 Chr.13: 102793613-102793613 on Build GRCh38
rs1026713186 Chr.13: 102793563-102793563 on Build GRCh38
rs750428332 Chr.13: 102793627-102793627 on Build GRCh38
rs747765386 Chr.13: 102793666-102793666 on Build GRCh38
rs958328275 Chr.13: 102793502-102793502 on Build GRCh38
rs759761193 Chr.13: 102793477-102793477 on Build GRCh38
rs766268960 Chr.13: 102793635-102793635 on Build GRCh38
rs144061173 Chr.13: 102793640-102793640 on Build GRCh38
rs528476532 Chr.13: 102793525-102793525 on Build GRCh38
rs373431763 Chr.13: 102793563-102793564 on Build GRCh38
rs763342305 Chr.13: 102793579-102793579 on Build GRCh38
rs556925485 Chr.13: 102793555-102793556 on Build GRCh38
rs757975884 Chr.13: 102793671-102793671 on Build GRCh38
rs775715783 Chr.13: 102793553-102793553 on Build GRCh38
rs755997491 Chr.13: 102793632-102793632 on Build GRCh38
rs912406584 Chr.13: 102793651-102793651 on Build GRCh38
rs951122362 Chr.13: 102793591-102793591 on Build GRCh38
rs767467151 Chr.13: 102793624-102793624 on Build GRCh38
rs369648880 Chr.13: 102793647-102793647 on Build GRCh38
rs367564202 Chr.13: 102793509-102793509 on Build GRCh38
rs778566690 Chr.13: 102793655-102793655 on Build GRCh38
rs776994119 Chr.13: 102793581-102793582 on Build GRCh38
rs576494656 Chr.13: 102793470-102793470 on Build GRCh38

View Full Product Details