Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14511119-14511392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLBD1
Entrez Gene ID
79887
Gene Name
phospholipase B domain containing 1
Gene Aliases
-
UniGene
Hs.131933
Gene Chromosome Location
Chr.12: 14503663-14567857 on Build GRCh38
Transcripts
NM_024829.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4040555 c.1186G>T p.G396* Chr.12: 14511260-14511260 on Build GRCh38
4040556 c.1148A>G p.Y383C Chr.12: 14511298-14511298 on Build GRCh38
5962045 c.1109A>G p.D370G Chr.12: 14511337-14511337 on Build GRCh38
5007253 c.1075G>C p.D359H Chr.12: 14511371-14511371 on Build GRCh38
5051478 c.1134G>A p.E378E Chr.12: 14511312-14511312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028038363 Chr.12: 14511280-14511280 on Build GRCh38
rs1045683007 Chr.12: 14511224-14511224 on Build GRCh38
rs548006259 Chr.12: 14511169-14511169 on Build GRCh38
rs749413227 Chr.12: 14511153-14511153 on Build GRCh38
rs773524183 Chr.12: 14511236-14511236 on Build GRCh38
rs553944681 Chr.12: 14511305-14511305 on Build GRCh38
rs149403554 Chr.12: 14511267-14511267 on Build GRCh38
rs2287541 Chr.12: 14511316-14511316 on Build GRCh38
rs755274209 Chr.12: 14511343-14511343 on Build GRCh38
rs375514909 Chr.12: 14511179-14511179 on Build GRCh38
rs1016572226 Chr.12: 14511266-14511266 on Build GRCh38
rs758852712 Chr.12: 14511351-14511351 on Build GRCh38
rs2287540 Chr.12: 14511245-14511245 on Build GRCh38
rs529551905 Chr.12: 14511167-14511167 on Build GRCh38
rs772605033 Chr.12: 14511234-14511234 on Build GRCh38
rs753989795 Chr.12: 14511324-14511324 on Build GRCh38
rs948523896 Chr.12: 14511192-14511192 on Build GRCh38
rs767597101 Chr.12: 14511300-14511300 on Build GRCh38
rs11550115 Chr.12: 14511299-14511299 on Build GRCh38
rs894310856 Chr.12: 14511158-14511158 on Build GRCh38
rs533474389 Chr.12: 14511190-14511190 on Build GRCh38
rs367807814 Chr.12: 14511302-14511302 on Build GRCh38
rs1019906569 Chr.12: 14511354-14511354 on Build GRCh38
rs566093824 Chr.12: 14511171-14511171 on Build GRCh38
rs183359266 Chr.12: 14511240-14511240 on Build GRCh38
rs568013423 Chr.12: 14511265-14511265 on Build GRCh38
rs996457638 Chr.12: 14511297-14511297 on Build GRCh38
rs766561209 Chr.12: 14511320-14511320 on Build GRCh38
rs201667097 Chr.12: 14511244-14511244 on Build GRCh38
rs774884905 Chr.12: 14511298-14511299 on Build GRCh38
rs778824843 Chr.12: 14511347-14511347 on Build GRCh38
rs778264458 Chr.12: 14511356-14511356 on Build GRCh38
rs202168556 Chr.12: 14511243-14511243 on Build GRCh38

View Full Product Details