Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196318123-196318368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCTEX1D2
Entrez Gene ID
255758
Gene Name
Tctex1 domain containing 2
Gene Aliases
-
UniGene
Hs.733180
Gene Chromosome Location
Chr.3: 196291219-196318294 on Build GRCh38
Transcripts
NM_152773.4 NR_046724.1 NR_121665.1 NR_037950.1

Gene Symbol
TM4SF19-AS1
Entrez Gene ID
100874214
Gene Name
TM4SF19 antisense RNA 1
Gene Aliases
-
UniGene
Hs.672105
Gene Chromosome Location
Chr.3: 196318332-196325570 on Build GRCh38
Transcripts
NM_152773.4 NR_046724.1 NR_121665.1 NR_037950.1

Gene Symbol
TM4SF19-TCTEX1D2
Entrez Gene ID
100534611
Gene Name
TM4SF19-TCTEX1D2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.733180
Gene Chromosome Location
Chr.3: 196316085-196338420 on Build GRCh38
Transcripts
NM_152773.4 NR_046724.1 NR_121665.1 NR_037950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1617405 c.9G>C p.T3T Chr.3: 196318144-196318144 on Build GRCh38
1228802 c.4G>A p.A2T Chr.3: 196318149-196318149 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1047712469 Chr.3: 196318307-196318307 on Build GRCh38
rs929920078 Chr.3: 196318302-196318302 on Build GRCh38
rs917208597 Chr.3: 196318309-196318309 on Build GRCh38
rs753928443 Chr.3: 196318202-196318202 on Build GRCh38
rs752258974 Chr.3: 196318160-196318160 on Build GRCh38
rs1000357265 Chr.3: 196318352-196318352 on Build GRCh38
rs955109494 Chr.3: 196318200-196318200 on Build GRCh38
rs1029397534 Chr.3: 196318197-196318197 on Build GRCh38
rs769764856 Chr.3: 196318291-196318291 on Build GRCh38
rs766929757 Chr.3: 196318155-196318156 on Build GRCh38
rs553817724 Chr.3: 196318206-196318206 on Build GRCh38
rs202097551 Chr.3: 196318142-196318142 on Build GRCh38
rs910978262 Chr.3: 196318293-196318293 on Build GRCh38
rs550944554 Chr.3: 196318171-196318171 on Build GRCh38
rs929303126 Chr.3: 196318196-196318196 on Build GRCh38
rs779969432 Chr.3: 196318163-196318163 on Build GRCh38
rs746460801 Chr.3: 196318164-196318164 on Build GRCh38
rs187790075 Chr.3: 196318245-196318245 on Build GRCh38
rs955137717 Chr.3: 196318283-196318283 on Build GRCh38
rs577678348 Chr.3: 196318294-196318294 on Build GRCh38
rs973912396 Chr.3: 196318295-196318295 on Build GRCh38
rs1030456640 Chr.3: 196318352-196318352 on Build GRCh38
rs182319594 Chr.3: 196318185-196318185 on Build GRCh38
rs768191137 Chr.3: 196318165-196318165 on Build GRCh38
rs1045121920 Chr.3: 196318298-196318298 on Build GRCh38
rs766606492 Chr.3: 196318234-196318234 on Build GRCh38
rs759988226 Chr.3: 196318182-196318182 on Build GRCh38
rs370554455 Chr.3: 196318300-196318300 on Build GRCh38
rs375451728 Chr.3: 196318183-196318183 on Build GRCh38
rs573467978 Chr.3: 196318203-196318203 on Build GRCh38
rs985590808 Chr.3: 196318287-196318287 on Build GRCh38
rs979774051 Chr.3: 196318221-196318221 on Build GRCh38
rs906005453 Chr.3: 196318225-196318231 on Build GRCh38
rs1023060734 Chr.3: 196318223-196318223 on Build GRCh38
rs753282035 Chr.3: 196318190-196318190 on Build GRCh38
rs997664846 Chr.3: 196318349-196318349 on Build GRCh38
rs911018497 Chr.3: 196318183-196318183 on Build GRCh38
rs948127743 Chr.3: 196318292-196318292 on Build GRCh38
rs878887460 Chr.3: 196318262-196318262 on Build GRCh38
rs897087360 Chr.3: 196318194-196318194 on Build GRCh38
rs942019006 Chr.3: 196318342-196318342 on Build GRCh38
rs542186386 Chr.3: 196318345-196318345 on Build GRCh38
rs759573304 Chr.3: 196318148-196318152 on Build GRCh38
rs751794204 Chr.3: 196318304-196318304 on Build GRCh38
rs1044833143 Chr.3: 196318328-196318328 on Build GRCh38
rs544822399 Chr.3: 196318321-196318321 on Build GRCh38
rs956309122 Chr.3: 196318250-196318250 on Build GRCh38
rs898070744 Chr.3: 196318346-196318346 on Build GRCh38
rs573662432 Chr.3: 196318215-196318215 on Build GRCh38
rs200389535 Chr.3: 196318181-196318181 on Build GRCh38
rs771939405 Chr.3: 196318160-196318160 on Build GRCh38
rs868491776 Chr.3: 196318290-196318290 on Build GRCh38
rs935179370 Chr.3: 196318239-196318239 on Build GRCh38
rs769629292 Chr.3: 196318173-196318173 on Build GRCh38
rs538613478 Chr.3: 196318166-196318166 on Build GRCh38
rs918045376 Chr.3: 196318275-196318275 on Build GRCh38
rs1018363075 Chr.3: 196318214-196318214 on Build GRCh38
rs988472625 Chr.3: 196318257-196318257 on Build GRCh38
rs555279962 Chr.3: 196318192-196318192 on Build GRCh38
rs764201336 Chr.3: 196318199-196318199 on Build GRCh38
rs145146700 Chr.3: 196318143-196318143 on Build GRCh38
rs745720115 Chr.3: 196318158-196318158 on Build GRCh38
rs898018275 Chr.3: 196318149-196318149 on Build GRCh38
rs950096525 Chr.3: 196318316-196318316 on Build GRCh38

View Full Product Details