Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6456413-6456638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A23
Entrez Gene ID
79085
Gene Name
solute carrier family 25 member 23
Gene Aliases
APC2, MCSC2, SCaMC-3
UniGene
Hs.356231
Gene Chromosome Location
Chr.19: 6436081-6460221 on Build GRCh38
Transcripts
NM_024103.2 XM_011528274.1 XM_011528275.1 XM_011528276.1 XM_011528277.1 XM_011528278.1 XM_011528279.1 XM_011528280.1 XM_011528281.1 XM_011528282.1 XM_011528283.1 XM_011528284.1 XM_011528285.1 XM_017027285.1 XM_017027286.1 XM_017027287.1 XM_017027288.1 XM_017027289.1 XM_017027290.1 XM_017027291.1 XM_017027292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6780588 c.437C>T p.S146L Chr.19: 6456466-6456466 on Build GRCh38
1680552 c.412T>G p.W138G Chr.19: 6456491-6456491 on Build GRCh38
5417572 c.483G>A p.T161T Chr.19: 6456420-6456420 on Build GRCh38
4726820 c.384C>T p.D128D Chr.19: 6456519-6456519 on Build GRCh38
5417571 c.483G>A p.T161T Chr.19: 6456420-6456420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79304508 Chr.19: 6456486-6456486 on Build GRCh38
rs774049176 Chr.19: 6456466-6456466 on Build GRCh38
rs779208068 Chr.19: 6456439-6456439 on Build GRCh38
rs541607862 Chr.19: 6456594-6456594 on Build GRCh38
rs764467645 Chr.19: 6456485-6456485 on Build GRCh38
rs775332648 Chr.19: 6456538-6456538 on Build GRCh38
rs745955363 Chr.19: 6456441-6456441 on Build GRCh38
rs149525556 Chr.19: 6456447-6456447 on Build GRCh38
rs372420203 Chr.19: 6456476-6456476 on Build GRCh38
rs747024230 Chr.19: 6456520-6456520 on Build GRCh38
rs749918850 Chr.19: 6456533-6456534 on Build GRCh38
rs571110750 Chr.19: 6456468-6456468 on Build GRCh38
rs886847425 Chr.19: 6456512-6456512 on Build GRCh38
rs775952069 Chr.19: 6456453-6456453 on Build GRCh38
rs200758261 Chr.19: 6456547-6456547 on Build GRCh38
rs1046835568 Chr.19: 6456502-6456502 on Build GRCh38
rs756085750 Chr.19: 6456521-6456521 on Build GRCh38
rs750738525 Chr.19: 6456492-6456492 on Build GRCh38
rs757635721 Chr.19: 6456487-6456487 on Build GRCh38
rs963832365 Chr.19: 6456539-6456539 on Build GRCh38
rs777332654 Chr.19: 6456548-6456548 on Build GRCh38
rs767094850 Chr.19: 6456471-6456471 on Build GRCh38
rs772376419 Chr.19: 6456503-6456503 on Build GRCh38
rs773093918 Chr.19: 6456457-6456457 on Build GRCh38
rs376917916 Chr.19: 6456549-6456549 on Build GRCh38
rs765680503 Chr.19: 6456490-6456490 on Build GRCh38
rs765733338 Chr.19: 6456553-6456553 on Build GRCh38
rs367895517 Chr.19: 6456523-6456523 on Build GRCh38
rs769573506 Chr.19: 6456456-6456456 on Build GRCh38
rs555434256 Chr.19: 6456608-6456608 on Build GRCh38
rs939062045 Chr.19: 6456488-6456488 on Build GRCh38
rs761267155 Chr.19: 6456482-6456482 on Build GRCh38
rs368298555 Chr.19: 6456519-6456519 on Build GRCh38
rs927471010 Chr.19: 6456565-6456565 on Build GRCh38
rs375466548 Chr.19: 6456474-6456474 on Build GRCh38
rs772333416 Chr.19: 6456444-6456444 on Build GRCh38
rs10418327 Chr.19: 6456592-6456592 on Build GRCh38
rs758659098 Chr.19: 6456518-6456518 on Build GRCh38
rs750771685 Chr.19: 6456576-6456576 on Build GRCh38
rs755712184 Chr.19: 6456534-6456534 on Build GRCh38
rs779426800 Chr.19: 6456581-6456581 on Build GRCh38
rs369529733 Chr.19: 6456563-6456563 on Build GRCh38
rs770586281 Chr.19: 6456465-6456465 on Build GRCh38
rs372136336 Chr.19: 6456534-6456534 on Build GRCh38
rs779989407 Chr.19: 6456446-6456446 on Build GRCh38
rs745569166 Chr.19: 6456535-6456535 on Build GRCh38
rs762672435 Chr.19: 6456462-6456462 on Build GRCh38
rs201973405 Chr.19: 6456470-6456470 on Build GRCh38

View Full Product Details