Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26179051-26179276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMA8
Entrez Gene ID
143471
Gene Name
proteasome subunit alpha 8
Gene Aliases
PSMA7L
UniGene
Hs.464813
Gene Chromosome Location
Chr.18: 26133847-26193355 on Build GRCh38
Transcripts
NM_001025096.1 NM_001025097.1 NM_001308188.1 NM_144662.2 XM_011525818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1388145 c.632_633insA p.N213fs*3 Chr.18: 26179084-26179085 on Build GRCh38
3672656 c.678+2T>C p.? Chr.18: 26179132-26179132 on Build GRCh38
4071625 c.661A>C p.R221R Chr.18: 26179113-26179113 on Build GRCh38
4438723 c.675G>C p.L225F Chr.18: 26179127-26179127 on Build GRCh38
3524674 c.617T>C p.V206A Chr.18: 26179069-26179069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750520056 Chr.18: 26179144-26179144 on Build GRCh38
rs760737822 Chr.18: 26179151-26179151 on Build GRCh38
rs1000724765 Chr.18: 26179231-26179231 on Build GRCh38
rs922817805 Chr.18: 26179218-26179218 on Build GRCh38
rs768853808 Chr.18: 26179101-26179101 on Build GRCh38
rs747333158 Chr.18: 26179211-26179211 on Build GRCh38
rs967520020 Chr.18: 26179130-26179130 on Build GRCh38
rs758374723 Chr.18: 26179158-26179158 on Build GRCh38
rs762061479 Chr.18: 26179138-26179138 on Build GRCh38
rs778313261 Chr.18: 26179150-26179150 on Build GRCh38
rs979235144 Chr.18: 26179131-26179131 on Build GRCh38
rs142265503 Chr.18: 26179081-26179081 on Build GRCh38
rs750609770 Chr.18: 26179150-26179150 on Build GRCh38
rs756843945 Chr.18: 26179170-26179170 on Build GRCh38
rs765193893 Chr.18: 26179154-26179154 on Build GRCh38
rs763355221 Chr.18: 26179100-26179100 on Build GRCh38
rs751357277 Chr.18: 26179164-26179164 on Build GRCh38
rs767869320 Chr.18: 26179141-26179141 on Build GRCh38
rs913731108 Chr.18: 26179095-26179095 on Build GRCh38
rs752622201 Chr.18: 26179157-26179157 on Build GRCh38
rs777500957 Chr.18: 26179163-26179163 on Build GRCh38
rs903748871 Chr.18: 26179227-26179227 on Build GRCh38
rs1039937527 Chr.18: 26179216-26179216 on Build GRCh38
rs114468957 Chr.18: 26179195-26179195 on Build GRCh38
rs377451012 Chr.18: 26179137-26179137 on Build GRCh38
rs774889972 Chr.18: 26179109-26179109 on Build GRCh38
rs17852261 Chr.18: 26179087-26179087 on Build GRCh38

View Full Product Details