Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84496011-84496272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPF1
Entrez Gene ID
80135
Gene Name
ribosome production factor 1 homolog
Gene Aliases
BXDC5
UniGene
Hs.481202
Gene Chromosome Location
Chr.1: 84479237-84498350 on Build GRCh38
Transcripts
NM_025065.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4010227 c.881+2T>C p.? Chr.1: 84496065-84496065 on Build GRCh38
7323647 c.891C>G p.F297L Chr.1: 84496253-84496253 on Build GRCh38
4882401 c.867C>A p.F289L Chr.1: 84496049-84496049 on Build GRCh38
912575 c.903G>T p.K301N Chr.1: 84496265-84496265 on Build GRCh38
3866160 c.853C>A p.R285R Chr.1: 84496035-84496035 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886092144 Chr.1: 84496072-84496072 on Build GRCh38
rs1011193116 Chr.1: 84496137-84496137 on Build GRCh38
rs748978580 Chr.1: 84496236-84496236 on Build GRCh38
rs565891899 Chr.1: 84496175-84496175 on Build GRCh38
rs748262403 Chr.1: 84496206-84496206 on Build GRCh38
rs199978767 Chr.1: 84496047-84496047 on Build GRCh38
rs529910681 Chr.1: 84496044-84496044 on Build GRCh38
rs72948448 Chr.1: 84496190-84496190 on Build GRCh38
rs753353389 Chr.1: 84496226-84496227 on Build GRCh38
rs1045337946 Chr.1: 84496232-84496232 on Build GRCh38
rs771809205 Chr.1: 84496238-84496238 on Build GRCh38
rs1003269145 Chr.1: 84496076-84496076 on Build GRCh38
rs959266355 Chr.1: 84496151-84496151 on Build GRCh38
rs918477746 Chr.1: 84496227-84496227 on Build GRCh38
rs774098385 Chr.1: 84496239-84496239 on Build GRCh38
rs978066096 Chr.1: 84496153-84496153 on Build GRCh38
rs895300290 Chr.1: 84496124-84496124 on Build GRCh38
rs773973055 Chr.1: 84496150-84496150 on Build GRCh38
rs1012457067 Chr.1: 84496157-84496157 on Build GRCh38
rs560629472 Chr.1: 84496205-84496205 on Build GRCh38
rs777811366 Chr.1: 84496210-84496210 on Build GRCh38
rs755237626 Chr.1: 84496202-84496202 on Build GRCh38
rs745848981 Chr.1: 84496211-84496211 on Build GRCh38
rs966724950 Chr.1: 84496147-84496147 on Build GRCh38
rs761366476 Chr.1: 84496037-84496037 on Build GRCh38
rs1033812960 Chr.1: 84496118-84496118 on Build GRCh38
rs768385168 Chr.1: 84496237-84496237 on Build GRCh38
rs368861828 Chr.1: 84496220-84496220 on Build GRCh38
rs760054903 Chr.1: 84496093-84496094 on Build GRCh38
rs774733749 Chr.1: 84496047-84496049 on Build GRCh38
rs756545807 Chr.1: 84496059-84496059 on Build GRCh38
rs1022542187 Chr.1: 84496143-84496143 on Build GRCh38
rs1029767464 Chr.1: 84496174-84496174 on Build GRCh38
rs756758387 Chr.1: 84496229-84496230 on Build GRCh38
rs941176137 Chr.1: 84496209-84496216 on Build GRCh38
rs774012681 Chr.1: 84496034-84496034 on Build GRCh38
rs767952587 Chr.1: 84496218-84496222 on Build GRCh38
rs369066394 Chr.1: 84496091-84496091 on Build GRCh38
rs758428600 Chr.1: 84496209-84496209 on Build GRCh38
rs985701882 Chr.1: 84496192-84496192 on Build GRCh38
rs769616392 Chr.1: 84496215-84496215 on Build GRCh38
rs911089843 Chr.1: 84496203-84496203 on Build GRCh38
rs754136089 Chr.1: 84496107-84496107 on Build GRCh38

View Full Product Details