Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161792173-161792398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF6
Entrez Gene ID
22926
Gene Name
activating transcription factor 6
Gene Aliases
ACHM7, ATF6A
UniGene
Hs.492740
Gene Chromosome Location
Chr.1: 161766244-161964070 on Build GRCh38
Transcripts
NM_007348.3 XM_006711224.1 XM_011509308.1 XM_011509309.1 XM_011509310.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6508081 c.590G>A p.S197N Chr.1: 161792229-161792229 on Build GRCh38
6606527 c.634A>G p.M212V Chr.1: 161792273-161792273 on Build GRCh38
5729276 c.652C>T p.Q218* Chr.1: 161792291-161792291 on Build GRCh38
898608 c.631C>T p.L211F Chr.1: 161792270-161792270 on Build GRCh38
3360442 c.663C>T p.I221I Chr.1: 161792302-161792302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747998336 Chr.1: 161792331-161792331 on Build GRCh38
rs914950753 Chr.1: 161792320-161792320 on Build GRCh38
rs779045495 Chr.1: 161792275-161792275 on Build GRCh38
rs376187541 Chr.1: 161792202-161792202 on Build GRCh38
rs572255096 Chr.1: 161792238-161792238 on Build GRCh38
rs749688488 Chr.1: 161792315-161792315 on Build GRCh38
rs750249727 Chr.1: 161792279-161792279 on Build GRCh38
rs764021639 Chr.1: 161792352-161792352 on Build GRCh38
rs370025619 Chr.1: 161792207-161792207 on Build GRCh38
rs138321248 Chr.1: 161792268-161792268 on Build GRCh38
rs540833998 Chr.1: 161792294-161792294 on Build GRCh38
rs374110554 Chr.1: 161792269-161792269 on Build GRCh38
rs774857491 Chr.1: 161792218-161792218 on Build GRCh38
rs769628164 Chr.1: 161792365-161792377 on Build GRCh38
rs368017110 Chr.1: 161792331-161792331 on Build GRCh38
rs759790425 Chr.1: 161792224-161792224 on Build GRCh38
rs767869777 Chr.1: 161792234-161792234 on Build GRCh38
rs112195242 Chr.1: 161792351-161792351 on Build GRCh38
rs778614001 Chr.1: 161792274-161792274 on Build GRCh38
rs768876285 Chr.1: 161792353-161792353 on Build GRCh38
rs757997210 Chr.1: 161792293-161792293 on Build GRCh38
rs764299597 Chr.1: 161792264-161792264 on Build GRCh38
rs766268875 Chr.1: 161792371-161792371 on Build GRCh38
rs760640737 Chr.1: 161792260-161792260 on Build GRCh38
rs924803205 Chr.1: 161792213-161792213 on Build GRCh38
rs146056942 Chr.1: 161792319-161792319 on Build GRCh38
rs771411815 Chr.1: 161792214-161792214 on Build GRCh38
rs780815661 Chr.1: 161792307-161792307 on Build GRCh38
rs768048617 Chr.1: 161792303-161792303 on Build GRCh38
rs369291362 Chr.1: 161792363-161792363 on Build GRCh38
rs758162439 Chr.1: 161792370-161792370 on Build GRCh38
rs759953675 Chr.1: 161792332-161792332 on Build GRCh38
rs746718839 Chr.1: 161792297-161792297 on Build GRCh38
rs115167829 Chr.1: 161792333-161792333 on Build GRCh38
rs138277547 Chr.1: 161792337-161792337 on Build GRCh38

View Full Product Details