Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23838217-23838452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERL3
Entrez Gene ID
91319
Gene Name
derlin 3
Gene Aliases
C22orf14, IZP6, LLN2, derlin-3
UniGene
Hs.593679
Gene Chromosome Location
Chr.22: 23834503-23839344 on Build GRCh38
Transcripts
NM_001002862.2 NM_001135751.1 NM_198440.3 XM_011530505.2 XM_011530506.2 XM_017029078.1 XM_017029079.1 XM_017029080.1 XM_017029081.1 XM_017029082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1751759 c.281C>G p.T94R Chr.22: 23838398-23838398 on Build GRCh38
4981825 c.263G>A p.G88D Chr.22: 23838416-23838416 on Build GRCh38
1203406 c.327+1G>A p.? Chr.22: 23838351-23838351 on Build GRCh38
5614860 c.291C>T p.F97F Chr.22: 23838388-23838388 on Build GRCh38
4103009 c.292G>A p.V98I Chr.22: 23838387-23838387 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371994369 Chr.22: 23838419-23838419 on Build GRCh38
rs967800614 Chr.22: 23838375-23838375 on Build GRCh38
rs759406442 Chr.22: 23838399-23838399 on Build GRCh38
rs755467434 Chr.22: 23838431-23838431 on Build GRCh38
rs201321388 Chr.22: 23838391-23838391 on Build GRCh38
rs773168556 Chr.22: 23838326-23838326 on Build GRCh38
rs774445766 Chr.22: 23838414-23838414 on Build GRCh38
rs912622007 Chr.22: 23838241-23838241 on Build GRCh38
rs562806647 Chr.22: 23838263-23838263 on Build GRCh38
rs867847111 Chr.22: 23838376-23838376 on Build GRCh38
rs759514404 Chr.22: 23838347-23838347 on Build GRCh38
rs141030731 Chr.22: 23838394-23838394 on Build GRCh38
rs750720698 Chr.22: 23838260-23838260 on Build GRCh38
rs770740955 Chr.22: 23838340-23838340 on Build GRCh38
rs751582211 Chr.22: 23838356-23838356 on Build GRCh38
rs147814749 Chr.22: 23838381-23838381 on Build GRCh38
rs766663143 Chr.22: 23838428-23838428 on Build GRCh38
rs756474104 Chr.22: 23838365-23838365 on Build GRCh38
rs754161997 Chr.22: 23838429-23838429 on Build GRCh38
rs760501541 Chr.22: 23838336-23838336 on Build GRCh38
rs773487659 Chr.22: 23838420-23838420 on Build GRCh38
rs1049111246 Chr.22: 23838369-23838369 on Build GRCh38
rs536282991 Chr.22: 23838365-23838365 on Build GRCh38
rs562256740 Chr.22: 23838361-23838361 on Build GRCh38
rs778566472 Chr.22: 23838377-23838377 on Build GRCh38
rs201924268 Chr.22: 23838397-23838397 on Build GRCh38
rs1049062603 Chr.22: 23838261-23838261 on Build GRCh38
rs143512856 Chr.22: 23838398-23838398 on Build GRCh38
rs750621124 Chr.22: 23838363-23838363 on Build GRCh38
rs747813250 Chr.22: 23838308-23838308 on Build GRCh38
rs576237830 Chr.22: 23838311-23838311 on Build GRCh38
rs117741234 Chr.22: 23838353-23838353 on Build GRCh38
rs6003907 Chr.22: 23838262-23838262 on Build GRCh38
rs768115854 Chr.22: 23838271-23838271 on Build GRCh38
rs1057018878 Chr.22: 23838306-23838306 on Build GRCh38
rs376592408 Chr.22: 23838423-23838423 on Build GRCh38
rs577144706 Chr.22: 23838254-23838254 on Build GRCh38
rs760439493 Chr.22: 23838426-23838427 on Build GRCh38
rs747617476 Chr.22: 23838379-23838379 on Build GRCh38
rs761929129 Chr.22: 23838418-23838418 on Build GRCh38
rs145966230 Chr.22: 23838370-23838370 on Build GRCh38
rs778469886 Chr.22: 23838278-23838278 on Build GRCh38
rs755452170 Chr.22: 23838372-23838372 on Build GRCh38
rs745603750 Chr.22: 23838367-23838367 on Build GRCh38
rs930610472 Chr.22: 23838255-23838255 on Build GRCh38
rs184851196 Chr.22: 23838401-23838401 on Build GRCh38

View Full Product Details