Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74844145-74844370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRRA1
Entrez Gene ID
143570
Gene Name
X-ray radiation resistance associated 1
Gene Aliases
-
UniGene
Hs.370145
Gene Chromosome Location
Chr.11: 74840910-74949187 on Build GRCh38
Transcripts
NM_001270380.1 NM_001270381.1 NM_182969.2 XM_005273765.2 XM_005273770.2 XM_011544755.1 XM_011544756.1 XM_011544757.1 XM_011544758.1 XM_011544759.1 XM_011544760.2 XM_011544761.1 XM_011544762.1 XM_011544763.1 XM_011544764.1 XM_011544765.1 XM_011544766.2 XM_011544767.1 XM_011544768.1 XM_017017221.1 XM_017017222.1 XM_017017223.1 XM_017017224.1 XM_017017225.1 XM_017017226.1 XM_017017227.1 XM_017017228.1 XM_017017229.1 XM_017017230.1 XM_017017231.1 XM_017017232.1 XM_017017233.1 XM_017017234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
690682 c.1987C>G p.L663V Chr.11: 74844200-74844200 on Build GRCh38
690683 c.2011G>C p.E671Q Chr.11: 74844176-74844176 on Build GRCh38
5767514 c.2013G>C p.E671D Chr.11: 74844174-74844174 on Build GRCh38
3700145 c.2019G>A p.R673R Chr.11: 74844168-74844168 on Build GRCh38
467414 c.1999T>C p.Y667H Chr.11: 74844188-74844188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763840100 Chr.11: 74844208-74844208 on Build GRCh38
rs759530967 Chr.11: 74844185-74844185 on Build GRCh38
rs767357091 Chr.11: 74844223-74844223 on Build GRCh38
rs570200367 Chr.11: 74844169-74844169 on Build GRCh38
rs779757323 Chr.11: 74844239-74844239 on Build GRCh38
rs549369299 Chr.11: 74844267-74844267 on Build GRCh38
rs76814368 Chr.11: 74844254-74844254 on Build GRCh38
rs771394885 Chr.11: 74844273-74844273 on Build GRCh38
rs763258273 Chr.11: 74844293-74844294 on Build GRCh38
rs750438556 Chr.11: 74844227-74844227 on Build GRCh38
rs776713627 Chr.11: 74844179-74844179 on Build GRCh38
rs373727157 Chr.11: 74844170-74844170 on Build GRCh38
rs764317281 Chr.11: 74844323-74844323 on Build GRCh38
rs774455989 Chr.11: 74844220-74844220 on Build GRCh38
rs535756655 Chr.11: 74844327-74844327 on Build GRCh38
rs199654451 Chr.11: 74844322-74844324 on Build GRCh38
rs1038772175 Chr.11: 74844341-74844341 on Build GRCh38
rs900224379 Chr.11: 74844344-74844344 on Build GRCh38
rs75226304 Chr.11: 74844333-74844333 on Build GRCh38
rs916445271 Chr.11: 74844217-74844217 on Build GRCh38
rs746438592 Chr.11: 74844283-74844283 on Build GRCh38
rs372286156 Chr.11: 74844282-74844282 on Build GRCh38
rs769834551 Chr.11: 74844288-74844288 on Build GRCh38
rs909072184 Chr.11: 74844328-74844328 on Build GRCh38
rs1046196823 Chr.11: 74844242-74844242 on Build GRCh38
rs940356769 Chr.11: 74844260-74844260 on Build GRCh38
rs775581934 Chr.11: 74844293-74844293 on Build GRCh38
rs370076583 Chr.11: 74844190-74844190 on Build GRCh38
rs996473821 Chr.11: 74844314-74844322 on Build GRCh38
rs771363741 Chr.11: 74844176-74844176 on Build GRCh38
rs766611957 Chr.11: 74844318-74844319 on Build GRCh38
rs375631338 Chr.11: 74844171-74844171 on Build GRCh38
rs771101702 Chr.11: 74844188-74844188 on Build GRCh38
rs763115211 Chr.11: 74844297-74844297 on Build GRCh38
rs751381819 Chr.11: 74844212-74844212 on Build GRCh38
rs928650889 Chr.11: 74844255-74844255 on Build GRCh38
rs755564067 Chr.11: 74844228-74844228 on Build GRCh38
rs775669740 Chr.11: 74844187-74844187 on Build GRCh38
rs747347205 Chr.11: 74844269-74844269 on Build GRCh38
rs773282100 Chr.11: 74844199-74844200 on Build GRCh38
rs774131976 Chr.11: 74844329-74844329 on Build GRCh38
rs765358616 Chr.11: 74844186-74844186 on Build GRCh38
rs757164192 Chr.11: 74844221-74844221 on Build GRCh38

View Full Product Details