Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27139210-27139455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H2BK
Entrez Gene ID
85236
Gene Name
histone cluster 1, H2bk
Gene Aliases
H2B/S, H2BFAiii, H2BFT, H2BK
UniGene
Hs.437275
Gene Chromosome Location
Chr.6: 27138293-27146858 on Build GRCh38
Transcripts
NM_080593.2 NM_003495.2

Gene Symbol
HIST1H4I
Entrez Gene ID
8294
Gene Name
histone cluster 1, H4i
Gene Aliases
H4/m, H4FM, H4M
UniGene
Hs.248172
Gene Chromosome Location
Chr.6: 27139309-27139678 on Build GRCh38
Transcripts
NM_080593.2 NM_003495.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4886805 c.11G>A p.R4H Chr.6: 27139319-27139319 on Build GRCh38
6173358 c.122G>T p.R41L Chr.6: 27139430-27139430 on Build GRCh38
4970404 c.10C>T p.R4C Chr.6: 27139318-27139318 on Build GRCh38
4609987 c.97C>T p.P33S Chr.6: 27139405-27139405 on Build GRCh38
4845995 c.106C>T p.R36W Chr.6: 27139414-27139414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751062963 Chr.6: 27139318-27139318 on Build GRCh38
rs777989743 Chr.6: 27139421-27139422 on Build GRCh38
rs991763555 Chr.6: 27139253-27139253 on Build GRCh38
rs771984935 Chr.6: 27139414-27139415 on Build GRCh38
rs748268091 Chr.6: 27139398-27139398 on Build GRCh38
rs958930223 Chr.6: 27139246-27139246 on Build GRCh38
rs753685066 Chr.6: 27139385-27139385 on Build GRCh38
rs564129943 Chr.6: 27139347-27139347 on Build GRCh38
rs1017330618 Chr.6: 27139240-27139240 on Build GRCh38
rs1039265007 Chr.6: 27139411-27139411 on Build GRCh38
rs970894517 Chr.6: 27139354-27139354 on Build GRCh38
rs755241311 Chr.6: 27139331-27139331 on Build GRCh38
rs761496441 Chr.6: 27139429-27139429 on Build GRCh38
rs763652001 Chr.6: 27139315-27139315 on Build GRCh38
rs778931172 Chr.6: 27139394-27139395 on Build GRCh38
rs913195920 Chr.6: 27139345-27139345 on Build GRCh38
rs373924281 Chr.6: 27139348-27139348 on Build GRCh38
rs757171424 Chr.6: 27139272-27139272 on Build GRCh38
rs202198731 Chr.6: 27139274-27139274 on Build GRCh38
rs768322550 Chr.6: 27139417-27139417 on Build GRCh38
rs773953710 Chr.6: 27139426-27139426 on Build GRCh38
rs773865845 Chr.6: 27139363-27139363 on Build GRCh38
rs775160286 Chr.6: 27139306-27139306 on Build GRCh38
rs369247236 Chr.6: 27139335-27139335 on Build GRCh38
rs182750738 Chr.6: 27139291-27139291 on Build GRCh38
rs143050720 Chr.6: 27139372-27139372 on Build GRCh38
rs772653571 Chr.6: 27139339-27139339 on Build GRCh38
rs773637987 Chr.6: 27139298-27139298 on Build GRCh38
rs924833981 Chr.6: 27139414-27139414 on Build GRCh38
rs560111648 Chr.6: 27139351-27139351 on Build GRCh38
rs761362967 Chr.6: 27139255-27139256 on Build GRCh38
rs764970467 Chr.6: 27139300-27139300 on Build GRCh38
rs757221402 Chr.6: 27139319-27139319 on Build GRCh38
rs749411146 Chr.6: 27139281-27139281 on Build GRCh38
rs201382302 Chr.6: 27139296-27139296 on Build GRCh38
rs755827089 Chr.6: 27139326-27139326 on Build GRCh38
rs746171478 Chr.6: 27139413-27139413 on Build GRCh38
rs754932977 Chr.6: 27139364-27139365 on Build GRCh38
rs751546994 Chr.6: 27139264-27139264 on Build GRCh38
rs771249191 Chr.6: 27139350-27139350 on Build GRCh38
rs756004217 Chr.6: 27139299-27139299 on Build GRCh38
rs376032611 Chr.6: 27139286-27139286 on Build GRCh38
rs766338156 Chr.6: 27139378-27139378 on Build GRCh38
rs970230173 Chr.6: 27139282-27139282 on Build GRCh38
rs753877604 Chr.6: 27139361-27139362 on Build GRCh38
rs761180213 Chr.6: 27139367-27139367 on Build GRCh38
rs779964017 Chr.6: 27139327-27139327 on Build GRCh38
rs767522132 Chr.6: 27139368-27139368 on Build GRCh38
rs747515954 Chr.6: 27139349-27139349 on Build GRCh38
rs762658565 Chr.6: 27139355-27139355 on Build GRCh38
rs760643847 Chr.6: 27139375-27139375 on Build GRCh38
rs762572412 Chr.6: 27139308-27139308 on Build GRCh38
rs151220380 Chr.6: 27139407-27139407 on Build GRCh38
rs750256756 Chr.6: 27139325-27139325 on Build GRCh38
rs963437703 Chr.6: 27139247-27139247 on Build GRCh38
rs763563038 Chr.6: 27139261-27139261 on Build GRCh38
rs760895572 Chr.6: 27139299-27139299 on Build GRCh38
rs758444195 Chr.6: 27139394-27139394 on Build GRCh38
rs777882267 Chr.6: 27139400-27139400 on Build GRCh38
rs994616347 Chr.6: 27139254-27139254 on Build GRCh38
rs377361526 Chr.6: 27139386-27139386 on Build GRCh38
rs763668364 Chr.6: 27139356-27139356 on Build GRCh38
rs781539533 Chr.6: 27139410-27139410 on Build GRCh38
rs372023652 Chr.6: 27139278-27139278 on Build GRCh38
rs936079641 Chr.6: 27139304-27139304 on Build GRCh38
rs779226325 Chr.6: 27139388-27139388 on Build GRCh38
rs752849854 Chr.6: 27139393-27139393 on Build GRCh38
rs749574795 Chr.6: 27139329-27139329 on Build GRCh38
rs996717639 Chr.6: 27139321-27139321 on Build GRCh38
rs779965916 Chr.6: 27139317-27139318 on Build GRCh38
rs780090802 Chr.6: 27139280-27139280 on Build GRCh38
rs575103151 Chr.6: 27139374-27139374 on Build GRCh38
rs767301868 Chr.6: 27139322-27139322 on Build GRCh38
rs140775845 Chr.6: 27139337-27139337 on Build GRCh38
rs1033744580 Chr.6: 27139366-27139366 on Build GRCh38
rs747527283 Chr.6: 27139404-27139404 on Build GRCh38

View Full Product Details