Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28616490-28616739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0100
Entrez Gene ID
9703
Gene Name
KIAA0100
Gene Aliases
BCOX, BCOX1, CT101, FMP27
UniGene
Hs.151761
Gene Chromosome Location
Chr.17: 28614440-28645159 on Build GRCh38
Transcripts
NM_014680.3 XM_005258073.2 XM_017025423.1 NR_040012.1

Gene Symbol
SPAG5-AS1
Entrez Gene ID
100506436
Gene Name
SPAG5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.743630
Gene Chromosome Location
Chr.17: 28599133-28617377 on Build GRCh38
Transcripts
NM_014680.3 XM_005258073.2 XM_017025423.1 NR_040012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4064961 c.6086T>A p.F2029Y Chr.17: 28616689-28616689 on Build GRCh38
3378105 c.6098G>A p.R2033Q Chr.17: 28616677-28616677 on Build GRCh38
3819114 c.6157+2T>G p.? Chr.17: 28616616-28616616 on Build GRCh38
6538691 c.6075G>C p.M2025I Chr.17: 28616700-28616700 on Build GRCh38
5663314 c.6098G>T p.R2033L Chr.17: 28616677-28616677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896040185 Chr.17: 28616542-28616542 on Build GRCh38
rs1028493243 Chr.17: 28616551-28616551 on Build GRCh38
rs553006736 Chr.17: 28616615-28616615 on Build GRCh38
rs749452404 Chr.17: 28616583-28616583 on Build GRCh38
rs953132441 Chr.17: 28616591-28616591 on Build GRCh38
rs754269381 Chr.17: 28616627-28616627 on Build GRCh38
rs772748667 Chr.17: 28616580-28616581 on Build GRCh38
rs756690315 Chr.17: 28616663-28616663 on Build GRCh38
rs373012241 Chr.17: 28616632-28616632 on Build GRCh38
rs763638904 Chr.17: 28616637-28616637 on Build GRCh38
rs200346281 Chr.17: 28616604-28616604 on Build GRCh38
rs768761789 Chr.17: 28616584-28616584 on Build GRCh38
rs748918145 Chr.17: 28616670-28616671 on Build GRCh38
rs1037245803 Chr.17: 28616519-28616519 on Build GRCh38
rs780968670 Chr.17: 28616548-28616548 on Build GRCh38
rs780781168 Chr.17: 28616685-28616685 on Build GRCh38
rs751273890 Chr.17: 28616641-28616641 on Build GRCh38
rs950780608 Chr.17: 28616679-28616679 on Build GRCh38
rs775281723 Chr.17: 28616575-28616575 on Build GRCh38
rs1023113363 Chr.17: 28616573-28616573 on Build GRCh38
rs769535213 Chr.17: 28616569-28616569 on Build GRCh38
rs750307079 Chr.17: 28616720-28616720 on Build GRCh38
rs766803007 Chr.17: 28616622-28616622 on Build GRCh38
rs117647565 Chr.17: 28616524-28616524 on Build GRCh38
rs756846954 Chr.17: 28616543-28616543 on Build GRCh38
rs370224353 Chr.17: 28616585-28616585 on Build GRCh38
rs372640102 Chr.17: 28616526-28616526 on Build GRCh38
rs777280354 Chr.17: 28616529-28616529 on Build GRCh38
rs773827434 Chr.17: 28616606-28616606 on Build GRCh38
rs761788929 Chr.17: 28616597-28616597 on Build GRCh38
rs1007304345 Chr.17: 28616661-28616661 on Build GRCh38
rs746494987 Chr.17: 28616536-28616536 on Build GRCh38

View Full Product Details