Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36177826-36178083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP7D1
Entrez Gene ID
55700
Gene Name
MAP7 domain containing 1
Gene Aliases
PARCC1, RPRC1
UniGene
Hs.356096
Gene Chromosome Location
Chr.1: 36155965-36180849 on Build GRCh38
Transcripts
NM_001286365.1 NM_001286366.1 NM_018067.4 XM_005271027.1 XM_006710759.1 XM_006710761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5157611 c.74delC p.C27fs*95 Chr.1: 36177929-36177929 on Build GRCh38
5448604 c.1461C>T p.G487G Chr.1: 36177954-36177954 on Build GRCh38
5135389 c.125delC p.R44fs*78 Chr.1: 36177980-36177980 on Build GRCh38
6126428 c.1562C>T p.A521V Chr.1: 36178055-36178055 on Build GRCh38
5448605 c.99C>T p.G33G Chr.1: 36177954-36177954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765761383 Chr.1: 36177938-36177938 on Build GRCh38
rs201548025 Chr.1: 36178019-36178019 on Build GRCh38
rs766311908 Chr.1: 36177851-36177851 on Build GRCh38
rs780204679 Chr.1: 36177899-36177899 on Build GRCh38
rs199802965 Chr.1: 36177984-36177984 on Build GRCh38
rs201908370 Chr.1: 36177963-36177963 on Build GRCh38
rs767160576 Chr.1: 36177859-36177859 on Build GRCh38
rs781072648 Chr.1: 36178034-36178034 on Build GRCh38
rs763077368 Chr.1: 36178054-36178054 on Build GRCh38
rs149009853 Chr.1: 36177996-36177996 on Build GRCh38
rs756959970 Chr.1: 36177850-36177856 on Build GRCh38
rs769546438 Chr.1: 36177921-36177921 on Build GRCh38
rs748578351 Chr.1: 36177910-36177910 on Build GRCh38
rs539606561 Chr.1: 36177886-36177886 on Build GRCh38
rs147794587 Chr.1: 36177906-36177906 on Build GRCh38
rs773737547 Chr.1: 36178001-36178001 on Build GRCh38
rs990801354 Chr.1: 36177903-36177903 on Build GRCh38
rs747416618 Chr.1: 36177905-36177905 on Build GRCh38
rs372974206 Chr.1: 36177909-36177909 on Build GRCh38
rs745916260 Chr.1: 36177897-36177897 on Build GRCh38
rs757467342 Chr.1: 36177885-36177885 on Build GRCh38
rs751567890 Chr.1: 36178055-36178055 on Build GRCh38
rs763905933 Chr.1: 36177980-36177980 on Build GRCh38
rs879414637 Chr.1: 36177908-36177908 on Build GRCh38
rs568854302 Chr.1: 36177854-36177854 on Build GRCh38
rs750592841 Chr.1: 36177981-36177981 on Build GRCh38
rs763817876 Chr.1: 36178051-36178051 on Build GRCh38
rs781776095 Chr.1: 36177992-36177992 on Build GRCh38
rs959761814 Chr.1: 36177902-36177902 on Build GRCh38
rs755155838 Chr.1: 36177856-36177856 on Build GRCh38
rs781456264 Chr.1: 36178061-36178061 on Build GRCh38
rs760627087 Chr.1: 36178049-36178049 on Build GRCh38
rs752494367 Chr.1: 36177868-36177868 on Build GRCh38
rs767558715 Chr.1: 36177958-36177958 on Build GRCh38
rs185056861 Chr.1: 36177957-36177957 on Build GRCh38
rs770622183 Chr.1: 36178016-36178016 on Build GRCh38
rs191169069 Chr.1: 36178057-36178057 on Build GRCh38
rs755424592 Chr.1: 36177988-36177988 on Build GRCh38
rs756706150 Chr.1: 36178062-36178062 on Build GRCh38
rs773042016 Chr.1: 36177925-36177925 on Build GRCh38
rs755953608 Chr.1: 36177977-36177977 on Build GRCh38
rs759145551 Chr.1: 36178027-36178027 on Build GRCh38
rs771884720 Chr.1: 36178029-36178029 on Build GRCh38
rs759326459 Chr.1: 36177955-36177955 on Build GRCh38
rs762729140 Chr.1: 36177931-36177931 on Build GRCh38
rs748839215 Chr.1: 36178013-36178013 on Build GRCh38
rs150251502 Chr.1: 36177974-36177974 on Build GRCh38
rs866615299 Chr.1: 36178058-36178058 on Build GRCh38
rs773928691 Chr.1: 36177948-36177948 on Build GRCh38

View Full Product Details