Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11111449-11111710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLR
Entrez Gene ID
3949
Gene Name
low density lipoprotein receptor
Gene Aliases
FH, FHC, LDLCQ2
UniGene
Hs.213289
Gene Chromosome Location
Chr.19: 11089362-11133830 on Build GRCh38
Transcripts
NM_000527.4 NM_001195798.1 NM_001195799.1 NM_001195800.1 NM_001195803.1 XM_011528010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1390294 c.1125C>T p.Y375Y Chr.19: 11111578-11111578 on Build GRCh38
6700303 c.1106T>C p.V369A Chr.19: 11111559-11111559 on Build GRCh38
3528466 c.1082C>T p.P361L Chr.19: 11111535-11111535 on Build GRCh38
7374889 c.1066G>T p.D356Y Chr.19: 11111519-11111519 on Build GRCh38
6569079 c.1101C>G p.L367L Chr.19: 11111554-11111554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553883598 Chr.19: 11111669-11111669 on Build GRCh38
rs879254785 Chr.19: 11111529-11111534 on Build GRCh38
rs758493253 Chr.19: 11111634-11111634 on Build GRCh38
rs879254779 Chr.19: 11111521-11111521 on Build GRCh38
rs756672317 Chr.19: 11111613-11111613 on Build GRCh38
rs879254790 Chr.19: 11111552-11111557 on Build GRCh38
rs149227308 Chr.19: 11111619-11111619 on Build GRCh38
rs567564778 Chr.19: 11111536-11111536 on Build GRCh38
rs879254787 Chr.19: 11111543-11111543 on Build GRCh38
rs760526525 Chr.19: 11111480-11111480 on Build GRCh38
rs879254784 Chr.19: 11111528-11111528 on Build GRCh38
rs879254789 Chr.19: 11111549-11111549 on Build GRCh38
rs996616174 Chr.19: 11111468-11111468 on Build GRCh38
rs879254809 Chr.19: 11111607-11111607 on Build GRCh38
rs765315456 Chr.19: 11111678-11111678 on Build GRCh38
rs879254799 Chr.19: 11111576-11111577 on Build GRCh38
rs730882097 Chr.19: 11111558-11111558 on Build GRCh38
rs879254773 Chr.19: 11111512-11111512 on Build GRCh38
rs879254819 Chr.19: 11111639-11111639 on Build GRCh38
rs879254782 Chr.19: 11111526-11111527 on Build GRCh38
rs572365788 Chr.19: 11111670-11111670 on Build GRCh38
rs767935341 Chr.19: 11111597-11111597 on Build GRCh38
rs879254780 Chr.19: 11111539-11111540 on Build GRCh38
rs879254821 Chr.19: 11111644-11111644 on Build GRCh38
rs771744014 Chr.19: 11111473-11111473 on Build GRCh38
rs560045570 Chr.19: 11111474-11111474 on Build GRCh38
rs879254798 Chr.19: 11111573-11111574 on Build GRCh38
rs879254796 Chr.19: 11111570-11111570 on Build GRCh38
rs769432647 Chr.19: 11111658-11111658 on Build GRCh38
rs879254783 Chr.19: 11111525-11111525 on Build GRCh38
rs768430352 Chr.19: 11111556-11111556 on Build GRCh38
rs773064328 Chr.19: 11111582-11111582 on Build GRCh38
rs779921498 Chr.19: 11111641-11111641 on Build GRCh38
rs960032925 Chr.19: 11111518-11111518 on Build GRCh38
rs748093803 Chr.19: 11111655-11111655 on Build GRCh38
rs879254778 Chr.19: 11111520-11111520 on Build GRCh38
rs763666203 Chr.19: 11111485-11111485 on Build GRCh38
rs928536522 Chr.19: 11111668-11111668 on Build GRCh38
rs200533979 Chr.19: 11111541-11111541 on Build GRCh38
rs756518976 Chr.19: 11111527-11111527 on Build GRCh38
rs746742171 Chr.19: 11111647-11111647 on Build GRCh38
rs751791403 Chr.19: 11111683-11111683 on Build GRCh38
rs879254808 Chr.19: 11111606-11111606 on Build GRCh38
rs752951310 Chr.19: 11111598-11111598 on Build GRCh38
rs879254818 Chr.19: 11111638-11111638 on Build GRCh38
rs879254781 Chr.19: 11111522-11111522 on Build GRCh38
rs774021285 Chr.19: 11111667-11111667 on Build GRCh38
rs879254795 Chr.19: 11111568-11111576 on Build GRCh38
rs768475018 Chr.19: 11111648-11111648 on Build GRCh38
rs730880131 Chr.19: 11111640-11111640 on Build GRCh38
rs536387604 Chr.19: 11111680-11111680 on Build GRCh38
rs879254820 Chr.19: 11111639-11111639 on Build GRCh38
rs879254807 Chr.19: 11111604-11111604 on Build GRCh38
rs750649426 Chr.19: 11111629-11111629 on Build GRCh38
rs879254817 Chr.19: 11111635-11111635 on Build GRCh38
rs138315511 Chr.19: 11111538-11111538 on Build GRCh38
rs879254777 Chr.19: 11111520-11111520 on Build GRCh38
rs996689598 Chr.19: 11111656-11111656 on Build GRCh38
rs746982741 Chr.19: 11111550-11111550 on Build GRCh38
rs875989915 Chr.19: 11111526-11111526 on Build GRCh38
rs875989917 Chr.19: 11111615-11111615 on Build GRCh38
rs202054816 Chr.19: 11111469-11111469 on Build GRCh38
rs781336025 Chr.19: 11111649-11111649 on Build GRCh38
rs749322464 Chr.19: 11111533-11111533 on Build GRCh38
rs200740958 Chr.19: 11111659-11111659 on Build GRCh38
rs764551797 Chr.19: 11111615-11111615 on Build GRCh38
rs879254804 Chr.19: 11111589-11111589 on Build GRCh38
rs11669576 Chr.19: 11111624-11111624 on Build GRCh38
rs879254806 Chr.19: 11111604-11111612 on Build GRCh38
rs892938153 Chr.19: 11111676-11111676 on Build GRCh38
rs1045407543 Chr.19: 11111661-11111661 on Build GRCh38
rs879254822 Chr.19: 11111650-11111650 on Build GRCh38
rs879254776 Chr.19: 11111516-11111516 on Build GRCh38
rs879254794 Chr.19: 11111566-11111567 on Build GRCh38
rs879254800 Chr.19: 11111577-11111577 on Build GRCh38
rs879254803 Chr.19: 11111588-11111588 on Build GRCh38
rs879254816 Chr.19: 11111632-11111632 on Build GRCh38
rs879254812 Chr.19: 11111626-11111626 on Build GRCh38
rs983617416 Chr.19: 11111591-11111591 on Build GRCh38
rs767767730 Chr.19: 11111519-11111519 on Build GRCh38
rs879254793 Chr.19: 11111563-11111563 on Build GRCh38
rs879254813 Chr.19: 11111627-11111628 on Build GRCh38
rs879254815 Chr.19: 11111631-11111631 on Build GRCh38
rs764826590 Chr.19: 11111572-11111572 on Build GRCh38
rs875989916 Chr.19: 11111574-11111575 on Build GRCh38
rs750454097 Chr.19: 11111679-11111679 on Build GRCh38
rs879254792 Chr.19: 11111562-11111562 on Build GRCh38
rs113669610 Chr.19: 11111557-11111557 on Build GRCh38
rs755449669 Chr.19: 11111514-11111514 on Build GRCh38
rs765020792 Chr.19: 11111505-11111505 on Build GRCh38
rs879254805 Chr.19: 11111603-11111603 on Build GRCh38
rs527401868 Chr.19: 11111492-11111492 on Build GRCh38
rs777926251 Chr.19: 11111531-11111531 on Build GRCh38
rs879254811 Chr.19: 11111621-11111621 on Build GRCh38
rs770448033 Chr.19: 11111665-11111665 on Build GRCh38
rs879254802 Chr.19: 11111585-11111585 on Build GRCh38
rs139066906 Chr.19: 11111620-11111620 on Build GRCh38
rs879254814 Chr.19: 11111627-11111627 on Build GRCh38
rs879254791 Chr.19: 11111555-11111555 on Build GRCh38
rs753248521 Chr.19: 11111523-11111523 on Build GRCh38
rs879254774 Chr.19: 11111513-11111513 on Build GRCh38
rs879254775 Chr.19: 11111515-11111516 on Build GRCh38
rs761581036 Chr.19: 11111495-11111495 on Build GRCh38
rs879254788 Chr.19: 11111544-11111544 on Build GRCh38
rs72658861 Chr.19: 11111506-11111506 on Build GRCh38
rs879254801 Chr.19: 11111583-11111583 on Build GRCh38
rs879254797 Chr.19: 11111571-11111571 on Build GRCh38
rs201971341 Chr.19: 11111668-11111668 on Build GRCh38
rs878854024 Chr.19: 11111551-11111562 on Build GRCh38
rs879254810 Chr.19: 11111611-11111611 on Build GRCh38
rs730882098 Chr.19: 11111586-11111586 on Build GRCh38
rs879254786 Chr.19: 11111538-11111538 on Build GRCh38

View Full Product Details