Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32737363-32737630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1
Entrez Gene ID
94104
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
BM020, C21orf66, FSAP105, GCFC, GCFC1
UniGene
Hs.644004
Gene Chromosome Location
Chr.21: 32733899-32771858 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Gene Symbol
PAXBP1-AS1
Entrez Gene ID
100506215
Gene Name
PAXBP1 antisense RNA 1
Gene Aliases
GCFC1-AS1
UniGene
Hs.657123
Gene Chromosome Location
Chr.21: 32728114-32743122 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030240 c.2505A>C p.Q835H Chr.21: 32737385-32737385 on Build GRCh38
4101263 c.2524G>A p.G842R Chr.21: 32737366-32737366 on Build GRCh38
6738308 c.2499_2500insC p.K834fs*32 Chr.21: 32737390-32737391 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs549339725 Chr.21: 32737458-32737458 on Build GRCh38
rs568313583 Chr.21: 32737511-32737511 on Build GRCh38
rs774096522 Chr.21: 32737571-32737571 on Build GRCh38
rs1026991454 Chr.21: 32737530-32737530 on Build GRCh38
rs760041666 Chr.21: 32737422-32737422 on Build GRCh38
rs924680284 Chr.21: 32737568-32737568 on Build GRCh38
rs761318734 Chr.21: 32737412-32737412 on Build GRCh38
rs914738015 Chr.21: 32737452-32737452 on Build GRCh38
rs779176388 Chr.21: 32737445-32737445 on Build GRCh38
rs1025050773 Chr.21: 32737599-32737599 on Build GRCh38
rs947590297 Chr.21: 32737479-32737479 on Build GRCh38
rs763726562 Chr.21: 32737527-32737527 on Build GRCh38
rs568680311 Chr.21: 32737602-32737602 on Build GRCh38
rs746856382 Chr.21: 32737428-32737428 on Build GRCh38
rs535356129 Chr.21: 32737518-32737518 on Build GRCh38
rs775447295 Chr.21: 32737394-32737394 on Build GRCh38
rs746060549 Chr.21: 32737446-32737446 on Build GRCh38
rs753881379 Chr.21: 32737439-32737439 on Build GRCh38
rs1012351577 Chr.21: 32737586-32737586 on Build GRCh38
rs757348900 Chr.21: 32737441-32737441 on Build GRCh38
rs370348660 Chr.21: 32737401-32737401 on Build GRCh38
rs776036603 Chr.21: 32737405-32737405 on Build GRCh38
rs144102538 Chr.21: 32737403-32737403 on Build GRCh38
rs750084791 Chr.21: 32737417-32737417 on Build GRCh38
rs376033585 Chr.21: 32737415-32737415 on Build GRCh38
rs755227711 Chr.21: 32737417-32737420 on Build GRCh38
rs556737710 Chr.21: 32737553-32737553 on Build GRCh38
rs190913591 Chr.21: 32737425-32737425 on Build GRCh38
rs778279679 Chr.21: 32737438-32737438 on Build GRCh38
rs779077920 Chr.21: 32737427-32737431 on Build GRCh38
rs756597122 Chr.21: 32737432-32737432 on Build GRCh38
rs959338519 Chr.21: 32737582-32737582 on Build GRCh38
rs375035076 Chr.21: 32737426-32737426 on Build GRCh38

View Full Product Details