Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64822355-64822592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIF1
Entrez Gene ID
80119
Gene Name
PIF1 5'-to-3' DNA helicase
Gene Aliases
C15orf20, PIF
UniGene
Hs.112160
Gene Chromosome Location
Chr.15: 64815630-64827173 on Build GRCh38
Transcripts
NM_001286496.1 NM_001286497.1 NM_001286499.1 NM_025049.3 XM_011522083.2 XM_011522084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4056240 c.668G>A p.S223N Chr.15: 64822501-64822501 on Build GRCh38
1301329 c.652G>A p.V218I Chr.15: 64822517-64822517 on Build GRCh38
6142522 c.651C>A p.A217A Chr.15: 64822518-64822518 on Build GRCh38
7383868 c.677T>A p.F226Y Chr.15: 64822492-64822492 on Build GRCh38
6846424 c.672C>G p.I224M Chr.15: 64822497-64822497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778406904 Chr.15: 64822443-64822443 on Build GRCh38
rs745897224 Chr.15: 64822535-64822535 on Build GRCh38
rs760464296 Chr.15: 64822496-64822496 on Build GRCh38
rs764706842 Chr.15: 64822507-64822507 on Build GRCh38
rs921945209 Chr.15: 64822453-64822453 on Build GRCh38
rs772201758 Chr.15: 64822538-64822538 on Build GRCh38
rs974792648 Chr.15: 64822444-64822444 on Build GRCh38
rs371480665 Chr.15: 64822518-64822518 on Build GRCh38
rs749441992 Chr.15: 64822451-64822451 on Build GRCh38
rs768410631 Chr.15: 64822569-64822569 on Build GRCh38
rs779169058 Chr.15: 64822528-64822528 on Build GRCh38
rs764077234 Chr.15: 64822497-64822497 on Build GRCh38
rs749907190 Chr.15: 64822509-64822509 on Build GRCh38
rs778461909 Chr.15: 64822523-64822523 on Build GRCh38
rs748478402 Chr.15: 64822425-64822425 on Build GRCh38
rs753359912 Chr.15: 64822379-64822379 on Build GRCh38
rs201060080 Chr.15: 64822411-64822411 on Build GRCh38
rs781439749 Chr.15: 64822414-64822414 on Build GRCh38
rs374923447 Chr.15: 64822517-64822517 on Build GRCh38
rs762683264 Chr.15: 64822383-64822418 on Build GRCh38
rs984009166 Chr.15: 64822384-64822384 on Build GRCh38
rs772300460 Chr.15: 64822468-64822468 on Build GRCh38
rs139868280 Chr.15: 64822501-64822501 on Build GRCh38
rs756747881 Chr.15: 64822395-64822395 on Build GRCh38
rs746102563 Chr.15: 64822461-64822461 on Build GRCh38
rs757793016 Chr.15: 64822525-64822525 on Build GRCh38
rs201269313 Chr.15: 64822436-64822436 on Build GRCh38
rs72744798 Chr.15: 64822557-64822557 on Build GRCh38
rs140985919 Chr.15: 64822493-64822493 on Build GRCh38
rs775155544 Chr.15: 64822483-64822483 on Build GRCh38
rs766025004 Chr.15: 64822384-64822384 on Build GRCh38
rs757989843 Chr.15: 64822514-64822514 on Build GRCh38
rs923835366 Chr.15: 64822474-64822474 on Build GRCh38
rs761354683 Chr.15: 64822503-64822503 on Build GRCh38
rs142382246 Chr.15: 64822524-64822524 on Build GRCh38
rs780310704 Chr.15: 64822544-64822544 on Build GRCh38
rs764938355 Chr.15: 64822396-64822396 on Build GRCh38
rs774761195 Chr.15: 64822452-64822452 on Build GRCh38
rs369793024 Chr.15: 64822400-64822400 on Build GRCh38
rs751389992 Chr.15: 64822491-64822493 on Build GRCh38

View Full Product Details