Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27912104-27912351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2B2
Entrez Gene ID
81697
Gene Name
olfactory receptor family 2 subfamily B member 2
Gene Aliases
OR2B2Q, OR2B9, OR6-1, dJ193B12.4, hs6M1-10
UniGene
Hs.307111
Gene Chromosome Location
Chr.6: 27911185-27912396 on Build GRCh38
Transcripts
NM_033057.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1169073 c.181T>A p.F61I Chr.6: 27912139-27912139 on Build GRCh38
6216111 c.134T>G p.I45R Chr.6: 27912186-27912186 on Build GRCh38
1260115 c.83T>G p.F28C Chr.6: 27912237-27912237 on Build GRCh38
4709752 c.23T>C p.V8A Chr.6: 27912297-27912297 on Build GRCh38
354520 c.62C>T p.P21L Chr.6: 27912258-27912258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054683888 Chr.6: 27912202-27912202 on Build GRCh38
rs750943860 Chr.6: 27912265-27912265 on Build GRCh38
rs763258479 Chr.6: 27912230-27912230 on Build GRCh38
rs762502395 Chr.6: 27912300-27912301 on Build GRCh38
rs764201927 Chr.6: 27912231-27912231 on Build GRCh38
rs759113107 Chr.6: 27912134-27912134 on Build GRCh38
rs1045916413 Chr.6: 27912152-27912152 on Build GRCh38
rs150184028 Chr.6: 27912211-27912211 on Build GRCh38
rs189300279 Chr.6: 27912154-27912154 on Build GRCh38
rs893486075 Chr.6: 27912187-27912187 on Build GRCh38
rs889077279 Chr.6: 27912216-27912216 on Build GRCh38
rs771572070 Chr.6: 27912172-27912172 on Build GRCh38
rs752446861 Chr.6: 27912279-27912280 on Build GRCh38
rs780415776 Chr.6: 27912314-27912314 on Build GRCh38
rs1028037966 Chr.6: 27912270-27912270 on Build GRCh38
rs527922738 Chr.6: 27912224-27912224 on Build GRCh38
rs61742093 Chr.6: 27912204-27912204 on Build GRCh38
rs772824979 Chr.6: 27912173-27912173 on Build GRCh38
rs746470182 Chr.6: 27912174-27912174 on Build GRCh38
rs756460482 Chr.6: 27912148-27912148 on Build GRCh38
rs376074748 Chr.6: 27912196-27912196 on Build GRCh38
rs946761525 Chr.6: 27912209-27912209 on Build GRCh38
rs937294127 Chr.6: 27912195-27912195 on Build GRCh38
rs764618877 Chr.6: 27912259-27912260 on Build GRCh38
rs746302286 Chr.6: 27912145-27912145 on Build GRCh38
rs935123308 Chr.6: 27912170-27912170 on Build GRCh38
rs770967801 Chr.6: 27912176-27912176 on Build GRCh38
rs751705341 Chr.6: 27912241-27912241 on Build GRCh38
rs756570686 Chr.6: 27912275-27912275 on Build GRCh38
rs781644838 Chr.6: 27912127-27912127 on Build GRCh38
rs768073385 Chr.6: 27912262-27912262 on Build GRCh38
rs138657607 Chr.6: 27912320-27912320 on Build GRCh38
rs776694562 Chr.6: 27912181-27912181 on Build GRCh38
rs952084257 Chr.6: 27912299-27912299 on Build GRCh38
rs746305654 Chr.6: 27912149-27912149 on Build GRCh38
rs144903141 Chr.6: 27912260-27912260 on Build GRCh38
rs181039572 Chr.6: 27912157-27912157 on Build GRCh38

View Full Product Details