Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36156267-36156506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP7D1
Entrez Gene ID
55700
Gene Name
MAP7 domain containing 1
Gene Aliases
PARCC1, RPRC1
UniGene
Hs.356096
Gene Chromosome Location
Chr.1: 36155965-36180849 on Build GRCh38
Transcripts
NM_001286365.1 NM_001286366.1 NM_018067.4 XM_005271027.1 XM_006710759.1 XM_006710761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5419481 c.46+6C>G p.? Chr.1: 36156469-36156469 on Build GRCh38
4988639 c.4G>A p.E2K Chr.1: 36156421-36156421 on Build GRCh38
238534 c.44delC p.P15fs*41 Chr.1: 36156461-36156461 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs957075659 Chr.1: 36156391-36156391 on Build GRCh38
rs975023593 Chr.1: 36156308-36156309 on Build GRCh38
rs764383171 Chr.1: 36156461-36156461 on Build GRCh38
rs970959837 Chr.1: 36156339-36156339 on Build GRCh38
rs1003793923 Chr.1: 36156367-36156367 on Build GRCh38
rs770592856 Chr.1: 36156399-36156399 on Build GRCh38
rs905194082 Chr.1: 36156364-36156365 on Build GRCh38
rs1036836295 Chr.1: 36156316-36156316 on Build GRCh38
rs946478150 Chr.1: 36156296-36156296 on Build GRCh38
rs530814067 Chr.1: 36156421-36156421 on Build GRCh38
rs988304787 Chr.1: 36156393-36156393 on Build GRCh38
rs899223697 Chr.1: 36156329-36156329 on Build GRCh38
rs12730614 Chr.1: 36156396-36156396 on Build GRCh38
rs1023604410 Chr.1: 36156354-36156354 on Build GRCh38
rs2274166 Chr.1: 36156309-36156309 on Build GRCh38
rs879295803 Chr.1: 36156431-36156431 on Build GRCh38
rs745390462 Chr.1: 36156455-36156455 on Build GRCh38
rs939899897 Chr.1: 36156315-36156315 on Build GRCh38
rs1031318771 Chr.1: 36156375-36156375 on Build GRCh38
rs552588202 Chr.1: 36156450-36156450 on Build GRCh38
rs886935700 Chr.1: 36156314-36156314 on Build GRCh38
rs986667315 Chr.1: 36156482-36156482 on Build GRCh38
rs966905526 Chr.1: 36156346-36156346 on Build GRCh38
rs1051002828 Chr.1: 36156310-36156310 on Build GRCh38
rs778228155 Chr.1: 36156418-36156418 on Build GRCh38
rs571047755 Chr.1: 36156469-36156469 on Build GRCh38
rs975135452 Chr.1: 36156423-36156423 on Build GRCh38
rs957999016 Chr.1: 36156390-36156390 on Build GRCh38
rs911233697 Chr.1: 36156429-36156429 on Build GRCh38
rs964084920 Chr.1: 36156432-36156432 on Build GRCh38
rs1022901524 Chr.1: 36156403-36156403 on Build GRCh38
rs371643371 Chr.1: 36156309-36156313 on Build GRCh38
rs999396456 Chr.1: 36156347-36156347 on Build GRCh38
rs771901097 Chr.1: 36156473-36156473 on Build GRCh38

View Full Product Details