Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43361749-43362016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN29
Entrez Gene ID
146050
Gene Name
zinc finger and SCAN domain containing 29
Gene Aliases
ZNF690, Zfp690
UniGene
Hs.418287
Gene Chromosome Location
Chr.15: 43358172-43371032 on Build GRCh38
Transcripts
NM_152455.3 XM_006720401.3 XM_011521266.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3672049 c.1714G>T p.E572* Chr.15: 43361918-43361918 on Build GRCh38
4054821 c.1825A>C p.S609R Chr.15: 43361807-43361807 on Build GRCh38
1373008 c.1879A>C p.K627Q Chr.15: 43361753-43361753 on Build GRCh38
2188181 c.1822G>C p.E608Q Chr.15: 43361810-43361810 on Build GRCh38
3501404 c.1784G>A p.R595K Chr.15: 43361848-43361848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572072217 Chr.15: 43361825-43361825 on Build GRCh38
rs775996530 Chr.15: 43361970-43361970 on Build GRCh38
rs528044686 Chr.15: 43361947-43361947 on Build GRCh38
rs756391602 Chr.15: 43361861-43361861 on Build GRCh38
rs549066454 Chr.15: 43361924-43361924 on Build GRCh38
rs780048155 Chr.15: 43361797-43361797 on Build GRCh38
rs1037149140 Chr.15: 43361905-43361905 on Build GRCh38
rs772593046 Chr.15: 43361813-43361813 on Build GRCh38
rs1022681611 Chr.15: 43361990-43361990 on Build GRCh38
rs368170187 Chr.15: 43361810-43361810 on Build GRCh38
rs138731321 Chr.15: 43361871-43361871 on Build GRCh38
rs200716301 Chr.15: 43361863-43361863 on Build GRCh38
rs542634391 Chr.15: 43361826-43361826 on Build GRCh38
rs764178423 Chr.15: 43361899-43361899 on Build GRCh38
rs551085809 Chr.15: 43361979-43361979 on Build GRCh38
rs112463491 Chr.15: 43361853-43361853 on Build GRCh38
rs368938451 Chr.15: 43361922-43361922 on Build GRCh38
rs771903211 Chr.15: 43361791-43361791 on Build GRCh38
rs141880595 Chr.15: 43361836-43361836 on Build GRCh38
rs545133458 Chr.15: 43361903-43361903 on Build GRCh38
rs888510264 Chr.15: 43361902-43361902 on Build GRCh38
rs146267149 Chr.15: 43361844-43361844 on Build GRCh38
rs921257955 Chr.15: 43361872-43361872 on Build GRCh38
rs192532525 Chr.15: 43361837-43361837 on Build GRCh38
rs749446945 Chr.15: 43361772-43361772 on Build GRCh38
rs768198434 Chr.15: 43361957-43361957 on Build GRCh38
rs375945705 Chr.15: 43361906-43361906 on Build GRCh38
rs746726024 Chr.15: 43361799-43361799 on Build GRCh38
rs142581706 Chr.15: 43361929-43361929 on Build GRCh38
rs554873450 Chr.15: 43361968-43361971 on Build GRCh38
rs748000591 Chr.15: 43361912-43361912 on Build GRCh38
rs769369588 Chr.15: 43361919-43361919 on Build GRCh38
rs6493085 Chr.15: 43361984-43361984 on Build GRCh38
rs536928990 Chr.15: 43361987-43361987 on Build GRCh38
rs374784714 Chr.15: 43361945-43361945 on Build GRCh38
rs768468072 Chr.15: 43361800-43361800 on Build GRCh38
rs757374709 Chr.15: 43361873-43361873 on Build GRCh38
rs776448467 Chr.15: 43361809-43361809 on Build GRCh38
rs371680057 Chr.15: 43361884-43361884 on Build GRCh38
rs763732688 Chr.15: 43361938-43361984 on Build GRCh38
rs138379738 Chr.15: 43361812-43361812 on Build GRCh38
rs1033216558 Chr.15: 43361910-43361910 on Build GRCh38

View Full Product Details