Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 167242504-167242741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBBX
Entrez Gene ID
79740
Gene Name
zinc finger B-box domain containing
Gene Aliases
-
UniGene
Hs.478143
Gene Chromosome Location
Chr.3: 167178399-167380405 on Build GRCh38
Transcripts
NM_001199201.1 NM_001199202.1 NM_024687.3 XM_011513144.1 XM_011513145.2 XM_011513148.2 XM_011513149.2 XM_011513151.2 XM_011513152.2 XM_011513153.2 XM_017007186.1 XM_017007187.1 XM_017007188.1 XM_017007189.1 XM_017007190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3702355 c.2226T>C p.D742D Chr.3: 167242555-167242555 on Build GRCh38
3589664 c.2187C>T p.F729F Chr.3: 167242594-167242594 on Build GRCh38
4115016 c.2223A>C p.T741T Chr.3: 167242558-167242558 on Build GRCh38
3783679 c.2380G>T p.V794F Chr.3: 167242518-167242518 on Build GRCh38
729268 c.2326A>G p.S776G Chr.3: 167242572-167242572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014415427 Chr.3: 167242698-167242698 on Build GRCh38
rs763497869 Chr.3: 167242566-167242566 on Build GRCh38
rs777106380 Chr.3: 167242535-167242535 on Build GRCh38
rs372050245 Chr.3: 167242694-167242694 on Build GRCh38
rs773706541 Chr.3: 167242664-167242664 on Build GRCh38
rs202081660 Chr.3: 167242655-167242655 on Build GRCh38
rs931965118 Chr.3: 167242536-167242536 on Build GRCh38
rs377025827 Chr.3: 167242540-167242540 on Build GRCh38
rs746733565 Chr.3: 167242530-167242530 on Build GRCh38
rs1052589247 Chr.3: 167242662-167242662 on Build GRCh38
rs35857002 Chr.3: 167242624-167242624 on Build GRCh38
rs757021422 Chr.3: 167242600-167242600 on Build GRCh38
rs545786776 Chr.3: 167242580-167242580 on Build GRCh38
rs905903701 Chr.3: 167242611-167242611 on Build GRCh38
rs748071593 Chr.3: 167242656-167242660 on Build GRCh38
rs752921130 Chr.3: 167242599-167242599 on Build GRCh38
rs759561737 Chr.3: 167242677-167242677 on Build GRCh38
rs374545588 Chr.3: 167242616-167242616 on Build GRCh38
rs767602444 Chr.3: 167242585-167242585 on Build GRCh38
rs770069712 Chr.3: 167242647-167242647 on Build GRCh38
rs781297155 Chr.3: 167242529-167242529 on Build GRCh38
rs1020759360 Chr.3: 167242710-167242710 on Build GRCh38
rs747789308 Chr.3: 167242645-167242645 on Build GRCh38
rs766631435 Chr.3: 167242568-167242568 on Build GRCh38
rs768185206 Chr.3: 167242534-167242534 on Build GRCh38
rs746504363 Chr.3: 167242630-167242630 on Build GRCh38
rs762053057 Chr.3: 167242538-167242538 on Build GRCh38
rs1043747814 Chr.3: 167242601-167242601 on Build GRCh38
rs776179745 Chr.3: 167242635-167242635 on Build GRCh38
rs779601131 Chr.3: 167242629-167242629 on Build GRCh38
rs773880341 Chr.3: 167242559-167242559 on Build GRCh38
rs370917356 Chr.3: 167242571-167242571 on Build GRCh38
rs766969757 Chr.3: 167242675-167242675 on Build GRCh38
rs950141767 Chr.3: 167242592-167242592 on Build GRCh38
rs768395438 Chr.3: 167242634-167242634 on Build GRCh38
rs148568021 Chr.3: 167242529-167242529 on Build GRCh38
rs896693235 Chr.3: 167242673-167242673 on Build GRCh38
rs763611801 Chr.3: 167242670-167242670 on Build GRCh38
rs1046211989 Chr.3: 167242605-167242605 on Build GRCh38
rs778619813 Chr.3: 167242613-167242613 on Build GRCh38
rs774612396 Chr.3: 167242676-167242676 on Build GRCh38
rs755276561 Chr.3: 167242575-167242575 on Build GRCh38

View Full Product Details