Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81354494-81354727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAN
Entrez Gene ID
8139
Gene Name
gigaxonin
Gene Aliases
GAN1, KLHL16
UniGene
Hs.112569
Gene Chromosome Location
Chr.16: 81314941-81380198 on Build GRCh38
Transcripts
NM_022041.3 XM_017023734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669207 c.484C>A p.R162R Chr.16: 81354606-81354606 on Build GRCh38
6669210 c.533T>G p.L178R Chr.16: 81354655-81354655 on Build GRCh38
6079874 c.565G>T p.V189F Chr.16: 81354687-81354687 on Build GRCh38
5596764 c.484C>T p.R162* Chr.16: 81354606-81354606 on Build GRCh38
6079875 c.412C>G p.R138G Chr.16: 81354534-81354534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762452364 Chr.16: 81354575-81354575 on Build GRCh38
rs754108287 Chr.16: 81354611-81354611 on Build GRCh38
rs763438490 Chr.16: 81354562-81354562 on Build GRCh38
rs754860983 Chr.16: 81354513-81354513 on Build GRCh38
rs771476213 Chr.16: 81354527-81354527 on Build GRCh38
rs773126137 Chr.16: 81354666-81354666 on Build GRCh38
rs993338852 Chr.16: 81354692-81354692 on Build GRCh38
rs766014086 Chr.16: 81354626-81354626 on Build GRCh38
rs143187097 Chr.16: 81354622-81354622 on Build GRCh38
rs745558424 Chr.16: 81354646-81354646 on Build GRCh38
rs768074318 Chr.16: 81354660-81354660 on Build GRCh38
rs143704621 Chr.16: 81354593-81354593 on Build GRCh38
rs148122732 Chr.16: 81354675-81354675 on Build GRCh38
rs758366654 Chr.16: 81354619-81354619 on Build GRCh38
rs748121633 Chr.16: 81354525-81354525 on Build GRCh38
rs767483792 Chr.16: 81354587-81354587 on Build GRCh38
rs373474009 Chr.16: 81354601-81354601 on Build GRCh38
rs775953461 Chr.16: 81354548-81354548 on Build GRCh38
rs776404697 Chr.16: 81354624-81354624 on Build GRCh38
rs139748896 Chr.16: 81354694-81354694 on Build GRCh38
rs762169549 Chr.16: 81354652-81354652 on Build GRCh38
rs899127418 Chr.16: 81354642-81354642 on Build GRCh38
rs778744880 Chr.16: 81354612-81354612 on Build GRCh38
rs991469553 Chr.16: 81354572-81354572 on Build GRCh38
rs1034377391 Chr.16: 81354654-81354654 on Build GRCh38
rs555585848 Chr.16: 81354607-81354607 on Build GRCh38
rs201884522 Chr.16: 81354686-81354686 on Build GRCh38
rs119485095 Chr.16: 81354627-81354627 on Build GRCh38
rs772833509 Chr.16: 81354534-81354534 on Build GRCh38
rs764605890 Chr.16: 81354566-81354566 on Build GRCh38
rs145789614 Chr.16: 81354521-81354521 on Build GRCh38
rs770618198 Chr.16: 81354539-81354539 on Build GRCh38
rs119485092 Chr.16: 81354535-81354535 on Build GRCh38
rs553692036 Chr.16: 81354614-81354614 on Build GRCh38
rs369703737 Chr.16: 81354567-81354567 on Build GRCh38
rs1022894311 Chr.16: 81354578-81354578 on Build GRCh38
rs766484755 Chr.16: 81354687-81354687 on Build GRCh38
rs112693832 Chr.16: 81354650-81354650 on Build GRCh38
rs1035955786 Chr.16: 81354546-81354546 on Build GRCh38
rs770663433 Chr.16: 81354623-81354623 on Build GRCh38
rs755250146 Chr.16: 81354693-81354693 on Build GRCh38

View Full Product Details