Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28616682-28616953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0100
Entrez Gene ID
9703
Gene Name
KIAA0100
Gene Aliases
BCOX, BCOX1, CT101, FMP27
UniGene
Hs.151761
Gene Chromosome Location
Chr.17: 28614440-28645159 on Build GRCh38
Transcripts
NM_014680.3 XM_005258073.2 XM_017025423.1 NR_040012.1

Gene Symbol
SPAG5-AS1
Entrez Gene ID
100506436
Gene Name
SPAG5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.743630
Gene Chromosome Location
Chr.17: 28599133-28617377 on Build GRCh38
Transcripts
NM_014680.3 XM_005258073.2 XM_017025423.1 NR_040012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4064961 c.6086T>A p.F2029Y Chr.17: 28616689-28616689 on Build GRCh38
4064963 c.5980C>T p.R1994W Chr.17: 28616930-28616930 on Build GRCh38
4064962 c.6086T>A p.F2029Y Chr.17: 28616689-28616689 on Build GRCh38
6692953 c.5982G>C p.R1994R Chr.17: 28616928-28616928 on Build GRCh38
4501912 c.6002C>T p.S2001F Chr.17: 28616908-28616908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199533260 Chr.17: 28616795-28616795 on Build GRCh38
rs372910327 Chr.17: 28616778-28616778 on Build GRCh38
rs779957103 Chr.17: 28616849-28616849 on Build GRCh38
rs938109117 Chr.17: 28616812-28616812 on Build GRCh38
rs748270972 Chr.17: 28616860-28616860 on Build GRCh38
rs749080893 Chr.17: 28616755-28616755 on Build GRCh38
rs747071615 Chr.17: 28616898-28616898 on Build GRCh38
rs925690132 Chr.17: 28616841-28616841 on Build GRCh38
rs776641490 Chr.17: 28616858-28616858 on Build GRCh38
rs369125189 Chr.17: 28616772-28616772 on Build GRCh38
rs768556642 Chr.17: 28616760-28616760 on Build GRCh38
rs754702224 Chr.17: 28616853-28616853 on Build GRCh38
rs748445942 Chr.17: 28616771-28616771 on Build GRCh38
rs988468386 Chr.17: 28616758-28616758 on Build GRCh38
rs771182650 Chr.17: 28616906-28616906 on Build GRCh38
rs375361944 Chr.17: 28616838-28616838 on Build GRCh38
rs772351072 Chr.17: 28616855-28616855 on Build GRCh38
rs867044347 Chr.17: 28616867-28616867 on Build GRCh38
rs201896448 Chr.17: 28616738-28616738 on Build GRCh38
rs578041145 Chr.17: 28616781-28616781 on Build GRCh38
rs866318934 Chr.17: 28616929-28616929 on Build GRCh38
rs375780767 Chr.17: 28616768-28616768 on Build GRCh38
rs376115781 Chr.17: 28616792-28616792 on Build GRCh38
rs749938329 Chr.17: 28616840-28616840 on Build GRCh38
rs750307079 Chr.17: 28616720-28616720 on Build GRCh38
rs779754747 Chr.17: 28616748-28616748 on Build GRCh38
rs777246681 Chr.17: 28616921-28616921 on Build GRCh38
rs761545535 Chr.17: 28616801-28616801 on Build GRCh38
rs753825105 Chr.17: 28616852-28616852 on Build GRCh38
rs778984368 Chr.17: 28616765-28616765 on Build GRCh38
rs755685175 Chr.17: 28616847-28616847 on Build GRCh38
rs531142947 Chr.17: 28616885-28616885 on Build GRCh38
rs772353763 Chr.17: 28616865-28616865 on Build GRCh38
rs1048449244 Chr.17: 28616819-28616819 on Build GRCh38
rs551308663 Chr.17: 28616880-28616880 on Build GRCh38
rs921129618 Chr.17: 28616786-28616786 on Build GRCh38

View Full Product Details