Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32396238-32396467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL2
Entrez Gene ID
56244
Gene Name
butyrophilin like 2
Gene Aliases
BTL-II, BTN7, HSBLMHC1, SS2
UniGene
Hs.534471
Gene Chromosome Location
Chr.6: 32393339-32408879 on Build GRCh38
Transcripts
NM_001304561.1 XM_017011057.1 NR_136245.1

Gene Symbol
LOC101929163
Entrez Gene ID
101929163
Gene Name
uncharacterized LOC101929163
Gene Aliases
-
UniGene
Hs.57692
Gene Chromosome Location
Chr.6: 32255173-32407730 on Build GRCh38
Transcripts
NM_001304561.1 XM_017011057.1 NR_136245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4805725 c.783A>C p.E261D Chr.6: 32396334-32396334 on Build GRCh38
5427670 c.448A>G p.I150V Chr.6: 32396437-32396437 on Build GRCh38
4825764 c.768C>T p.L256L Chr.6: 32396349-32396349 on Build GRCh38
3625431 c.734C>T p.S245F Chr.6: 32396383-32396383 on Build GRCh38
461803 c.791A>G p.Q264R Chr.6: 32396326-32396326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144609103 Chr.6: 32396356-32396356 on Build GRCh38
rs768964504 Chr.6: 32396395-32396395 on Build GRCh38
rs899412781 Chr.6: 32396414-32396414 on Build GRCh38
rs2076527 Chr.6: 32396437-32396437 on Build GRCh38
rs778673342 Chr.6: 32396257-32396257 on Build GRCh38
rs762192832 Chr.6: 32396405-32396405 on Build GRCh38
rs769715885 Chr.6: 32396268-32396268 on Build GRCh38
rs769143380 Chr.6: 32396342-32396343 on Build GRCh38
rs767747252 Chr.6: 32396415-32396418 on Build GRCh38
rs28366191 Chr.6: 32396413-32396413 on Build GRCh38
rs146677249 Chr.6: 32396360-32396360 on Build GRCh38
rs777649107 Chr.6: 32396349-32396349 on Build GRCh38
rs34423804 Chr.6: 32396269-32396269 on Build GRCh38
rs2076528 Chr.6: 32396417-32396417 on Build GRCh38
rs116715584 Chr.6: 32396315-32396315 on Build GRCh38
rs1046295795 Chr.6: 32396404-32396404 on Build GRCh38
rs761256978 Chr.6: 32396305-32396305 on Build GRCh38
rs36110770 Chr.6: 32396267-32396267 on Build GRCh38
rs765808783 Chr.6: 32396309-32396309 on Build GRCh38
rs774754847 Chr.6: 32396392-32396394 on Build GRCh38
rs764919831 Chr.6: 32396326-32396326 on Build GRCh38
rs779234639 Chr.6: 32396399-32396399 on Build GRCh38
rs41355746 Chr.6: 32396275-32396275 on Build GRCh38
rs753412728 Chr.6: 32396313-32396313 on Build GRCh38
rs747994941 Chr.6: 32396258-32396258 on Build GRCh38
rs41333546 Chr.6: 32396280-32396280 on Build GRCh38
rs558755529 Chr.6: 32396284-32396284 on Build GRCh38
rs987041303 Chr.6: 32396298-32396298 on Build GRCh38
rs138515121 Chr.6: 32396299-32396299 on Build GRCh38
rs887038963 Chr.6: 32396366-32396366 on Build GRCh38
rs752459935 Chr.6: 32396345-32396345 on Build GRCh38
rs758333883 Chr.6: 32396266-32396266 on Build GRCh38
rs114609366 Chr.6: 32396371-32396371 on Build GRCh38
rs749425282 Chr.6: 32396387-32396387 on Build GRCh38
rs749458249 Chr.6: 32396273-32396273 on Build GRCh38
rs772265296 Chr.6: 32396416-32396416 on Build GRCh38
rs769178206 Chr.6: 32396348-32396348 on Build GRCh38

View Full Product Details