Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135749943-135750186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNT1
Entrez Gene ID
57582
Gene Name
potassium sodium-activated channel subfamily T member 1
Gene Aliases
EIEE14, ENFL5, KCa4.1, SLACK, Slo2.2, bA100C15.2
UniGene
Hs.104950
Gene Chromosome Location
Chr.9: 135700807-135793147 on Build GRCh38
Transcripts
NM_001272003.1 NM_020822.2 XM_011518877.2 XM_011518878.2 XM_011518879.2 XM_011518880.1 XM_017014931.1 XM_017014932.1 XM_017014933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6874091 c.205G>T p.V69L Chr.9: 135750105-135750105 on Build GRCh38
7373252 c.242G>A p.R81Q Chr.9: 135750142-135750142 on Build GRCh38
5459419 c.216C>T p.Y72Y Chr.9: 135750116-135750116 on Build GRCh38
1330641 c.313T>C p.F105L Chr.9: 135750156-135750156 on Build GRCh38
5459418 c.273C>T p.Y91Y Chr.9: 135750116-135750116 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030934422 Chr.9: 135750037-135750037 on Build GRCh38
rs979723093 Chr.9: 135750030-135750030 on Build GRCh38
rs764463118 Chr.9: 135750062-135750062 on Build GRCh38
rs776233220 Chr.9: 135750117-135750117 on Build GRCh38
rs764504563 Chr.9: 135750131-135750131 on Build GRCh38
rs202149775 Chr.9: 135750077-135750077 on Build GRCh38
rs1007587081 Chr.9: 135749962-135749962 on Build GRCh38
rs769300779 Chr.9: 135750119-135750119 on Build GRCh38
rs762249620 Chr.9: 135750143-135750143 on Build GRCh38
rs746788900 Chr.9: 135750105-135750105 on Build GRCh38
rs183022730 Chr.9: 135750122-135750122 on Build GRCh38
rs748057688 Chr.9: 135750087-135750087 on Build GRCh38
rs901393645 Chr.9: 135750008-135750008 on Build GRCh38
rs558966732 Chr.9: 135750128-135750128 on Build GRCh38
rs952480594 Chr.9: 135750036-135750036 on Build GRCh38
rs767894239 Chr.9: 135750152-135750152 on Build GRCh38
rs988832610 Chr.9: 135750041-135750041 on Build GRCh38
rs544408039 Chr.9: 135750038-135750038 on Build GRCh38
rs1025233216 Chr.9: 135750032-135750032 on Build GRCh38
rs753205513 Chr.9: 135750051-135750051 on Build GRCh38
rs778749299 Chr.9: 135750086-135750086 on Build GRCh38
rs917179079 Chr.9: 135750110-135750110 on Build GRCh38
rs878953565 Chr.9: 135750024-135750024 on Build GRCh38
rs772478192 Chr.9: 135749996-135749996 on Build GRCh38
rs756234145 Chr.9: 135750085-135750085 on Build GRCh38
rs868560158 Chr.9: 135750161-135750161 on Build GRCh38
rs752032951 Chr.9: 135750142-135750142 on Build GRCh38
rs770619151 Chr.9: 135750116-135750116 on Build GRCh38
rs993799473 Chr.9: 135750012-135750012 on Build GRCh38
rs993195922 Chr.9: 135750081-135750081 on Build GRCh38
rs772811461 Chr.9: 135750048-135750064 on Build GRCh38
rs763250731 Chr.9: 135750060-135750060 on Build GRCh38
rs191265575 Chr.9: 135750076-135750076 on Build GRCh38
rs760287830 Chr.9: 135750082-135750083 on Build GRCh38
rs868064538 Chr.9: 135750146-135750146 on Build GRCh38
rs927413004 Chr.9: 135750089-135750089 on Build GRCh38
rs757619543 Chr.9: 135750072-135750072 on Build GRCh38
rs377025248 Chr.9: 135750063-135750063 on Build GRCh38
rs759075818 Chr.9: 135750118-135750118 on Build GRCh38
rs567041377 Chr.9: 135750102-135750102 on Build GRCh38
rs930841801 Chr.9: 135750113-135750113 on Build GRCh38
rs1049280147 Chr.9: 135750115-135750115 on Build GRCh38
rs777687658 Chr.9: 135750104-135750104 on Build GRCh38

View Full Product Details