Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40962321-40962562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT39
Entrez Gene ID
390792
Gene Name
keratin 39
Gene Aliases
CK-39, K39, KA35
UniGene
Hs.28467
Gene Chromosome Location
Chr.17: 40958417-40966892 on Build GRCh38
Transcripts
NM_213656.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3680433 c.796C>A p.Q266K Chr.17: 40962476-40962476 on Build GRCh38
979009 c.719C>A p.S240Y Chr.17: 40962553-40962553 on Build GRCh38
3680432 c.811C>A p.Q271K Chr.17: 40962461-40962461 on Build GRCh38
4696842 c.844G>T p.D282Y Chr.17: 40962428-40962428 on Build GRCh38
979006 c.839G>A p.R280H Chr.17: 40962433-40962433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763029784 Chr.17: 40962474-40962474 on Build GRCh38
rs755276857 Chr.17: 40962406-40962406 on Build GRCh38
rs767555189 Chr.17: 40962372-40962372 on Build GRCh38
rs371490393 Chr.17: 40962521-40962521 on Build GRCh38
rs750490854 Chr.17: 40962377-40962377 on Build GRCh38
rs762983081 Chr.17: 40962370-40962370 on Build GRCh38
rs946506019 Chr.17: 40962450-40962450 on Build GRCh38
rs775415813 Chr.17: 40962463-40962463 on Build GRCh38
rs779395217 Chr.17: 40962408-40962408 on Build GRCh38
rs760842448 Chr.17: 40962380-40962380 on Build GRCh38
rs530492702 Chr.17: 40962401-40962401 on Build GRCh38
rs1010389266 Chr.17: 40962513-40962513 on Build GRCh38
rs1045091853 Chr.17: 40962430-40962430 on Build GRCh38
rs868415451 Chr.17: 40962402-40962402 on Build GRCh38
rs753269338 Chr.17: 40962422-40962422 on Build GRCh38
rs927849189 Chr.17: 40962399-40962399 on Build GRCh38
rs780842043 Chr.17: 40962447-40962447 on Build GRCh38
rs759356520 Chr.17: 40962351-40962351 on Build GRCh38
rs754220746 Chr.17: 40962405-40962405 on Build GRCh38
rs746487960 Chr.17: 40962439-40962439 on Build GRCh38
rs375964584 Chr.17: 40962354-40962354 on Build GRCh38
rs766908475 Chr.17: 40962393-40962394 on Build GRCh38
rs1046822301 Chr.17: 40962350-40962350 on Build GRCh38
rs371874219 Chr.17: 40962495-40962495 on Build GRCh38
rs367785023 Chr.17: 40962482-40962482 on Build GRCh38
rs199857411 Chr.17: 40962498-40962498 on Build GRCh38
rs143164542 Chr.17: 40962456-40962456 on Build GRCh38
rs766581614 Chr.17: 40962391-40962391 on Build GRCh38
rs530818487 Chr.17: 40962433-40962433 on Build GRCh38
rs773193671 Chr.17: 40962491-40962491 on Build GRCh38
rs139147632 Chr.17: 40962434-40962434 on Build GRCh38
rs775503976 Chr.17: 40962361-40962361 on Build GRCh38
rs776766858 Chr.17: 40962514-40962514 on Build GRCh38
rs982057122 Chr.17: 40962398-40962398 on Build GRCh38

View Full Product Details