Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32383741-32384000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSDC1
Entrez Gene ID
55108
Gene Name
BSD domain containing 1
Gene Aliases
-
UniGene
Hs.353454
Gene Chromosome Location
Chr.1: 32364633-32394461 on Build GRCh38
Transcripts
NM_001143888.2 NM_001143889.2 NM_001143890.2 NM_001300958.1 NM_018045.7 NR_125352.1 XM_005270986.4 XM_011541677.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
426106 c.301G>A p.V101M Chr.1: 32383937-32383937 on Build GRCh38
7296922 c.375G>A p.P125P Chr.1: 32383863-32383863 on Build GRCh38
3488178 c.374C>G p.P125R Chr.1: 32383864-32383864 on Build GRCh38
2078028 c.348C>T p.C116C Chr.1: 32383890-32383890 on Build GRCh38
1742100 c.254C>G p.S85* Chr.1: 32383984-32383984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992116645 Chr.1: 32383769-32383769 on Build GRCh38
rs374512700 Chr.1: 32383801-32383801 on Build GRCh38
rs753978607 Chr.1: 32383890-32383890 on Build GRCh38
rs916686046 Chr.1: 32383770-32383770 on Build GRCh38
rs902497133 Chr.1: 32383777-32383777 on Build GRCh38
rs746274192 Chr.1: 32383788-32383788 on Build GRCh38
rs766045450 Chr.1: 32383964-32383964 on Build GRCh38
rs773953698 Chr.1: 32383966-32383967 on Build GRCh38
rs868328633 Chr.1: 32383916-32383916 on Build GRCh38
rs979896089 Chr.1: 32383857-32383857 on Build GRCh38
rs772626969 Chr.1: 32383792-32383792 on Build GRCh38
rs762685415 Chr.1: 32383956-32383956 on Build GRCh38
rs761391756 Chr.1: 32383826-32383826 on Build GRCh38
rs764887301 Chr.1: 32383859-32383859 on Build GRCh38
rs751401179 Chr.1: 32383887-32383887 on Build GRCh38
rs757537648 Chr.1: 32383895-32383895 on Build GRCh38
rs750222067 Chr.1: 32383864-32383864 on Build GRCh38
rs35849659 Chr.1: 32383932-32383932 on Build GRCh38
rs747435307 Chr.1: 32383915-32383915 on Build GRCh38
rs964831323 Chr.1: 32383934-32383934 on Build GRCh38
rs762726759 Chr.1: 32383869-32383869 on Build GRCh38
rs551508208 Chr.1: 32383937-32383937 on Build GRCh38
rs779344928 Chr.1: 32383896-32383896 on Build GRCh38
rs138922587 Chr.1: 32383912-32383912 on Build GRCh38
rs1039369757 Chr.1: 32383966-32383966 on Build GRCh38
rs772785486 Chr.1: 32383941-32383941 on Build GRCh38
rs569122318 Chr.1: 32383911-32383911 on Build GRCh38
rs769435395 Chr.1: 32383940-32383940 on Build GRCh38
rs1052357916 Chr.1: 32383922-32383922 on Build GRCh38
rs371418658 Chr.1: 32383785-32383785 on Build GRCh38
rs763546418 Chr.1: 32383812-32383812 on Build GRCh38
rs746211783 Chr.1: 32383899-32383899 on Build GRCh38
rs776150379 Chr.1: 32383814-32383814 on Build GRCh38
rs777929685 Chr.1: 32383889-32383889 on Build GRCh38
rs751371743 Chr.1: 32383979-32383979 on Build GRCh38
rs760212606 Chr.1: 32383806-32383806 on Build GRCh38
rs759487244 Chr.1: 32383981-32383981 on Build GRCh38
rs752337157 Chr.1: 32383791-32383794 on Build GRCh38
rs368930325 Chr.1: 32383888-32383888 on Build GRCh38
rs769157227 Chr.1: 32383917-32383917 on Build GRCh38
rs766315858 Chr.1: 32383873-32383873 on Build GRCh38

View Full Product Details