Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 150793590-150793861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD99L2
Entrez Gene ID
83692
Gene Name
CD99 molecule like 2
Gene Aliases
CD99B, MIC2L1
UniGene
Hs.522805
Gene Chromosome Location
Chr.X: 150766336-150898816 on Build GRCh38
Transcripts
NM_001184808.1 NM_001242614.1 NM_031462.3 NM_134445.3 NM_134446.3 XM_017029890.1 XM_017029891.1

SNP Detail

SNP ID Chromosome Location
rs375119285 Chr.X: 150793642-150793642 on Build GRCh38
rs782344876 Chr.X: 150793788-150793788 on Build GRCh38
rs781855312 Chr.X: 150793684-150793684 on Build GRCh38
rs199756881 Chr.X: 150793722-150793722 on Build GRCh38
rs961913040 Chr.X: 150793627-150793627 on Build GRCh38
rs369653086 Chr.X: 150793674-150793674 on Build GRCh38
rs782209079 Chr.X: 150793736-150793736 on Build GRCh38
rs374053534 Chr.X: 150793677-150793677 on Build GRCh38
rs782379231 Chr.X: 150793741-150793741 on Build GRCh38
rs782058248 Chr.X: 150793808-150793808 on Build GRCh38
rs782662398 Chr.X: 150793718-150793718 on Build GRCh38
rs782715387 Chr.X: 150793662-150793662 on Build GRCh38
rs377051493 Chr.X: 150793777-150793777 on Build GRCh38
rs782050961 Chr.X: 150793787-150793787 on Build GRCh38
rs782415402 Chr.X: 150793775-150793775 on Build GRCh38
rs1030236879 Chr.X: 150793632-150793632 on Build GRCh38
rs781802970 Chr.X: 150793791-150793791 on Build GRCh38
rs782245993 Chr.X: 150793720-150793720 on Build GRCh38
rs782447228 Chr.X: 150793727-150793727 on Build GRCh38
rs781944496 Chr.X: 150793641-150793641 on Build GRCh38
rs782030612 Chr.X: 150793652-150793652 on Build GRCh38
rs782003527 Chr.X: 150793792-150793792 on Build GRCh38
rs190716813 Chr.X: 150793803-150793803 on Build GRCh38
rs782385956 Chr.X: 150793668-150793668 on Build GRCh38
rs782749743 Chr.X: 150793648-150793648 on Build GRCh38
rs782285952 Chr.X: 150793772-150793772 on Build GRCh38
rs973040342 Chr.X: 150793631-150793631 on Build GRCh38
rs782549830 Chr.X: 150793656-150793656 on Build GRCh38
rs782205280 Chr.X: 150793724-150793724 on Build GRCh38
rs111664885 Chr.X: 150793668-150793668 on Build GRCh38
rs782806358 Chr.X: 150793798-150793798 on Build GRCh38
rs1016629372 Chr.X: 150793623-150793623 on Build GRCh38
rs936099003 Chr.X: 150793814-150793814 on Build GRCh38
rs782625112 Chr.X: 150793730-150793730 on Build GRCh38
rs782568224 Chr.X: 150793702-150793702 on Build GRCh38
rs782690185 Chr.X: 150793790-150793791 on Build GRCh38
rs782610964 Chr.X: 150793681-150793681 on Build GRCh38
rs782637207 Chr.X: 150793749-150793749 on Build GRCh38
rs782115289 Chr.X: 150793796-150793796 on Build GRCh38

View Full Product Details