Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1798555-1798794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL36
Entrez Gene ID
64979
Gene Name
mitochondrial ribosomal protein L36
Gene Aliases
BRIP1, L36mt, MRP-L36, PRPL36, RPMJ
UniGene
Hs.719465
Gene Chromosome Location
Chr.5: 1798385-1801432 on Build GRCh38
Transcripts
NM_032479.3 XM_011514079.2 XM_011514080.2 XM_017009750.1 XM_017009751.1 XM_017009752.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5061004 c.212T>C p.V71A Chr.5: 1798724-1798724 on Build GRCh38
5437151 c.162C>T p.P54P Chr.5: 1798774-1798774 on Build GRCh38
5688027 c.146G>T p.R49L Chr.5: 1798790-1798790 on Build GRCh38
5561199 c.202A>C p.N68H Chr.5: 1798734-1798734 on Build GRCh38
5868325 c.260G>A p.R87Q Chr.5: 1798676-1798676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780020402 Chr.5: 1798621-1798621 on Build GRCh38
rs746490953 Chr.5: 1798624-1798624 on Build GRCh38
rs761929388 Chr.5: 1798721-1798721 on Build GRCh38
rs776083699 Chr.5: 1798629-1798629 on Build GRCh38
rs750696700 Chr.5: 1798714-1798716 on Build GRCh38
rs373722244 Chr.5: 1798654-1798654 on Build GRCh38
rs11543343 Chr.5: 1798767-1798767 on Build GRCh38
rs775801553 Chr.5: 1798693-1798693 on Build GRCh38
rs543103340 Chr.5: 1798653-1798653 on Build GRCh38
rs761269289 Chr.5: 1798695-1798695 on Build GRCh38
rs765336685 Chr.5: 1798616-1798618 on Build GRCh38
rs766211981 Chr.5: 1798732-1798732 on Build GRCh38
rs771966252 Chr.5: 1798619-1798619 on Build GRCh38
rs559763911 Chr.5: 1798762-1798762 on Build GRCh38
rs141000957 Chr.5: 1798678-1798678 on Build GRCh38
rs370132229 Chr.5: 1798612-1798612 on Build GRCh38
rs753959556 Chr.5: 1798649-1798649 on Build GRCh38
rs760603990 Chr.5: 1798648-1798648 on Build GRCh38
rs767999482 Chr.5: 1798671-1798671 on Build GRCh38
rs921331125 Chr.5: 1798581-1798581 on Build GRCh38
rs371776792 Chr.5: 1798696-1798696 on Build GRCh38
rs756275231 Chr.5: 1798602-1798602 on Build GRCh38
rs375979480 Chr.5: 1798595-1798595 on Build GRCh38
rs761545644 Chr.5: 1798632-1798632 on Build GRCh38
rs747647816 Chr.5: 1798676-1798676 on Build GRCh38
rs147308643 Chr.5: 1798666-1798666 on Build GRCh38
rs142443551 Chr.5: 1798723-1798723 on Build GRCh38
rs770832296 Chr.5: 1798773-1798773 on Build GRCh38
rs781036110 Chr.5: 1798744-1798744 on Build GRCh38
rs916804294 Chr.5: 1798692-1798692 on Build GRCh38
rs746972875 Chr.5: 1798669-1798669 on Build GRCh38
rs773367035 Chr.5: 1798774-1798774 on Build GRCh38
rs769184351 Chr.5: 1798770-1798770 on Build GRCh38
rs746962313 Chr.5: 1798747-1798747 on Build GRCh38
rs775956445 Chr.5: 1798766-1798766 on Build GRCh38
rs906178098 Chr.5: 1798633-1798633 on Build GRCh38
rs777500156 Chr.5: 1798748-1798748 on Build GRCh38
rs778703689 Chr.5: 1798614-1798614 on Build GRCh38
rs765436174 Chr.5: 1798722-1798722 on Build GRCh38
rs939916219 Chr.5: 1798620-1798620 on Build GRCh38
rs750200839 Chr.5: 1798658-1798658 on Build GRCh38
rs893553572 Chr.5: 1798644-1798644 on Build GRCh38
rs768124862 Chr.5: 1798645-1798645 on Build GRCh38
rs201364735 Chr.5: 1798593-1798593 on Build GRCh38
rs367987549 Chr.5: 1798683-1798683 on Build GRCh38
rs762260198 Chr.5: 1798614-1798615 on Build GRCh38
rs752061979 Chr.5: 1798592-1798592 on Build GRCh38
rs376804257 Chr.5: 1798590-1798590 on Build GRCh38
rs575765382 Chr.5: 1798627-1798627 on Build GRCh38
rs780782830 Chr.5: 1798674-1798674 on Build GRCh38
rs560726356 Chr.5: 1798617-1798617 on Build GRCh38
rs139443193 Chr.5: 1798771-1798771 on Build GRCh38
rs200540489 Chr.5: 1798668-1798668 on Build GRCh38
rs751409392 Chr.5: 1798741-1798741 on Build GRCh38
rs751251864 Chr.5: 1798751-1798751 on Build GRCh38
rs371641771 Chr.5: 1798626-1798626 on Build GRCh38
rs530341830 Chr.5: 1798772-1798772 on Build GRCh38
rs760027234 Chr.5: 1798639-1798639 on Build GRCh38
rs992860537 Chr.5: 1798680-1798680 on Build GRCh38
rs748960433 Chr.5: 1798761-1798761 on Build GRCh38
rs143983305 Chr.5: 1798625-1798625 on Build GRCh38
rs73024103 Chr.5: 1798586-1798586 on Build GRCh38
rs192549235 Chr.5: 1798677-1798677 on Build GRCh38
rs752801788 Chr.5: 1798598-1798598 on Build GRCh38
rs758373620 Chr.5: 1798638-1798638 on Build GRCh38
rs368590462 Chr.5: 1798609-1798609 on Build GRCh38
rs758125732 Chr.5: 1798726-1798726 on Build GRCh38

View Full Product Details