Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 33372652-33372877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT10
Entrez Gene ID
84750
Gene Name
fucosyltransferase 10
Gene Aliases
FUCTX
UniGene
Hs.458713
Gene Chromosome Location
Chr.8: 33308102-33473492 on Build GRCh38
Transcripts
NM_032664.3 XM_011544676.2 XM_011544677.2 XM_011544678.2 XM_011544679.2 XM_011544680.2 XM_011544681.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6852093 c.1318C>T p.R440* Chr.8: 33372699-33372699 on Build GRCh38
4416878 c.1332T>A p.I444I Chr.8: 33372685-33372685 on Build GRCh38
750586 c.1319G>T p.R440L Chr.8: 33372698-33372698 on Build GRCh38
3899766 c.1294C>T p.R432W Chr.8: 33372723-33372723 on Build GRCh38
5939825 c.1213-7C>T p.? Chr.8: 33372811-33372811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749318349 Chr.8: 33372726-33372726 on Build GRCh38
rs767896583 Chr.8: 33372829-33372829 on Build GRCh38
rs753229916 Chr.8: 33372720-33372720 on Build GRCh38
rs143828192 Chr.8: 33372699-33372699 on Build GRCh38
rs779100469 Chr.8: 33372765-33372765 on Build GRCh38
rs759870361 Chr.8: 33372819-33372819 on Build GRCh38
rs768540999 Chr.8: 33372692-33372692 on Build GRCh38
rs532598203 Chr.8: 33372723-33372723 on Build GRCh38
rs371902825 Chr.8: 33372780-33372780 on Build GRCh38
rs975346416 Chr.8: 33372679-33372679 on Build GRCh38
rs769565637 Chr.8: 33372793-33372793 on Build GRCh38
rs2292747 Chr.8: 33372685-33372685 on Build GRCh38
rs200029664 Chr.8: 33372738-33372738 on Build GRCh38
rs543575527 Chr.8: 33372693-33372693 on Build GRCh38
rs372535165 Chr.8: 33372774-33372774 on Build GRCh38
rs752963856 Chr.8: 33372842-33372842 on Build GRCh38
rs760406391 Chr.8: 33372691-33372691 on Build GRCh38
rs766203380 Chr.8: 33372805-33372805 on Build GRCh38
rs146845613 Chr.8: 33372707-33372707 on Build GRCh38
rs746766311 Chr.8: 33372700-33372700 on Build GRCh38
rs368563609 Chr.8: 33372716-33372716 on Build GRCh38
rs746862509 Chr.8: 33372769-33372769 on Build GRCh38
rs751246405 Chr.8: 33372808-33372808 on Build GRCh38
rs771103436 Chr.8: 33372680-33372680 on Build GRCh38
rs150685638 Chr.8: 33372677-33372677 on Build GRCh38
rs751974018 Chr.8: 33372709-33372709 on Build GRCh38
rs559762514 Chr.8: 33372744-33372744 on Build GRCh38
rs375251022 Chr.8: 33372786-33372786 on Build GRCh38
rs762482815 Chr.8: 33372798-33372798 on Build GRCh38
rs776371439 Chr.8: 33372814-33372814 on Build GRCh38
rs935337312 Chr.8: 33372776-33372776 on Build GRCh38
rs764772859 Chr.8: 33372694-33372694 on Build GRCh38
rs749908771 Chr.8: 33372698-33372698 on Build GRCh38
rs185056122 Chr.8: 33372722-33372722 on Build GRCh38
rs772859460 Chr.8: 33372795-33372795 on Build GRCh38
rs755392414 Chr.8: 33372714-33372714 on Build GRCh38

View Full Product Details