Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169854582-169854831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCYL3
Entrez Gene ID
57147
Gene Name
SCY1 like pseudokinase 3
Gene Aliases
PACE-1, PACE1
UniGene
Hs.435560
Gene Chromosome Location
Chr.1: 169853074-169894267 on Build GRCh38
Transcripts
NM_020423.6 NM_181093.3 XM_006711465.1 XM_011509801.1 XM_011509802.1 XM_011509803.1 XM_017001862.1 XM_017001863.1 XM_017001864.1 XM_017001865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4024992 c.1722G>A p.E574E Chr.1: 169854717-169854717 on Build GRCh38
1683808 c.1547_1548delGG p.W516fs*1 Chr.1: 169854729-169854730 on Build GRCh38
6534023 c.1543T>C p.S515P Chr.1: 169854734-169854734 on Build GRCh38
899371 c.1528G>T p.D510Y Chr.1: 169854749-169854749 on Build GRCh38
5511590 c.1601T>C p.I534T Chr.1: 169854676-169854676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55686910 Chr.1: 169854774-169854774 on Build GRCh38
rs200214247 Chr.1: 169854690-169854690 on Build GRCh38
rs140613939 Chr.1: 169854654-169854654 on Build GRCh38
rs745445136 Chr.1: 169854738-169854738 on Build GRCh38
rs560076666 Chr.1: 169854764-169854764 on Build GRCh38
rs1027596296 Chr.1: 169854661-169854661 on Build GRCh38
rs12143301 Chr.1: 169854649-169854649 on Build GRCh38
rs201470077 Chr.1: 169854637-169854637 on Build GRCh38
rs773017990 Chr.1: 169854738-169854739 on Build GRCh38
rs542836824 Chr.1: 169854711-169854711 on Build GRCh38
rs750523786 Chr.1: 169854797-169854797 on Build GRCh38
rs1034855400 Chr.1: 169854695-169854695 on Build GRCh38
rs769212971 Chr.1: 169854632-169854632 on Build GRCh38
rs934933624 Chr.1: 169854801-169854801 on Build GRCh38
rs149664958 Chr.1: 169854639-169854639 on Build GRCh38
rs769498962 Chr.1: 169854755-169854755 on Build GRCh38
rs368350801 Chr.1: 169854793-169854793 on Build GRCh38
rs775063262 Chr.1: 169854759-169854759 on Build GRCh38
rs533455766 Chr.1: 169854720-169854720 on Build GRCh38
rs188212739 Chr.1: 169854724-169854724 on Build GRCh38
rs761949057 Chr.1: 169854701-169854701 on Build GRCh38
rs760754172 Chr.1: 169854806-169854806 on Build GRCh38
rs948775009 Chr.1: 169854712-169854712 on Build GRCh38
rs201750145 Chr.1: 169854630-169854630 on Build GRCh38
rs770408514 Chr.1: 169854731-169854731 on Build GRCh38
rs201275933 Chr.1: 169854650-169854650 on Build GRCh38
rs780218486 Chr.1: 169854714-169854714 on Build GRCh38
rs146013542 Chr.1: 169854766-169854766 on Build GRCh38
rs754247618 Chr.1: 169854716-169854716 on Build GRCh38
rs769823190 Chr.1: 169854690-169854691 on Build GRCh38
rs776992376 Chr.1: 169854719-169854719 on Build GRCh38
rs149208605 Chr.1: 169854783-169854783 on Build GRCh38
rs750559330 Chr.1: 169854708-169854708 on Build GRCh38
rs780486621 Chr.1: 169854612-169854612 on Build GRCh38
rs950599617 Chr.1: 169854683-169854683 on Build GRCh38
rs967853716 Chr.1: 169854624-169854624 on Build GRCh38
rs780703947 Chr.1: 169854733-169854733 on Build GRCh38
rs562875528 Chr.1: 169854709-169854709 on Build GRCh38
rs764109119 Chr.1: 169854659-169854659 on Build GRCh38
rs767752383 Chr.1: 169854703-169854703 on Build GRCh38

View Full Product Details