Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 33818076-33818347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC3
Entrez Gene ID
5983
Gene Name
replication factor C subunit 3
Gene Aliases
RFC38
UniGene
Hs.115474
Gene Chromosome Location
Chr.13: 33818049-33973946 on Build GRCh38
Transcripts
NM_002915.3 NM_181558.2 XM_011535172.2 XM_011535173.2 XM_011535174.2 XM_011535175.1 XM_017020680.1 XM_017020681.1 XM_017020682.1 XM_017020683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3356392 c.43C>T p.R15W Chr.13: 33818221-33818221 on Build GRCh38
5012658 c.34T>A p.S12T Chr.13: 33818212-33818212 on Build GRCh38
3356391 c.43C>T p.R15W Chr.13: 33818221-33818221 on Build GRCh38
5970971 c.18C>T p.D6D Chr.13: 33818196-33818196 on Build GRCh38
4876806 c.41G>A p.G14E Chr.13: 33818219-33818219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147166048 Chr.13: 33818184-33818184 on Build GRCh38
rs770245095 Chr.13: 33818258-33818258 on Build GRCh38
rs1052945423 Chr.13: 33818102-33818102 on Build GRCh38
rs1038892595 Chr.13: 33818202-33818202 on Build GRCh38
rs775443068 Chr.13: 33818214-33818214 on Build GRCh38
rs994875055 Chr.13: 33818119-33818119 on Build GRCh38
rs760085394 Chr.13: 33818168-33818168 on Build GRCh38
rs766834467 Chr.13: 33818160-33818160 on Build GRCh38
rs934727133 Chr.13: 33818167-33818167 on Build GRCh38
rs762554467 Chr.13: 33818207-33818207 on Build GRCh38
rs770558001 Chr.13: 33818303-33818304 on Build GRCh38
rs369722964 Chr.13: 33818196-33818196 on Build GRCh38
rs745840273 Chr.13: 33818153-33818153 on Build GRCh38
rs763643347 Chr.13: 33818285-33818285 on Build GRCh38
rs765099642 Chr.13: 33818191-33818195 on Build GRCh38
rs772138530 Chr.13: 33818187-33818187 on Build GRCh38
rs763552185 Chr.13: 33818212-33818212 on Build GRCh38
rs769916582 Chr.13: 33818183-33818183 on Build GRCh38
rs916125237 Chr.13: 33818141-33818141 on Build GRCh38
rs773939145 Chr.13: 33818304-33818328 on Build GRCh38
rs754312759 Chr.13: 33818162-33818162 on Build GRCh38
rs781125577 Chr.13: 33818231-33818231 on Build GRCh38
rs927280915 Chr.13: 33818185-33818185 on Build GRCh38
rs992975559 Chr.13: 33818138-33818138 on Build GRCh38
rs149708296 Chr.13: 33818253-33818253 on Build GRCh38
rs774626588 Chr.13: 33818158-33818158 on Build GRCh38
rs750197079 Chr.13: 33818314-33818314 on Build GRCh38
rs748863625 Chr.13: 33818304-33818304 on Build GRCh38
rs949006915 Chr.13: 33818157-33818158 on Build GRCh38
rs996398004 Chr.13: 33818300-33818300 on Build GRCh38
rs994805798 Chr.13: 33818239-33818239 on Build GRCh38
rs772568550 Chr.13: 33818276-33818276 on Build GRCh38
rs777426991 Chr.13: 33818148-33818148 on Build GRCh38
rs200346948 Chr.13: 33818132-33818132 on Build GRCh38
rs1043065500 Chr.13: 33818099-33818099 on Build GRCh38
rs368005710 Chr.13: 33818114-33818114 on Build GRCh38
rs3135532 Chr.13: 33818157-33818157 on Build GRCh38
rs199707785 Chr.13: 33818180-33818180 on Build GRCh38
rs953852866 Chr.13: 33818142-33818142 on Build GRCh38
rs777039237 Chr.13: 33818199-33818199 on Build GRCh38
rs576857924 Chr.13: 33818243-33818243 on Build GRCh38
rs1044355610 Chr.13: 33818108-33818108 on Build GRCh38
rs778435967 Chr.13: 33818133-33818133 on Build GRCh38
rs140507898 Chr.13: 33818219-33818219 on Build GRCh38
rs772163574 Chr.13: 33818244-33818244 on Build GRCh38
rs752271919 Chr.13: 33818174-33818174 on Build GRCh38
rs750246024 Chr.13: 33818233-33818233 on Build GRCh38
rs368066565 Chr.13: 33818135-33818135 on Build GRCh38
rs1004994421 Chr.13: 33818236-33818236 on Build GRCh38
rs372032624 Chr.13: 33818127-33818127 on Build GRCh38
rs570937436 Chr.13: 33818234-33818234 on Build GRCh38
rs752098925 Chr.13: 33818206-33818206 on Build GRCh38
rs41556117 Chr.13: 33818129-33818129 on Build GRCh38
rs752874822 Chr.13: 33818171-33818171 on Build GRCh38
rs200516496 Chr.13: 33818238-33818238 on Build GRCh38
rs955846317 Chr.13: 33818164-33818164 on Build GRCh38
rs777765038 Chr.13: 33818245-33818245 on Build GRCh38
rs771211872 Chr.13: 33818250-33818250 on Build GRCh38
rs371916362 Chr.13: 33818151-33818151 on Build GRCh38
rs947332756 Chr.13: 33818105-33818105 on Build GRCh38
rs960447288 Chr.13: 33818325-33818326 on Build GRCh38
rs776741309 Chr.13: 33818152-33818152 on Build GRCh38
rs3135533 Chr.13: 33818224-33818224 on Build GRCh38
rs1005072895 Chr.13: 33818126-33818126 on Build GRCh38
rs545549965 Chr.13: 33818116-33818116 on Build GRCh38
rs3135531 Chr.13: 33818103-33818103 on Build GRCh38
rs769675392 Chr.13: 33818150-33818150 on Build GRCh38
rs765739264 Chr.13: 33818205-33818205 on Build GRCh38
rs371311426 Chr.13: 33818178-33818178 on Build GRCh38
rs186969174 Chr.13: 33818119-33818119 on Build GRCh38
rs1034708309 Chr.13: 33818318-33818318 on Build GRCh38
rs376501033 Chr.13: 33818194-33818194 on Build GRCh38
rs776916714 Chr.13: 33818251-33818251 on Build GRCh38
rs142273480 Chr.13: 33818100-33818100 on Build GRCh38
rs112921767 Chr.13: 33818159-33818159 on Build GRCh38
rs964658868 Chr.13: 33818118-33818118 on Build GRCh38
rs760034304 Chr.13: 33818203-33818203 on Build GRCh38
rs1027981971 Chr.13: 33818140-33818140 on Build GRCh38
rs776034156 Chr.13: 33818270-33818270 on Build GRCh38
rs781178872 Chr.13: 33818179-33818179 on Build GRCh38
rs1053164827 Chr.13: 33818176-33818176 on Build GRCh38
rs747242551 Chr.13: 33818249-33818249 on Build GRCh38
rs773797037 Chr.13: 33818304-33818304 on Build GRCh38
rs900407577 Chr.13: 33818218-33818218 on Build GRCh38
rs751272132 Chr.13: 33818177-33818177 on Build GRCh38
rs905896678 Chr.13: 33818111-33818111 on Build GRCh38
rs763419199 Chr.13: 33818280-33818280 on Build GRCh38
rs769820964 Chr.13: 33818226-33818227 on Build GRCh38
rs372362680 Chr.13: 33818303-33818303 on Build GRCh38
rs777501925 Chr.13: 33818175-33818175 on Build GRCh38
rs767288692 Chr.13: 33818309-33818309 on Build GRCh38

View Full Product Details