Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18510725-18510972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC23B
Entrez Gene ID
10483
Gene Name
Sec23 homolog B, coat complex II component
Gene Aliases
CDA-II, CDAII, CDAN2, CWS7, HEMPAS
UniGene
Hs.369373
Gene Chromosome Location
Chr.20: 18507544-18561415 on Build GRCh38
Transcripts
NM_001172745.1 NM_001172746.1 NM_006363.4 NM_032985.4 NM_032986.3 XM_017027593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1024904 c.5C>T p.A2V Chr.20: 18510840-18510840 on Build GRCh38
5646803 c.111C>T p.P37P Chr.20: 18510946-18510946 on Build GRCh38
3737642 c.35A>G p.E12G Chr.20: 18510870-18510870 on Build GRCh38
6774136 c.67G>A p.V23M Chr.20: 18510902-18510902 on Build GRCh38
6159126 c.91G>T p.A31S Chr.20: 18510926-18510926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375299261 Chr.20: 18510792-18510792 on Build GRCh38
rs200836567 Chr.20: 18510918-18510918 on Build GRCh38
rs139419360 Chr.20: 18510916-18510916 on Build GRCh38
rs752214160 Chr.20: 18510849-18510849 on Build GRCh38
rs940982122 Chr.20: 18510924-18510924 on Build GRCh38
rs768881779 Chr.20: 18510887-18510887 on Build GRCh38
rs756961943 Chr.20: 18510772-18510772 on Build GRCh38
rs905074313 Chr.20: 18510888-18510888 on Build GRCh38
rs772572358 Chr.20: 18510906-18510906 on Build GRCh38
rs766083236 Chr.20: 18510828-18510828 on Build GRCh38
rs139738956 Chr.20: 18510761-18510761 on Build GRCh38
rs190721221 Chr.20: 18510840-18510840 on Build GRCh38
rs751530265 Chr.20: 18510937-18510937 on Build GRCh38
rs144542988 Chr.20: 18510901-18510901 on Build GRCh38
rs748200634 Chr.20: 18510902-18510902 on Build GRCh38
rs967988810 Chr.20: 18510821-18510821 on Build GRCh38
rs1009563752 Chr.20: 18510773-18510773 on Build GRCh38
rs992663914 Chr.20: 18510853-18510853 on Build GRCh38
rs761082788 Chr.20: 18510912-18510912 on Build GRCh38
rs758435412 Chr.20: 18510851-18510851 on Build GRCh38
rs764194823 Chr.20: 18510931-18510931 on Build GRCh38
rs369908885 Chr.20: 18510852-18510852 on Build GRCh38
rs771260913 Chr.20: 18510913-18510913 on Build GRCh38
rs147576961 Chr.20: 18510938-18510938 on Build GRCh38
rs773579541 Chr.20: 18510819-18510819 on Build GRCh38
rs749449660 Chr.20: 18510879-18510879 on Build GRCh38
rs749901355 Chr.20: 18510824-18510824 on Build GRCh38
rs145142114 Chr.20: 18510923-18510923 on Build GRCh38
rs774972983 Chr.20: 18510813-18510813 on Build GRCh38
rs139616572 Chr.20: 18510860-18510860 on Build GRCh38
rs760962295 Chr.20: 18510826-18510826 on Build GRCh38
rs749451298 Chr.20: 18510793-18510793 on Build GRCh38
rs767961083 Chr.20: 18510816-18510816 on Build GRCh38
rs368833951 Chr.20: 18510799-18510799 on Build GRCh38
rs762490578 Chr.20: 18510928-18510928 on Build GRCh38
rs780821154 Chr.20: 18510943-18510943 on Build GRCh38
rs377153618 Chr.20: 18510814-18510814 on Build GRCh38
rs757114358 Chr.20: 18510870-18510870 on Build GRCh38
rs376365322 Chr.20: 18510839-18510839 on Build GRCh38
rs750181418 Chr.20: 18510948-18510948 on Build GRCh38
rs759146033 Chr.20: 18510917-18510917 on Build GRCh38
rs886056525 Chr.20: 18510920-18510920 on Build GRCh38
rs755624074 Chr.20: 18510948-18510948 on Build GRCh38
rs121918222 Chr.20: 18510875-18510875 on Build GRCh38
rs764619034 Chr.20: 18510841-18510841 on Build GRCh38
rs372080875 Chr.20: 18510876-18510876 on Build GRCh38
rs6045440 Chr.20: 18510909-18510909 on Build GRCh38
rs768608178 Chr.20: 18510806-18510806 on Build GRCh38

View Full Product Details