Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15318960-15319233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEKT3
Entrez Gene ID
64518
Gene Name
tektin 3
Gene Aliases
-
UniGene
Hs.414648
Gene Chromosome Location
Chr.17: 15303811-15343682 on Build GRCh38
Transcripts
NM_031898.2 XM_011523988.2 XM_011523989.1 XM_011523990.2 XM_017024954.1 XM_017024955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4427816 c.712G>T p.D238Y Chr.17: 15319099-15319099 on Build GRCh38
5470614 c.734C>T p.A245V Chr.17: 15319077-15319077 on Build GRCh38
1228903 c.666A>C p.E222D Chr.17: 15319145-15319145 on Build GRCh38
3514474 c.716A>G p.K239R Chr.17: 15319095-15319095 on Build GRCh38
7393451 c.686G>T p.C229F Chr.17: 15319125-15319125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377335456 Chr.17: 15319187-15319187 on Build GRCh38
rs773354751 Chr.17: 15319149-15319149 on Build GRCh38
rs1045092037 Chr.17: 15319107-15319107 on Build GRCh38
rs781295921 Chr.17: 15319127-15319127 on Build GRCh38
rs780415379 Chr.17: 15319022-15319022 on Build GRCh38
rs750760890 Chr.17: 15319090-15319090 on Build GRCh38
rs758735582 Chr.17: 15319094-15319094 on Build GRCh38
rs955097346 Chr.17: 15318997-15318997 on Build GRCh38
rs755889882 Chr.17: 15319085-15319085 on Build GRCh38
rs202131392 Chr.17: 15319072-15319072 on Build GRCh38
rs373341211 Chr.17: 15319057-15319057 on Build GRCh38
rs371141816 Chr.17: 15319086-15319086 on Build GRCh38
rs570443858 Chr.17: 15319053-15319053 on Build GRCh38
rs1022563589 Chr.17: 15318988-15318988 on Build GRCh38
rs939287267 Chr.17: 15319128-15319128 on Build GRCh38
rs770720258 Chr.17: 15319156-15319156 on Build GRCh38
rs767177901 Chr.17: 15319077-15319077 on Build GRCh38
rs768728527 Chr.17: 15319037-15319037 on Build GRCh38
rs769118187 Chr.17: 15319112-15319112 on Build GRCh38
rs987604211 Chr.17: 15319014-15319014 on Build GRCh38
rs201125307 Chr.17: 15319154-15319154 on Build GRCh38
rs112235939 Chr.17: 15319056-15319056 on Build GRCh38
rs567388550 Chr.17: 15319159-15319160 on Build GRCh38
rs151050579 Chr.17: 15319101-15319101 on Build GRCh38
rs760470635 Chr.17: 15319076-15319076 on Build GRCh38
rs375675542 Chr.17: 15319177-15319177 on Build GRCh38
rs775209132 Chr.17: 15319183-15319183 on Build GRCh38
rs761393708 Chr.17: 15319151-15319151 on Build GRCh38
rs759569155 Chr.17: 15319175-15319175 on Build GRCh38
rs769207032 Chr.17: 15319093-15319093 on Build GRCh38
rs940854673 Chr.17: 15319024-15319024 on Build GRCh38
rs748211346 Chr.17: 15319132-15319132 on Build GRCh38
rs961416711 Chr.17: 15319104-15319104 on Build GRCh38
rs376637238 Chr.17: 15319168-15319168 on Build GRCh38
rs927905650 Chr.17: 15319109-15319109 on Build GRCh38
rs920549817 Chr.17: 15319160-15319160 on Build GRCh38
rs750846246 Chr.17: 15319168-15319168 on Build GRCh38
rs775285388 Chr.17: 15319074-15319075 on Build GRCh38
rs770067285 Chr.17: 15319135-15319135 on Build GRCh38
rs776865745 Chr.17: 15319052-15319052 on Build GRCh38
rs376304969 Chr.17: 15319059-15319059 on Build GRCh38
rs369103733 Chr.17: 15319069-15319069 on Build GRCh38
rs753485358 Chr.17: 15319196-15319196 on Build GRCh38
rs377740406 Chr.17: 15319076-15319076 on Build GRCh38
rs185536477 Chr.17: 15319189-15319189 on Build GRCh38

View Full Product Details