Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44275397-44275638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM151B
Entrez Gene ID
441151
Gene Name
transmembrane protein 151B
Gene Aliases
C6orf137, TMEM193, bA444E17.5
UniGene
Hs.632851
Gene Chromosome Location
Chr.6: 44270743-44279445 on Build GRCh38
Transcripts
NM_001137560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4639200 c.762G>A p.A254A Chr.6: 44275588-44275588 on Build GRCh38
5023401 c.768C>T p.F256F Chr.6: 44275594-44275594 on Build GRCh38
3782072 c.636C>T p.C212C Chr.6: 44275462-44275462 on Build GRCh38
4637154 c.649G>A p.V217M Chr.6: 44275475-44275475 on Build GRCh38
3749980 c.591C>T p.R197R Chr.6: 44275417-44275417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913175813 Chr.6: 44275567-44275567 on Build GRCh38
rs565577088 Chr.6: 44275444-44275444 on Build GRCh38
rs747257129 Chr.6: 44275540-44275540 on Build GRCh38
rs781324874 Chr.6: 44275561-44275561 on Build GRCh38
rs12194552 Chr.6: 44275417-44275417 on Build GRCh38
rs377642130 Chr.6: 44275599-44275599 on Build GRCh38
rs778179686 Chr.6: 44275463-44275463 on Build GRCh38
rs921774077 Chr.6: 44275536-44275536 on Build GRCh38
rs911634217 Chr.6: 44275439-44275439 on Build GRCh38
rs535594597 Chr.6: 44275434-44275434 on Build GRCh38
rs138321819 Chr.6: 44275453-44275453 on Build GRCh38
rs769817811 Chr.6: 44275597-44275597 on Build GRCh38
rs987306818 Chr.6: 44275570-44275570 on Build GRCh38
rs745962224 Chr.6: 44275562-44275562 on Build GRCh38
rs935908967 Chr.6: 44275546-44275546 on Build GRCh38
rs553795413 Chr.6: 44275435-44275435 on Build GRCh38
rs912618143 Chr.6: 44275585-44275585 on Build GRCh38
rs974567855 Chr.6: 44275509-44275509 on Build GRCh38
rs771142344 Chr.6: 44275558-44275558 on Build GRCh38
rs557789408 Chr.6: 44275484-44275484 on Build GRCh38

View Full Product Details