Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29306756-29306981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR14J1
Entrez Gene ID
442191
Gene Name
olfactory receptor family 14 subfamily J member 1
Gene Aliases
OR5U1, OR6-25, bA150A6.4, hs6M1-28
UniGene
Hs.632004
Gene Chromosome Location
Chr.6: 29306690-29307655 on Build GRCh38
Transcripts
NM_030946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4406022 c.194C>T p.S65F Chr.6: 29306883-29306883 on Build GRCh38
741565 c.280C>G p.Q94E Chr.6: 29306969-29306969 on Build GRCh38
3623961 c.159C>T p.L53L Chr.6: 29306848-29306848 on Build GRCh38
1286797 c.272C>A p.S91Y Chr.6: 29306961-29306961 on Build GRCh38
3623960 c.150C>T p.D50D Chr.6: 29306839-29306839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772262186 Chr.6: 29306838-29306838 on Build GRCh38
rs750087103 Chr.6: 29306804-29306804 on Build GRCh38
rs776850760 Chr.6: 29306825-29306825 on Build GRCh38
rs947209841 Chr.6: 29306840-29306840 on Build GRCh38
rs748711210 Chr.6: 29306836-29306836 on Build GRCh38
rs147062235 Chr.6: 29306914-29306914 on Build GRCh38
rs766948464 Chr.6: 29306928-29306928 on Build GRCh38
rs538970330 Chr.6: 29306817-29306817 on Build GRCh38
rs769244586 Chr.6: 29306834-29306834 on Build GRCh38
rs147681955 Chr.6: 29306952-29306952 on Build GRCh38
rs763928160 Chr.6: 29306901-29306901 on Build GRCh38
rs775318817 Chr.6: 29306859-29306859 on Build GRCh38
rs780740384 Chr.6: 29306803-29306803 on Build GRCh38
rs1052890824 Chr.6: 29306827-29306827 on Build GRCh38
rs765977363 Chr.6: 29306951-29306951 on Build GRCh38
rs372401214 Chr.6: 29306841-29306841 on Build GRCh38
rs368777812 Chr.6: 29306801-29306801 on Build GRCh38
rs779649648 Chr.6: 29306925-29306925 on Build GRCh38
rs755742835 Chr.6: 29306950-29306950 on Build GRCh38
rs756739940 Chr.6: 29306789-29306789 on Build GRCh38
rs749636074 Chr.6: 29306824-29306824 on Build GRCh38
rs375688699 Chr.6: 29306917-29306917 on Build GRCh38
rs762902645 Chr.6: 29306786-29306786 on Build GRCh38
rs760898355 Chr.6: 29306844-29306844 on Build GRCh38
rs763505322 Chr.6: 29306788-29306788 on Build GRCh38
rs762707548 Chr.6: 29306866-29306866 on Build GRCh38
rs749914573 Chr.6: 29306937-29306937 on Build GRCh38
rs771083965 Chr.6: 29306850-29306850 on Build GRCh38
rs780407143 Chr.6: 29306823-29306823 on Build GRCh38
rs761806559 Chr.6: 29306916-29306916 on Build GRCh38
rs746096141 Chr.6: 29306852-29306852 on Build GRCh38

View Full Product Details