Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42698275-42698500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HHATL
Entrez Gene ID
57467
Gene Name
hedgehog acyltransferase-like
Gene Aliases
C3orf3, GUP1, MBOAT3, MSTP002, OACT3
UniGene
Hs.476041
Gene Chromosome Location
Chr.3: 42692663-42702849 on Build GRCh38
Transcripts
NM_020707.3 NR_027753.1 XM_006713274.2 XM_006713275.2 XM_011533969.2 XM_011533970.2 XM_017006935.1 XM_017006936.1 XM_017006937.1 XM_017006938.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1044771 c.533G>T p.G178V Chr.3: 42698302-42698302 on Build GRCh38
6886254 c.503C>T p.T168I Chr.3: 42698332-42698332 on Build GRCh38
71055 c.500G>A p.G167D Chr.3: 42698335-42698335 on Build GRCh38
5834669 c.507_508insT p.D170fs*1 Chr.3: 42698327-42698328 on Build GRCh38
3594082 c.488G>A p.G163E Chr.3: 42698347-42698347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751439585 Chr.3: 42698425-42698425 on Build GRCh38
rs774480376 Chr.3: 42698354-42698354 on Build GRCh38
rs764753074 Chr.3: 42698387-42698387 on Build GRCh38
rs984622852 Chr.3: 42698334-42698334 on Build GRCh38
rs80118338 Chr.3: 42698459-42698459 on Build GRCh38
rs986031376 Chr.3: 42698472-42698472 on Build GRCh38
rs750063269 Chr.3: 42698402-42698402 on Build GRCh38
rs756986939 Chr.3: 42698457-42698457 on Build GRCh38
rs374432449 Chr.3: 42698366-42698366 on Build GRCh38
rs761274628 Chr.3: 42698381-42698381 on Build GRCh38
rs892652357 Chr.3: 42698443-42698443 on Build GRCh38
rs1011045267 Chr.3: 42698445-42698445 on Build GRCh38
rs758176064 Chr.3: 42698295-42698295 on Build GRCh38
rs142024513 Chr.3: 42698415-42698415 on Build GRCh38
rs1000654399 Chr.3: 42698386-42698386 on Build GRCh38
rs1052665104 Chr.3: 42698416-42698416 on Build GRCh38
rs777846259 Chr.3: 42698340-42698340 on Build GRCh38
rs143853080 Chr.3: 42698293-42698293 on Build GRCh38
rs114515184 Chr.3: 42698458-42698458 on Build GRCh38
rs746978470 Chr.3: 42698323-42698323 on Build GRCh38
rs777269417 Chr.3: 42698391-42698391 on Build GRCh38
rs764781162 Chr.3: 42698394-42698394 on Build GRCh38
rs139658999 Chr.3: 42698349-42698349 on Build GRCh38
rs754844693 Chr.3: 42698336-42698336 on Build GRCh38
rs111524014 Chr.3: 42698333-42698333 on Build GRCh38
rs1029907721 Chr.3: 42698465-42698465 on Build GRCh38
rs763909073 Chr.3: 42698421-42698421 on Build GRCh38
rs749428104 Chr.3: 42698348-42698348 on Build GRCh38
rs758379867 Chr.3: 42698404-42698404 on Build GRCh38
rs746084786 Chr.3: 42698357-42698357 on Build GRCh38
rs1023490876 Chr.3: 42698373-42698373 on Build GRCh38
rs182917333 Chr.3: 42698467-42698467 on Build GRCh38
rs779847922 Chr.3: 42698305-42698305 on Build GRCh38
rs911429256 Chr.3: 42698475-42698475 on Build GRCh38
rs746902049 Chr.3: 42698454-42698454 on Build GRCh38
rs370156172 Chr.3: 42698355-42698355 on Build GRCh38
rs570266146 Chr.3: 42698411-42698411 on Build GRCh38
rs1039703999 Chr.3: 42698401-42698401 on Build GRCh38
rs772348816 Chr.3: 42698358-42698358 on Build GRCh38

View Full Product Details