Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13586976-13587249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF18
Entrez Gene ID
8559
Gene Name
pre-mRNA processing factor 18
Gene Aliases
PRP18, hPrp18
UniGene
Hs.660649
Gene Chromosome Location
Chr.10: 13586811-13645279 on Build GRCh38
Transcripts
NM_003675.3 XM_011519754.2 XM_017016860.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4854468 c.28C>A p.R10R Chr.10: 13587114-13587114 on Build GRCh38
3436106 c.24C>T p.I8I Chr.10: 13587110-13587110 on Build GRCh38
5999092 c.13A>G p.K5E Chr.10: 13587099-13587099 on Build GRCh38
1171675 c.28C>T p.R10W Chr.10: 13587114-13587114 on Build GRCh38
6348147 c.36G>T p.R12R Chr.10: 13587122-13587122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775068941 Chr.10: 13587091-13587091 on Build GRCh38
rs751175281 Chr.10: 13587188-13587191 on Build GRCh38
rs754144647 Chr.10: 13587043-13587043 on Build GRCh38
rs540135860 Chr.10: 13587207-13587207 on Build GRCh38
rs746794249 Chr.10: 13587061-13587061 on Build GRCh38
rs757591037 Chr.10: 13587019-13587019 on Build GRCh38
rs544148729 Chr.10: 13587080-13587080 on Build GRCh38
rs748801723 Chr.10: 13587193-13587193 on Build GRCh38
rs564234998 Chr.10: 13587128-13587128 on Build GRCh38
rs756803382 Chr.10: 13587197-13587198 on Build GRCh38
rs779624841 Chr.10: 13587192-13587192 on Build GRCh38
rs761280908 Chr.10: 13587210-13587210 on Build GRCh38
rs371450941 Chr.10: 13587155-13587155 on Build GRCh38
rs770749874 Chr.10: 13587063-13587063 on Build GRCh38
rs555406056 Chr.10: 13587054-13587054 on Build GRCh38
rs751478825 Chr.10: 13587169-13587169 on Build GRCh38
rs771358630 Chr.10: 13587229-13587229 on Build GRCh38
rs780977840 Chr.10: 13587173-13587173 on Build GRCh38
rs568746423 Chr.10: 13587102-13587102 on Build GRCh38
rs975569618 Chr.10: 13587144-13587144 on Build GRCh38
rs577558671 Chr.10: 13587170-13587170 on Build GRCh38
rs910129271 Chr.10: 13587066-13587066 on Build GRCh38
rs889476141 Chr.10: 13587051-13587051 on Build GRCh38
rs41291325 Chr.10: 13587028-13587028 on Build GRCh38
rs893718940 Chr.10: 13586996-13586996 on Build GRCh38
rs768047945 Chr.10: 13587098-13587098 on Build GRCh38
rs112938116 Chr.10: 13587181-13587181 on Build GRCh38
rs11540074 Chr.10: 13587017-13587017 on Build GRCh38
rs368466889 Chr.10: 13587166-13587166 on Build GRCh38
rs959402354 Chr.10: 13587190-13587191 on Build GRCh38
rs903807056 Chr.10: 13587003-13587003 on Build GRCh38
rs766877479 Chr.10: 13587125-13587125 on Build GRCh38
rs926697735 Chr.10: 13587179-13587179 on Build GRCh38
rs553643811 Chr.10: 13587007-13587007 on Build GRCh38
rs777626278 Chr.10: 13587056-13587056 on Build GRCh38
rs769234953 Chr.10: 13587083-13587083 on Build GRCh38
rs768083160 Chr.10: 13587209-13587209 on Build GRCh38
rs758454254 Chr.10: 13587160-13587160 on Build GRCh38
rs750788775 Chr.10: 13587037-13587037 on Build GRCh38
rs755063591 Chr.10: 13587044-13587044 on Build GRCh38
rs113351847 Chr.10: 13587015-13587015 on Build GRCh38
rs982788170 Chr.10: 13587058-13587058 on Build GRCh38
rs367761319 Chr.10: 13587055-13587055 on Build GRCh38
rs779150942 Chr.10: 13587050-13587050 on Build GRCh38
rs933727632 Chr.10: 13587161-13587161 on Build GRCh38
rs761082227 Chr.10: 13587109-13587109 on Build GRCh38
rs1002034983 Chr.10: 13587005-13587005 on Build GRCh38
rs373644439 Chr.10: 13587042-13587042 on Build GRCh38
rs765625085 Chr.10: 13587147-13587147 on Build GRCh38
rs958777443 Chr.10: 13587026-13587026 on Build GRCh38
rs780732119 Chr.10: 13587198-13587199 on Build GRCh38
rs200277491 Chr.10: 13587081-13587081 on Build GRCh38
rs779336224 Chr.10: 13587010-13587010 on Build GRCh38
rs755259491 Chr.10: 13587134-13587134 on Build GRCh38
rs998355444 Chr.10: 13587033-13587033 on Build GRCh38
rs769430970 Chr.10: 13587175-13587175 on Build GRCh38
rs573470945 Chr.10: 13587027-13587027 on Build GRCh38
rs773633606 Chr.10: 13587108-13587108 on Build GRCh38
rs762435084 Chr.10: 13587094-13587094 on Build GRCh38

View Full Product Details