Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55274387-55274660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPBP1
Entrez Gene ID
23640
Gene Name
HSPA (Hsp70) binding protein 1
Gene Aliases
FES1
UniGene
Hs.53066
Gene Chromosome Location
Chr.19: 55262222-55280383 on Build GRCh38
Transcripts
NM_001130106.1 NM_001297600.1 NM_012267.4 XM_005258700.2 XM_005258701.2 XM_011526691.1 XM_017026543.1 XM_017026544.1 XM_017026545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1190156 c.593G>A p.R198H Chr.19: 55274445-55274445 on Build GRCh38
4127895 c.445C>T p.L149L Chr.19: 55274593-55274593 on Build GRCh38
4081525 c.437T>C p.M146T Chr.19: 55274601-55274601 on Build GRCh38
5776253 c.477G>A p.A159A Chr.19: 55274561-55274561 on Build GRCh38
5495482 c.582G>T p.R194R Chr.19: 55274456-55274456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776670838 Chr.19: 55274544-55274544 on Build GRCh38
rs764503929 Chr.19: 55274562-55274562 on Build GRCh38
rs768939458 Chr.19: 55274446-55274446 on Build GRCh38
rs374807828 Chr.19: 55274458-55274458 on Build GRCh38
rs765682977 Chr.19: 55274504-55274504 on Build GRCh38
rs150595412 Chr.19: 55274570-55274570 on Build GRCh38
rs572927428 Chr.19: 55274563-55274563 on Build GRCh38
rs569499026 Chr.19: 55274534-55274534 on Build GRCh38
rs148458387 Chr.19: 55274537-55274537 on Build GRCh38
rs769861452 Chr.19: 55274440-55274440 on Build GRCh38
rs146963187 Chr.19: 55274513-55274513 on Build GRCh38
rs762143896 Chr.19: 55274457-55274457 on Build GRCh38
rs76377742 Chr.19: 55274559-55274559 on Build GRCh38
rs778790704 Chr.19: 55274529-55274529 on Build GRCh38
rs141743392 Chr.19: 55274526-55274526 on Build GRCh38
rs548533606 Chr.19: 55274547-55274547 on Build GRCh38
rs1042830564 Chr.19: 55274441-55274441 on Build GRCh38
rs755740157 Chr.19: 55274516-55274516 on Build GRCh38
rs894373446 Chr.19: 55274421-55274421 on Build GRCh38
rs141770023 Chr.19: 55274561-55274561 on Build GRCh38
rs186436823 Chr.19: 55274566-55274566 on Build GRCh38
rs771093424 Chr.19: 55274612-55274612 on Build GRCh38
rs756969293 Chr.19: 55274510-55274510 on Build GRCh38
rs780613773 Chr.19: 55274554-55274554 on Build GRCh38
rs751265692 Chr.19: 55274429-55274429 on Build GRCh38
rs778647232 Chr.19: 55274584-55274584 on Build GRCh38
rs754410794 Chr.19: 55274472-55274472 on Build GRCh38
rs866614018 Chr.19: 55274604-55274604 on Build GRCh38
rs776728065 Chr.19: 55274615-55274615 on Build GRCh38
rs150486738 Chr.19: 55274425-55274425 on Build GRCh38
rs754973817 Chr.19: 55274583-55274583 on Build GRCh38
rs890527922 Chr.19: 55274558-55274558 on Build GRCh38
rs1027133433 Chr.19: 55274523-55274523 on Build GRCh38
rs369191820 Chr.19: 55274553-55274553 on Build GRCh38
rs747986218 Chr.19: 55274593-55274593 on Build GRCh38
rs777515038 Chr.19: 55274602-55274602 on Build GRCh38
rs768559005 Chr.19: 55274450-55274450 on Build GRCh38
rs747855866 Chr.19: 55274530-55274530 on Build GRCh38
rs756841813 Chr.19: 55274430-55274430 on Build GRCh38
rs1000876135 Chr.19: 55274465-55274465 on Build GRCh38
rs138905885 Chr.19: 55274525-55274525 on Build GRCh38
rs760005842 Chr.19: 55274483-55274483 on Build GRCh38
rs142792055 Chr.19: 55274508-55274508 on Build GRCh38
rs747281033 Chr.19: 55274609-55274609 on Build GRCh38
rs767671307 Chr.19: 55274407-55274407 on Build GRCh38
rs749222436 Chr.19: 55274446-55274446 on Build GRCh38
rs139452756 Chr.19: 55274594-55274594 on Build GRCh38
rs766830096 Chr.19: 55274422-55274422 on Build GRCh38
rs754216716 Chr.19: 55274423-55274423 on Build GRCh38
rs987976437 Chr.19: 55274599-55274599 on Build GRCh38
rs373339365 Chr.19: 55274571-55274571 on Build GRCh38
rs773370646 Chr.19: 55274462-55274462 on Build GRCh38
rs904897256 Chr.19: 55274469-55274469 on Build GRCh38
rs779704098 Chr.19: 55274521-55274521 on Build GRCh38
rs755395443 Chr.19: 55274424-55274424 on Build GRCh38
rs202077081 Chr.19: 55274444-55274444 on Build GRCh38
rs772341333 Chr.19: 55274533-55274533 on Build GRCh38
rs750447388 Chr.19: 55274408-55274408 on Build GRCh38
rs780780798 Chr.19: 55274434-55274434 on Build GRCh38
rs780645577 Chr.19: 55274512-55274512 on Build GRCh38
rs546713376 Chr.19: 55274445-55274445 on Build GRCh38
rs949111959 Chr.19: 55274636-55274636 on Build GRCh38
rs138127293 Chr.19: 55274435-55274435 on Build GRCh38
rs773982890 Chr.19: 55274456-55274456 on Build GRCh38
rs770350529 Chr.19: 55274630-55274630 on Build GRCh38
rs760678461 Chr.19: 55274470-55274470 on Build GRCh38
rs745872133 Chr.19: 55274625-55274625 on Build GRCh38
rs771040201 Chr.19: 55274543-55274543 on Build GRCh38

View Full Product Details