Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 38688683-38688908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLF3
Entrez Gene ID
51274
Gene Name
Kruppel like factor 3
Gene Aliases
BKLF
UniGene
Hs.298658
Gene Chromosome Location
Chr.4: 38664050-38701508 on Build GRCh38
Transcripts
NM_016531.5 XM_017008279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2846236 c.195G>A p.T65T Chr.4: 38688722-38688722 on Build GRCh38
2846239 c.299C>T p.S100F Chr.4: 38688826-38688826 on Build GRCh38
7296971 c.306A>G p.P102P Chr.4: 38688833-38688833 on Build GRCh38
4653813 c.190C>A p.L64I Chr.4: 38688717-38688717 on Build GRCh38
1055095 c.194C>T p.T65M Chr.4: 38688721-38688721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778961008 Chr.4: 38688732-38688732 on Build GRCh38
rs148740827 Chr.4: 38688707-38688707 on Build GRCh38
rs201591446 Chr.4: 38688887-38688887 on Build GRCh38
rs150818202 Chr.4: 38688748-38688748 on Build GRCh38
rs761766407 Chr.4: 38688836-38688836 on Build GRCh38
rs775966171 Chr.4: 38688747-38688747 on Build GRCh38
rs746543079 Chr.4: 38688740-38688740 on Build GRCh38
rs761153626 Chr.4: 38688793-38688793 on Build GRCh38
rs17854688 Chr.4: 38688874-38688874 on Build GRCh38
rs117461542 Chr.4: 38688766-38688766 on Build GRCh38
rs200138091 Chr.4: 38688867-38688867 on Build GRCh38
rs200788104 Chr.4: 38688829-38688829 on Build GRCh38
rs781573258 Chr.4: 38688704-38688704 on Build GRCh38
rs765180141 Chr.4: 38688764-38688764 on Build GRCh38
rs373502495 Chr.4: 38688722-38688722 on Build GRCh38
rs758507634 Chr.4: 38688735-38688736 on Build GRCh38
rs17857398 Chr.4: 38688781-38688781 on Build GRCh38
rs374745533 Chr.4: 38688864-38688864 on Build GRCh38
rs369311053 Chr.4: 38688760-38688760 on Build GRCh38
rs756476726 Chr.4: 38688782-38688782 on Build GRCh38
rs767805661 Chr.4: 38688759-38688759 on Build GRCh38
rs577513330 Chr.4: 38688803-38688803 on Build GRCh38
rs145278110 Chr.4: 38688827-38688827 on Build GRCh38
rs368422949 Chr.4: 38688802-38688802 on Build GRCh38
rs768992554 Chr.4: 38688808-38688808 on Build GRCh38
rs375331147 Chr.4: 38688744-38688744 on Build GRCh38
rs748083085 Chr.4: 38688733-38688733 on Build GRCh38
rs752385388 Chr.4: 38688858-38688858 on Build GRCh38
rs372969202 Chr.4: 38688757-38688757 on Build GRCh38
rs569064361 Chr.4: 38688749-38688749 on Build GRCh38
rs745581332 Chr.4: 38688801-38688801 on Build GRCh38
rs767752575 Chr.4: 38688839-38688839 on Build GRCh38
rs374324359 Chr.4: 38688789-38688789 on Build GRCh38
rs780750739 Chr.4: 38688879-38688879 on Build GRCh38
rs377495024 Chr.4: 38688743-38688743 on Build GRCh38
rs745389848 Chr.4: 38688882-38688882 on Build GRCh38
rs1002536838 Chr.4: 38688857-38688857 on Build GRCh38
rs955759740 Chr.4: 38688869-38688869 on Build GRCh38
rs766713070 Chr.4: 38688853-38688853 on Build GRCh38
rs754796742 Chr.4: 38688761-38688761 on Build GRCh38
rs562428356 Chr.4: 38688875-38688875 on Build GRCh38
rs548049272 Chr.4: 38688866-38688866 on Build GRCh38
rs780803803 Chr.4: 38688799-38688799 on Build GRCh38
rs201836837 Chr.4: 38688735-38688735 on Build GRCh38
rs553313192 Chr.4: 38688849-38688849 on Build GRCh38
rs375983014 Chr.4: 38688771-38688771 on Build GRCh38
rs771597394 Chr.4: 38688734-38688734 on Build GRCh38
rs531970339 Chr.4: 38688878-38688878 on Build GRCh38
rs371074043 Chr.4: 38688852-38688852 on Build GRCh38
rs766215913 Chr.4: 38688783-38688785 on Build GRCh38

View Full Product Details