Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15564384-15564657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HACL1
Entrez Gene ID
26061
Gene Name
2-hydroxyacyl-CoA lyase 1
Gene Aliases
2-HPCL, HPCL, HPCL2, PHYH2
UniGene
Hs.63290
Gene Chromosome Location
Chr.3: 15560704-15601852 on Build GRCh38
Transcripts
NM_001284413.1 NM_001284415.1 NM_001284416.1 NM_012260.3 NR_104315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5398507 c.1458C>T p.Y486Y Chr.3: 15564610-15564610 on Build GRCh38
3005116 c.1482G>T p.W494C Chr.3: 15564586-15564586 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781144590 Chr.3: 15564493-15564493 on Build GRCh38
rs148867538 Chr.3: 15564477-15564477 on Build GRCh38
rs777097303 Chr.3: 15564616-15564616 on Build GRCh38
rs1017078201 Chr.3: 15564475-15564475 on Build GRCh38
rs1001499099 Chr.3: 15564419-15564419 on Build GRCh38
rs752598573 Chr.3: 15564600-15564600 on Build GRCh38
rs562347592 Chr.3: 15564579-15564579 on Build GRCh38
rs746250245 Chr.3: 15564512-15564512 on Build GRCh38
rs372054175 Chr.3: 15564620-15564620 on Build GRCh38
rs776139566 Chr.3: 15564523-15564523 on Build GRCh38
rs751295536 Chr.3: 15564556-15564556 on Build GRCh38
rs115616776 Chr.3: 15564527-15564527 on Build GRCh38
rs182631548 Chr.3: 15564548-15564548 on Build GRCh38
rs772624464 Chr.3: 15564522-15564522 on Build GRCh38
rs878948616 Chr.3: 15564459-15564459 on Build GRCh38
rs562471532 Chr.3: 15564509-15564509 on Build GRCh38
rs1028346409 Chr.3: 15564547-15564547 on Build GRCh38
rs968424519 Chr.3: 15564408-15564408 on Build GRCh38
rs931349703 Chr.3: 15564553-15564553 on Build GRCh38
rs988343415 Chr.3: 15564468-15564468 on Build GRCh38
rs377588785 Chr.3: 15564510-15564510 on Build GRCh38
rs764062741 Chr.3: 15564534-15564534 on Build GRCh38
rs142650709 Chr.3: 15564436-15564436 on Build GRCh38
rs375070346 Chr.3: 15564545-15564545 on Build GRCh38
rs868008631 Chr.3: 15564586-15564586 on Build GRCh38
rs911808240 Chr.3: 15564617-15564617 on Build GRCh38
rs756802649 Chr.3: 15564624-15564624 on Build GRCh38
rs765496549 Chr.3: 15564555-15564555 on Build GRCh38
rs756737585 Chr.3: 15564441-15564441 on Build GRCh38
rs139470887 Chr.3: 15564562-15564562 on Build GRCh38
rs754796732 Chr.3: 15564540-15564552 on Build GRCh38
rs1004791002 Chr.3: 15564442-15564442 on Build GRCh38
rs147764927 Chr.3: 15564605-15564605 on Build GRCh38

View Full Product Details