Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37801291-37801516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF10
Entrez Gene ID
79269
Gene Name
DDB1 and CUL4 associated factor 10
Gene Aliases
WDR32
UniGene
Hs.118394
Gene Chromosome Location
Chr.9: 37800554-37867669 on Build GRCh38
Transcripts
NM_001286810.1 NM_024345.4 XM_005251577.3 XM_005251578.3 XM_017015128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
753415 c.538G>T p.G180W Chr.9: 37801404-37801404 on Build GRCh38
6423825 c.433T>A p.F145I Chr.9: 37801299-37801299 on Build GRCh38
5771250 c.510C>A p.A170A Chr.9: 37801376-37801376 on Build GRCh38
6640325 c.461C>T p.S154F Chr.9: 37801327-37801327 on Build GRCh38
5078673 c.431_432insG p.N144fs*7 Chr.9: 37801297-37801298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370190180 Chr.9: 37801388-37801388 on Build GRCh38
rs770921332 Chr.9: 37801451-37801451 on Build GRCh38
rs75986742 Chr.9: 37801488-37801488 on Build GRCh38
rs755837354 Chr.9: 37801446-37801446 on Build GRCh38
rs772963081 Chr.9: 37801403-37801403 on Build GRCh38
rs116379925 Chr.9: 37801465-37801465 on Build GRCh38
rs764701106 Chr.9: 37801414-37801414 on Build GRCh38
rs374346144 Chr.9: 37801400-37801400 on Build GRCh38
rs1006933986 Chr.9: 37801345-37801345 on Build GRCh38
rs760907572 Chr.9: 37801411-37801411 on Build GRCh38
rs573368194 Chr.9: 37801336-37801336 on Build GRCh38
rs188933688 Chr.9: 37801370-37801370 on Build GRCh38
rs772358218 Chr.9: 37801391-37801391 on Build GRCh38
rs148430370 Chr.9: 37801385-37801385 on Build GRCh38
rs778807521 Chr.9: 37801358-37801358 on Build GRCh38
rs559843565 Chr.9: 37801470-37801470 on Build GRCh38
rs575473033 Chr.9: 37801386-37801386 on Build GRCh38
rs149691291 Chr.9: 37801350-37801350 on Build GRCh38
rs762510038 Chr.9: 37801416-37801416 on Build GRCh38
rs938344323 Chr.9: 37801441-37801441 on Build GRCh38
rs946984942 Chr.9: 37801489-37801489 on Build GRCh38
rs965376632 Chr.9: 37801361-37801361 on Build GRCh38
rs752571640 Chr.9: 37801443-37801443 on Build GRCh38
rs973684771 Chr.9: 37801430-37801430 on Build GRCh38
rs990783446 Chr.9: 37801481-37801481 on Build GRCh38
rs749174659 Chr.9: 37801449-37801449 on Build GRCh38
rs963227758 Chr.9: 37801426-37801426 on Build GRCh38
rs780969667 Chr.9: 37801314-37801314 on Build GRCh38
rs201208093 Chr.9: 37801377-37801377 on Build GRCh38
rs181616247 Chr.9: 37801436-37801436 on Build GRCh38
rs1018781166 Chr.9: 37801351-37801351 on Build GRCh38
rs765870449 Chr.9: 37801422-37801422 on Build GRCh38
rs74987730 Chr.9: 37801447-37801447 on Build GRCh38
rs948228870 Chr.9: 37801421-37801421 on Build GRCh38
rs145533200 Chr.9: 37801373-37801373 on Build GRCh38
rs751004535 Chr.9: 37801433-37801433 on Build GRCh38
rs777534271 Chr.9: 37801365-37801365 on Build GRCh38
rs977176099 Chr.9: 37801367-37801367 on Build GRCh38
rs574334180 Chr.9: 37801462-37801462 on Build GRCh38
rs779142860 Chr.9: 37801454-37801454 on Build GRCh38

View Full Product Details