Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49417731-49418004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMT
Entrez Gene ID
275
Gene Name
aminomethyltransferase
Gene Aliases
GCE, GCST, GCVT, NKH
UniGene
Hs.102
Gene Chromosome Location
Chr.3: 49416778-49422678 on Build GRCh38
Transcripts
NM_000481.3 NM_001164710.1 NM_001164711.1 NM_001164712.1 NR_028435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1165174 c.878-9A>T p.? Chr.3: 49417982-49417982 on Build GRCh38
7089397 c.1020G>T p.E340D Chr.3: 49417831-49417831 on Build GRCh38
5565459 c.890G>A p.R297Q Chr.3: 49417961-49417961 on Build GRCh38
4158145 c.954G>A p.R318R Chr.3: 49417897-49417897 on Build GRCh38
4754820 c.992G>A p.R331Q Chr.3: 49417859-49417859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370432325 Chr.3: 49417750-49417750 on Build GRCh38
rs773415748 Chr.3: 49417958-49417958 on Build GRCh38
rs763216113 Chr.3: 49417960-49417960 on Build GRCh38
rs376165517 Chr.3: 49417923-49417923 on Build GRCh38
rs747454402 Chr.3: 49417933-49417933 on Build GRCh38
rs777376196 Chr.3: 49417832-49417832 on Build GRCh38
rs376366324 Chr.3: 49417907-49417907 on Build GRCh38
rs11715915 Chr.3: 49417897-49417897 on Build GRCh38
rs373269572 Chr.3: 49417754-49417754 on Build GRCh38
rs1030409681 Chr.3: 49417820-49417820 on Build GRCh38
rs762157837 Chr.3: 49417752-49417752 on Build GRCh38
rs777583902 Chr.3: 49417899-49417899 on Build GRCh38
rs769609020 Chr.3: 49417830-49417830 on Build GRCh38
rs751298230 Chr.3: 49417765-49417765 on Build GRCh38
rs762527539 Chr.3: 49417787-49417788 on Build GRCh38
rs371771461 Chr.3: 49417954-49417954 on Build GRCh38
rs776145886 Chr.3: 49417836-49417836 on Build GRCh38
rs181134220 Chr.3: 49417974-49417974 on Build GRCh38
rs866625610 Chr.3: 49417893-49417893 on Build GRCh38
rs772650939 Chr.3: 49417835-49417835 on Build GRCh38
rs976598152 Chr.3: 49417815-49417816 on Build GRCh38
rs1056820947 Chr.3: 49417965-49417965 on Build GRCh38
rs878933427 Chr.3: 49417775-49417775 on Build GRCh38
rs748831732 Chr.3: 49417834-49417834 on Build GRCh38
rs386833690 Chr.3: 49417964-49417964 on Build GRCh38
rs747733053 Chr.3: 49417808-49417808 on Build GRCh38
rs370016900 Chr.3: 49417843-49417843 on Build GRCh38
rs950546384 Chr.3: 49417862-49417862 on Build GRCh38
rs759349289 Chr.3: 49417967-49417967 on Build GRCh38
rs964737719 Chr.3: 49417773-49417773 on Build GRCh38
rs146114174 Chr.3: 49417901-49417901 on Build GRCh38
rs375727509 Chr.3: 49417800-49417800 on Build GRCh38
rs770048404 Chr.3: 49417961-49417961 on Build GRCh38
rs149457059 Chr.3: 49417890-49417890 on Build GRCh38
rs891522376 Chr.3: 49417968-49417968 on Build GRCh38
rs541011963 Chr.3: 49417860-49417860 on Build GRCh38
rs776823002 Chr.3: 49417952-49417952 on Build GRCh38
rs766422988 Chr.3: 49417962-49417962 on Build GRCh38
rs374539303 Chr.3: 49417851-49417851 on Build GRCh38
rs755780814 Chr.3: 49417898-49417898 on Build GRCh38
rs754720687 Chr.3: 49417769-49417769 on Build GRCh38
rs763020838 Chr.3: 49417880-49417880 on Build GRCh38
rs901123177 Chr.3: 49417811-49417811 on Build GRCh38
rs758281017 Chr.3: 49417755-49417755 on Build GRCh38
rs945551152 Chr.3: 49417978-49417978 on Build GRCh38
rs386833691 Chr.3: 49417879-49417881 on Build GRCh38
rs386833692 Chr.3: 49417868-49417869 on Build GRCh38
rs144971200 Chr.3: 49417953-49417953 on Build GRCh38
rs972670254 Chr.3: 49417972-49417972 on Build GRCh38
rs142183532 Chr.3: 49417910-49417910 on Build GRCh38
rs774367002 Chr.3: 49417980-49417980 on Build GRCh38
rs769249621 Chr.3: 49417945-49417945 on Build GRCh38
rs761911876 Chr.3: 49417865-49417865 on Build GRCh38
rs773215128 Chr.3: 49417879-49417879 on Build GRCh38
rs121964985 Chr.3: 49417892-49417892 on Build GRCh38
rs1009200275 Chr.3: 49417777-49417777 on Build GRCh38
rs766509002 Chr.3: 49417889-49417889 on Build GRCh38
rs766006954 Chr.3: 49417964-49417965 on Build GRCh38
rs765464808 Chr.3: 49417869-49417869 on Build GRCh38
rs779977631 Chr.3: 49417761-49417761 on Build GRCh38

View Full Product Details