Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 20075086-20075359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP61
Entrez Gene ID
26074
Gene Name
cilia and flagella associated protein 61
Gene Aliases
C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4
UniGene
Hs.729168
Gene Chromosome Location
Chr.20: 20052514-20360714 on Build GRCh38
Transcripts
NM_001167816.1 NM_015585.3 XM_005260688.3 XM_005260689.2 XM_005260690.3 XM_005260691.3 XM_005260692.3 XM_005260693.3 XM_005260697.1 XM_011529211.1 XM_011529212.2 XM_011529213.1 XM_017027786.1 XM_017027787.1 XM_017027788.1 XM_017027789.1 XM_017027790.1 XM_017027795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5391663 c.392C>T p.P131L Chr.20: 20075209-20075209 on Build GRCh38
7109122 c.394G>C p.E132Q Chr.20: 20075211-20075211 on Build GRCh38
4917570 c.413T>C p.L138P Chr.20: 20075230-20075230 on Build GRCh38
4917569 c.413T>C p.L138P Chr.20: 20075230-20075230 on Build GRCh38
4135243 c.373A>G p.T125A Chr.20: 20075190-20075190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779187638 Chr.20: 20075232-20075232 on Build GRCh38
rs201930326 Chr.20: 20075262-20075262 on Build GRCh38
rs370093815 Chr.20: 20075298-20075298 on Build GRCh38
rs73291608 Chr.20: 20075156-20075156 on Build GRCh38
rs772687935 Chr.20: 20075172-20075172 on Build GRCh38
rs762438394 Chr.20: 20075275-20075275 on Build GRCh38
rs569599145 Chr.20: 20075310-20075310 on Build GRCh38
rs771228955 Chr.20: 20075247-20075247 on Build GRCh38
rs368302962 Chr.20: 20075268-20075268 on Build GRCh38
rs995524638 Chr.20: 20075276-20075276 on Build GRCh38
rs74173348 Chr.20: 20075166-20075166 on Build GRCh38
rs529375058 Chr.20: 20075208-20075208 on Build GRCh38
rs928859411 Chr.20: 20075259-20075259 on Build GRCh38
rs372098405 Chr.20: 20075234-20075234 on Build GRCh38
rs898414150 Chr.20: 20075332-20075332 on Build GRCh38
rs779099599 Chr.20: 20075161-20075161 on Build GRCh38
rs754041807 Chr.20: 20075218-20075218 on Build GRCh38
rs147357752 Chr.20: 20075189-20075189 on Build GRCh38
rs767278773 Chr.20: 20075211-20075211 on Build GRCh38
rs778435358 Chr.20: 20075245-20075245 on Build GRCh38
rs776151243 Chr.20: 20075193-20075193 on Build GRCh38
rs749471321 Chr.20: 20075141-20075141 on Build GRCh38
rs199811958 Chr.20: 20075180-20075180 on Build GRCh38
rs557905071 Chr.20: 20075126-20075126 on Build GRCh38
rs878966541 Chr.20: 20075165-20075165 on Build GRCh38
rs764748072 Chr.20: 20075197-20075197 on Build GRCh38
rs371147283 Chr.20: 20075223-20075223 on Build GRCh38
rs759882003 Chr.20: 20075295-20075295 on Build GRCh38
rs771266368 Chr.20: 20075192-20075192 on Build GRCh38
rs111238282 Chr.20: 20075143-20075143 on Build GRCh38
rs1016612404 Chr.20: 20075112-20075112 on Build GRCh38
rs137869491 Chr.20: 20075207-20075207 on Build GRCh38
rs367870948 Chr.20: 20075204-20075204 on Build GRCh38
rs759028258 Chr.20: 20075243-20075243 on Build GRCh38
rs777004140 Chr.20: 20075248-20075248 on Build GRCh38
rs750213925 Chr.20: 20075287-20075287 on Build GRCh38
rs767240704 Chr.20: 20075286-20075286 on Build GRCh38
rs755206390 Chr.20: 20075231-20075231 on Build GRCh38
rs749235783 Chr.20: 20075169-20075169 on Build GRCh38
rs774958076 Chr.20: 20075201-20075201 on Build GRCh38
rs200993375 Chr.20: 20075265-20075265 on Build GRCh38
rs914840953 Chr.20: 20075220-20075220 on Build GRCh38
rs765538444 Chr.20: 20075297-20075297 on Build GRCh38
rs371978326 Chr.20: 20075277-20075277 on Build GRCh38
rs759186446 Chr.20: 20075195-20075195 on Build GRCh38
rs530536412 Chr.20: 20075317-20075317 on Build GRCh38
rs116380418 Chr.20: 20075266-20075266 on Build GRCh38
rs200178751 Chr.20: 20075280-20075280 on Build GRCh38

View Full Product Details