Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30555604-30555849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF764
Entrez Gene ID
92595
Gene Name
zinc finger protein 764
Gene Aliases
-
UniGene
Hs.132227
Gene Chromosome Location
Chr.16: 30553764-30558321 on Build GRCh38
Transcripts
NM_001172679.1 NM_033410.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3712010 c.694C>T p.R232C Chr.16: 30555727-30555727 on Build GRCh38
5484013 c.794G>A p.G265D Chr.16: 30555627-30555627 on Build GRCh38
6002196 c.771A>C p.K257N Chr.16: 30555650-30555650 on Build GRCh38
3690925 c.727C>T p.R243C Chr.16: 30555694-30555694 on Build GRCh38
4591561 c.797G>T p.R266L Chr.16: 30555624-30555624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781194698 Chr.16: 30555798-30555798 on Build GRCh38
rs752011232 Chr.16: 30555785-30555785 on Build GRCh38
rs143625356 Chr.16: 30555821-30555821 on Build GRCh38
rs372095147 Chr.16: 30555630-30555630 on Build GRCh38
rs768912068 Chr.16: 30555719-30555719 on Build GRCh38
rs780060820 Chr.16: 30555709-30555709 on Build GRCh38
rs776448500 Chr.16: 30555636-30555636 on Build GRCh38
rs200159165 Chr.16: 30555819-30555819 on Build GRCh38
rs773030364 Chr.16: 30555649-30555649 on Build GRCh38
rs770003508 Chr.16: 30555803-30555803 on Build GRCh38
rs978901229 Chr.16: 30555658-30555658 on Build GRCh38
rs773049269 Chr.16: 30555735-30555735 on Build GRCh38
rs750791064 Chr.16: 30555776-30555776 on Build GRCh38
rs752401106 Chr.16: 30555769-30555769 on Build GRCh38
rs200568262 Chr.16: 30555825-30555825 on Build GRCh38
rs747076946 Chr.16: 30555714-30555714 on Build GRCh38
rs201499017 Chr.16: 30555643-30555643 on Build GRCh38
rs964882330 Chr.16: 30555652-30555652 on Build GRCh38
rs761571463 Chr.16: 30555640-30555640 on Build GRCh38
rs773782971 Chr.16: 30555756-30555756 on Build GRCh38
rs766309839 Chr.16: 30555741-30555741 on Build GRCh38
rs746711799 Chr.16: 30555733-30555737 on Build GRCh38
rs748305347 Chr.16: 30555726-30555726 on Build GRCh38
rs866170437 Chr.16: 30555655-30555655 on Build GRCh38
rs1044297433 Chr.16: 30555742-30555742 on Build GRCh38
rs770584829 Chr.16: 30555816-30555816 on Build GRCh38
rs201700669 Chr.16: 30555637-30555637 on Build GRCh38
rs549155581 Chr.16: 30555638-30555638 on Build GRCh38
rs148272059 Chr.16: 30555674-30555674 on Build GRCh38
rs369017354 Chr.16: 30555635-30555635 on Build GRCh38
rs746374251 Chr.16: 30555696-30555696 on Build GRCh38
rs1018286223 Chr.16: 30555650-30555650 on Build GRCh38
rs993377050 Chr.16: 30555760-30555760 on Build GRCh38
rs374119072 Chr.16: 30555815-30555815 on Build GRCh38
rs546714053 Chr.16: 30555747-30555747 on Build GRCh38
rs748431861 Chr.16: 30555799-30555799 on Build GRCh38
rs139950542 Chr.16: 30555779-30555779 on Build GRCh38
rs1038200505 Chr.16: 30555817-30555817 on Build GRCh38
rs767257887 Chr.16: 30555686-30555686 on Build GRCh38
rs765578626 Chr.16: 30555770-30555770 on Build GRCh38
rs759478755 Chr.16: 30555823-30555823 on Build GRCh38
rs757725711 Chr.16: 30555688-30555688 on Build GRCh38
rs746344442 Chr.16: 30555625-30555625 on Build GRCh38
rs369303755 Chr.16: 30555677-30555677 on Build GRCh38
rs779583327 Chr.16: 30555624-30555624 on Build GRCh38
rs777470491 Chr.16: 30555805-30555805 on Build GRCh38
rs202069419 Chr.16: 30555645-30555645 on Build GRCh38
rs755058088 Chr.16: 30555797-30555797 on Build GRCh38
rs780584927 Chr.16: 30555782-30555782 on Build GRCh38
rs775418144 Chr.16: 30555628-30555629 on Build GRCh38
rs779353000 Chr.16: 30555694-30555694 on Build GRCh38
rs768508449 Chr.16: 30555822-30555823 on Build GRCh38
rs763766442 Chr.16: 30555829-30555829 on Build GRCh38
rs776859295 Chr.16: 30555725-30555725 on Build GRCh38
rs771720292 Chr.16: 30555824-30555824 on Build GRCh38
rs762894395 Chr.16: 30555668-30555668 on Build GRCh38
rs528564302 Chr.16: 30555687-30555687 on Build GRCh38
rs371525188 Chr.16: 30555757-30555757 on Build GRCh38
rs750799468 Chr.16: 30555830-30555830 on Build GRCh38
rs747249153 Chr.16: 30555634-30555634 on Build GRCh38
rs987727218 Chr.16: 30555646-30555646 on Build GRCh38
rs200281411 Chr.16: 30555642-30555642 on Build GRCh38
rs762870245 Chr.16: 30555739-30555739 on Build GRCh38
rs917246100 Chr.16: 30555693-30555693 on Build GRCh38
rs769593557 Chr.16: 30555730-30555730 on Build GRCh38
rs758930910 Chr.16: 30555708-30555708 on Build GRCh38
rs567984501 Chr.16: 30555828-30555828 on Build GRCh38
rs866905203 Chr.16: 30555705-30555705 on Build GRCh38
rs968442709 Chr.16: 30555788-30555788 on Build GRCh38

View Full Product Details