Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74543847-74544072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3B
Entrez Gene ID
10620
Gene Name
AT-rich interaction domain 3B
Gene Aliases
BDP, DRIL2
UniGene
Hs.647642
Gene Chromosome Location
Chr.15: 74541177-74598131 on Build GRCh38
Transcripts
NM_001307939.1 NM_006465.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6142745 c.42A>G p.Q14Q Chr.15: 74543978-74543978 on Build GRCh38
274041 c.80C>T p.A27V Chr.15: 74544016-74544016 on Build GRCh38
434292 c.61G>A p.A21T Chr.15: 74543997-74543997 on Build GRCh38
7235856 c.39A>G p.Q13Q Chr.15: 74543975-74543975 on Build GRCh38
3686104 c.13_15delCAG p.Q15delQ Chr.15: 74543949-74543951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759921078 Chr.15: 74544013-74544013 on Build GRCh38
rs562735419 Chr.15: 74544004-74544004 on Build GRCh38
rs529059345 Chr.15: 74543975-74543976 on Build GRCh38
rs767879408 Chr.15: 74543970-74543975 on Build GRCh38
rs772182871 Chr.15: 74543984-74543984 on Build GRCh38
rs8043032 Chr.15: 74543975-74543975 on Build GRCh38
rs756777862 Chr.15: 74543952-74543952 on Build GRCh38
rs762405564 Chr.15: 74543949-74543951 on Build GRCh38
rs779966496 Chr.15: 74543972-74543972 on Build GRCh38
rs777055147 Chr.15: 74544009-74544009 on Build GRCh38
rs750140517 Chr.15: 74543963-74543963 on Build GRCh38
rs765867777 Chr.15: 74544029-74544029 on Build GRCh38
rs751127718 Chr.15: 74543949-74543949 on Build GRCh38
rs775102156 Chr.15: 74543949-74543954 on Build GRCh38
rs766414838 Chr.15: 74543964-74543975 on Build GRCh38
rs755142232 Chr.15: 74543967-74543975 on Build GRCh38
rs760184502 Chr.15: 74543938-74543938 on Build GRCh38
rs773447867 Chr.15: 74543907-74543907 on Build GRCh38
rs754135975 Chr.15: 74543931-74543931 on Build GRCh38
rs760780683 Chr.15: 74543911-74543911 on Build GRCh38
rs763065575 Chr.15: 74543955-74543956 on Build GRCh38
rs540147284 Chr.15: 74543974-74543975 on Build GRCh38
rs958609373 Chr.15: 74543894-74543894 on Build GRCh38
rs539867129 Chr.15: 74543884-74543884 on Build GRCh38
rs199788980 Chr.15: 74543928-74543928 on Build GRCh38
rs1040677797 Chr.15: 74543901-74543901 on Build GRCh38
rs992709643 Chr.15: 74543926-74543926 on Build GRCh38
rs902077849 Chr.15: 74543916-74543916 on Build GRCh38
rs139869260 Chr.15: 74543985-74543985 on Build GRCh38
rs748260619 Chr.15: 74543978-74543978 on Build GRCh38
rs1049233481 Chr.15: 74543874-74543874 on Build GRCh38
rs760676916 Chr.15: 74543949-74543957 on Build GRCh38
rs756871703 Chr.15: 74543980-74544021 on Build GRCh38
rs751809525 Chr.15: 74543964-74543978 on Build GRCh38
rs765768391 Chr.15: 74543946-74543946 on Build GRCh38
rs776098624 Chr.15: 74544032-74544032 on Build GRCh38
rs772307541 Chr.15: 74543889-74543889 on Build GRCh38
rs763522240 Chr.15: 74544039-74544039 on Build GRCh38
rs770169890 Chr.15: 74543951-74543952 on Build GRCh38
rs201809126 Chr.15: 74543987-74543987 on Build GRCh38
rs748969981 Chr.15: 74543974-74543974 on Build GRCh38
rs755710615 Chr.15: 74543967-74543967 on Build GRCh38
rs148651467 Chr.15: 74543885-74543885 on Build GRCh38
rs574173186 Chr.15: 74543948-74543949 on Build GRCh38
rs752939260 Chr.15: 74543973-74543975 on Build GRCh38

View Full Product Details